Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26209483-26210047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HADHA
Entrez Gene ID
3030
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
Gene Aliases
ECHA, GBP, HADH, LCEH, LCHAD, MTPA, TP-ALPHA
UniGene
Hs.516032
Gene Chromosome Location
Chr.2: 26190635-26244726 on Build GRCh38
Transcripts
NM_000182.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7370633 c.1054A>T p.N352Y Chr.2: 26209811-26209811 on Build GRCh38
5134305 c.987G>A p.E329E Chr.2: 26209878-26209878 on Build GRCh38
7401205 c.996G>A p.M332I Chr.2: 26209869-26209869 on Build GRCh38
1209273 c.980T>G p.F327C Chr.2: 26209885-26209885 on Build GRCh38
442702 c.1005A>G p.E335E Chr.2: 26209860-26209860 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758024143 Chr.2: 26209957-26209957 on Build GRCh38
rs867116152 Chr.2: 26209945-26209945 on Build GRCh38
rs748021991 Chr.2: 26209851-26209851 on Build GRCh38
rs548370377 Chr.2: 26209680-26209680 on Build GRCh38
rs776500880 Chr.2: 26209751-26209751 on Build GRCh38
rs774955995 Chr.2: 26209737-26209737 on Build GRCh38
rs764286763 Chr.2: 26209807-26209807 on Build GRCh38
rs920136564 Chr.2: 26210006-26210006 on Build GRCh38
rs978708941 Chr.2: 26209753-26209753 on Build GRCh38
rs1037050696 Chr.2: 26209975-26209975 on Build GRCh38
rs775800474 Chr.2: 26209884-26209884 on Build GRCh38
rs779337335 Chr.2: 26209732-26209732 on Build GRCh38
rs959747626 Chr.2: 26209662-26209662 on Build GRCh38
rs1037313606 Chr.2: 26209545-26209545 on Build GRCh38
rs963046481 Chr.2: 26209596-26209596 on Build GRCh38
rs753001959 Chr.2: 26209785-26209785 on Build GRCh38
rs761815338 Chr.2: 26209752-26209752 on Build GRCh38
rs888827820 Chr.2: 26209633-26209633 on Build GRCh38
rs778667523 Chr.2: 26209797-26209797 on Build GRCh38
rs746561798 Chr.2: 26209739-26209739 on Build GRCh38
rs765809862 Chr.2: 26209915-26209915 on Build GRCh38
rs372895195 Chr.2: 26209733-26209733 on Build GRCh38
rs754004337 Chr.2: 26209903-26209903 on Build GRCh38
rs746686443 Chr.2: 26209842-26209842 on Build GRCh38
rs758013500 Chr.2: 26209834-26209834 on Build GRCh38
rs866324765 Chr.2: 26209930-26209930 on Build GRCh38
rs780409509 Chr.2: 26209929-26209929 on Build GRCh38
rs1005131255 Chr.2: 26209573-26209573 on Build GRCh38
rs759657537 Chr.2: 26209764-26209764 on Build GRCh38
rs764937403 Chr.2: 26209761-26209761 on Build GRCh38
rs775823974 Chr.2: 26209828-26209828 on Build GRCh38
rs779640035 Chr.2: 26209836-26209836 on Build GRCh38
rs900574180 Chr.2: 26209529-26209529 on Build GRCh38
rs565987699 Chr.2: 26209584-26209584 on Build GRCh38
rs979643559 Chr.2: 26209837-26209837 on Build GRCh38
rs780821979 Chr.2: 26209843-26209843 on Build GRCh38
rs750924075 Chr.2: 26209916-26209916 on Build GRCh38
rs760986303 Chr.2: 26209900-26209900 on Build GRCh38
rs141882266 Chr.2: 26209933-26209933 on Build GRCh38
rs532098288 Chr.2: 26209537-26209537 on Build GRCh38
rs754683307 Chr.2: 26209940-26209940 on Build GRCh38
rs148569363 Chr.2: 26209627-26209627 on Build GRCh38
rs770874291 Chr.2: 26209880-26209880 on Build GRCh38
rs370792678 Chr.2: 26209826-26209826 on Build GRCh38
rs1034118834 Chr.2: 26209639-26209639 on Build GRCh38
rs111383528 Chr.2: 26209504-26209504 on Build GRCh38
rs757845324 Chr.2: 26209816-26209816 on Build GRCh38
rs183045060 Chr.2: 26209763-26209763 on Build GRCh38
rs752285354 Chr.2: 26209870-26209870 on Build GRCh38
rs886601032 Chr.2: 26209993-26209993 on Build GRCh38
rs569842001 Chr.2: 26209736-26209736 on Build GRCh38
rs767895844 Chr.2: 26209766-26209766 on Build GRCh38
rs1052400919 Chr.2: 26209954-26209954 on Build GRCh38
rs754227963 Chr.2: 26209813-26209813 on Build GRCh38
rs757509173 Chr.2: 26209731-26209731 on Build GRCh38
rs931560999 Chr.2: 26210015-26210015 on Build GRCh38
rs1049927183 Chr.2: 26209592-26209592 on Build GRCh38
rs779130894 Chr.2: 26209820-26209820 on Build GRCh38
rs137852772 Chr.2: 26209840-26209840 on Build GRCh38
rs747383193 Chr.2: 26209707-26209707 on Build GRCh38
rs769821069 Chr.2: 26209862-26209862 on Build GRCh38
rs1016374936 Chr.2: 26209591-26209591 on Build GRCh38
rs556112081 Chr.2: 26209563-26209563 on Build GRCh38
rs913062752 Chr.2: 26209677-26209677 on Build GRCh38
rs552325549 Chr.2: 26209578-26209578 on Build GRCh38
rs773118436 Chr.2: 26209931-26209955 on Build GRCh38
rs764262123 Chr.2: 26209901-26209901 on Build GRCh38
rs899391685 Chr.2: 26210017-26210017 on Build GRCh38
rs191384588 Chr.2: 26209669-26209669 on Build GRCh38
rs965714346 Chr.2: 26209738-26209738 on Build GRCh38
rs372635243 Chr.2: 26209740-26209740 on Build GRCh38
rs2229420 Chr.2: 26209793-26209793 on Build GRCh38
rs796051970 Chr.2: 26209883-26209883 on Build GRCh38
rs940818355 Chr.2: 26210013-26210013 on Build GRCh38
rs1057516556 Chr.2: 26209813-26209813 on Build GRCh38
rs772164983 Chr.2: 26209804-26209804 on Build GRCh38
rs758711759 Chr.2: 26209925-26209925 on Build GRCh38
rs762771519 Chr.2: 26209874-26209874 on Build GRCh38

View Full Product Details