Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83292276-83292872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPSE
Entrez Gene ID
10855
Gene Name
heparanase
Gene Aliases
HPA, HPA1, HPR1, HPSE1, HSE1
UniGene
Hs.44227
Gene Chromosome Location
Chr.4: 83292461-83335153 on Build GRCh38
Transcripts
NM_001098540.2 NM_001166498.2 NM_001199830.1 NM_006665.5

SNP Detail

SNP ID Chromosome Location
rs563178146 Chr.4: 83292818-83292818 on Build GRCh38
rs537998280 Chr.4: 83292844-83292844 on Build GRCh38
rs1019805970 Chr.4: 83292318-83292318 on Build GRCh38
rs375652871 Chr.4: 83292626-83292626 on Build GRCh38
rs565189077 Chr.4: 83292435-83292435 on Build GRCh38
rs924128451 Chr.4: 83292746-83292746 on Build GRCh38
rs915748838 Chr.4: 83292614-83292615 on Build GRCh38
rs181542512 Chr.4: 83292807-83292807 on Build GRCh38
rs963081591 Chr.4: 83292573-83292573 on Build GRCh38
rs980913088 Chr.4: 83292682-83292682 on Build GRCh38
rs996610365 Chr.4: 83292404-83292404 on Build GRCh38
rs1053689408 Chr.4: 83292341-83292341 on Build GRCh38
rs75705669 Chr.4: 83292587-83292587 on Build GRCh38
rs935554293 Chr.4: 83292771-83292771 on Build GRCh38
rs4693602 Chr.4: 83292466-83292466 on Build GRCh38
rs528980635 Chr.4: 83292824-83292824 on Build GRCh38
rs117163003 Chr.4: 83292845-83292845 on Build GRCh38
rs959964487 Chr.4: 83292582-83292582 on Build GRCh38
rs948557940 Chr.4: 83292623-83292623 on Build GRCh38
rs1012711635 Chr.4: 83292365-83292365 on Build GRCh38
rs906598531 Chr.4: 83292831-83292831 on Build GRCh38
rs997413648 Chr.4: 83292844-83292844 on Build GRCh38
rs908515593 Chr.4: 83292789-83292789 on Build GRCh38
rs77205506 Chr.4: 83292436-83292436 on Build GRCh38
rs527358442 Chr.4: 83292711-83292711 on Build GRCh38
rs75138284 Chr.4: 83292424-83292424 on Build GRCh38
rs543616061 Chr.4: 83292359-83292359 on Build GRCh38
rs950591196 Chr.4: 83292458-83292458 on Build GRCh38
rs935606922 Chr.4: 83292303-83292303 on Build GRCh38
rs1041277948 Chr.4: 83292808-83292808 on Build GRCh38
rs548775644 Chr.4: 83292718-83292718 on Build GRCh38
rs750130730 Chr.4: 83292778-83292778 on Build GRCh38
rs1045157127 Chr.4: 83292376-83292376 on Build GRCh38
rs761524584 Chr.4: 83292666-83292666 on Build GRCh38
rs1016312587 Chr.4: 83292479-83292479 on Build GRCh38
rs112085278 Chr.4: 83292842-83292842 on Build GRCh38
rs948076344 Chr.4: 83292788-83292788 on Build GRCh38
rs893817982 Chr.4: 83292350-83292350 on Build GRCh38
rs1031572205 Chr.4: 83292579-83292579 on Build GRCh38
rs767289813 Chr.4: 83292719-83292719 on Build GRCh38
rs906648786 Chr.4: 83292432-83292432 on Build GRCh38
rs954488736 Chr.4: 83292694-83292694 on Build GRCh38
rs921696548 Chr.4: 83292677-83292677 on Build GRCh38
rs915187071 Chr.4: 83292780-83292780 on Build GRCh38

View Full Product Details