Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 175701372-175701896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPM6A
Entrez Gene ID
2823
Gene Name
glycoprotein M6A
Gene Aliases
GPM6, M6A
UniGene
Hs.75819
Gene Chromosome Location
Chr.4: 175632937-176002691 on Build GRCh38
Transcripts
NM_001261447.1 NM_001261448.1 NM_005277.4 NM_201591.2 NM_201592.2 NR_048571.1 XM_006714183.1 XM_011531877.1 XM_017008036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6099873 c.105C>A p.C35* Chr.4: 175701667-175701667 on Build GRCh38
138467 c.158G>A p.G53E Chr.4: 175701647-175701647 on Build GRCh38
6099872 c.138C>A p.C46* Chr.4: 175701667-175701667 on Build GRCh38
4687820 c.64C>A p.L22M Chr.4: 175701741-175701741 on Build GRCh38
6999951 c.106G>A p.G36S Chr.4: 175701666-175701666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011341677 Chr.4: 175701867-175701867 on Build GRCh38
rs753808225 Chr.4: 175701706-175701706 on Build GRCh38
rs150616485 Chr.4: 175701693-175701693 on Build GRCh38
rs144181937 Chr.4: 175701655-175701655 on Build GRCh38
rs767339992 Chr.4: 175701553-175701553 on Build GRCh38
rs1047905071 Chr.4: 175701789-175701789 on Build GRCh38
rs1028538880 Chr.4: 175701499-175701499 on Build GRCh38
rs373402926 Chr.4: 175701564-175701564 on Build GRCh38
rs773799350 Chr.4: 175701647-175701647 on Build GRCh38
rs867058508 Chr.4: 175701692-175701692 on Build GRCh38
rs752366922 Chr.4: 175701554-175701554 on Build GRCh38
rs748458884 Chr.4: 175701590-175701590 on Build GRCh38
rs760509346 Chr.4: 175701694-175701694 on Build GRCh38
rs371360656 Chr.4: 175701787-175701787 on Build GRCh38
rs780329543 Chr.4: 175701575-175701575 on Build GRCh38
rs745634928 Chr.4: 175701813-175701814 on Build GRCh38
rs1033371224 Chr.4: 175701419-175701419 on Build GRCh38
rs749839994 Chr.4: 175701612-175701612 on Build GRCh38
rs200605053 Chr.4: 175701818-175701819 on Build GRCh38
rs769049503 Chr.4: 175701750-175701751 on Build GRCh38
rs375144137 Chr.4: 175701599-175701599 on Build GRCh38
rs150260353 Chr.4: 175701760-175701760 on Build GRCh38
rs755987626 Chr.4: 175701558-175701558 on Build GRCh38
rs764074420 Chr.4: 175701561-175701561 on Build GRCh38
rs879717576 Chr.4: 175701788-175701788 on Build GRCh38
rs563782465 Chr.4: 175701767-175701767 on Build GRCh38
rs11545191 Chr.4: 175701727-175701727 on Build GRCh38
rs377524544 Chr.4: 175701566-175701566 on Build GRCh38
rs779487560 Chr.4: 175701795-175701795 on Build GRCh38
rs977978299 Chr.4: 175701541-175701541 on Build GRCh38
rs745314396 Chr.4: 175701799-175701799 on Build GRCh38
rs184260946 Chr.4: 175701437-175701437 on Build GRCh38
rs776925519 Chr.4: 175701811-175701815 on Build GRCh38
rs372684382 Chr.4: 175701792-175701792 on Build GRCh38
rs73002168 Chr.4: 175701394-175701394 on Build GRCh38
rs751531608 Chr.4: 175701754-175701754 on Build GRCh38
rs1006857241 Chr.4: 175701409-175701409 on Build GRCh38
rs756444332 Chr.4: 175701772-175701772 on Build GRCh38
rs759166792 Chr.4: 175701530-175701530 on Build GRCh38
rs757591832 Chr.4: 175701608-175701608 on Build GRCh38
rs774031576 Chr.4: 175701529-175701529 on Build GRCh38
rs778464205 Chr.4: 175701494-175701494 on Build GRCh38
rs770460573 Chr.4: 175701619-175701619 on Build GRCh38
rs747244424 Chr.4: 175701578-175701578 on Build GRCh38
rs368394410 Chr.4: 175701771-175701771 on Build GRCh38
rs761902239 Chr.4: 175701712-175701712 on Build GRCh38
rs771568010 Chr.4: 175701666-175701666 on Build GRCh38
rs754314228 Chr.4: 175701646-175701646 on Build GRCh38
rs754129798 Chr.4: 175701576-175701576 on Build GRCh38
rs762066365 Chr.4: 175701819-175701819 on Build GRCh38
rs781485424 Chr.4: 175701589-175701589 on Build GRCh38
rs755226339 Chr.4: 175701579-175701579 on Build GRCh38
rs778166413 Chr.4: 175701778-175701778 on Build GRCh38
rs771644231 Chr.4: 175701809-175701809 on Build GRCh38
rs764023022 Chr.4: 175701704-175701704 on Build GRCh38
rs1052981819 Chr.4: 175701818-175701818 on Build GRCh38
rs1050067511 Chr.4: 175701614-175701614 on Build GRCh38
rs766479791 Chr.4: 175701739-175701739 on Build GRCh38
rs917702074 Chr.4: 175701471-175701471 on Build GRCh38
rs966412318 Chr.4: 175701472-175701472 on Build GRCh38
rs1006477791 Chr.4: 175701807-175701807 on Build GRCh38
rs370813625 Chr.4: 175701594-175701594 on Build GRCh38
rs530214295 Chr.4: 175701422-175701422 on Build GRCh38

View Full Product Details