Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 153581295-153581885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0922
Entrez Gene ID
23240
Gene Name
KIAA0922
Gene Aliases
TMEM131L
UniGene
Hs.732450
Gene Chromosome Location
Chr.4: 153466346-153637608 on Build GRCh38
Transcripts
NM_001131007.1 NM_015196.3 XM_011531780.1 XM_011531781.1 XM_011531783.1 XM_017007924.1 XM_017007925.1 XM_017007926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5865901 c.892+1G>A p.? Chr.4: 153581561-153581561 on Build GRCh38
1149862 c.828C>T p.N276N Chr.4: 153581496-153581496 on Build GRCh38
732351 c.384C>T p.N128N Chr.4: 153581496-153581496 on Build GRCh38
6325110 c.850C>T p.L284F Chr.4: 153581518-153581518 on Build GRCh38
420158 c.400G>A p.D134N Chr.4: 153581512-153581512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772074628 Chr.4: 153581717-153581718 on Build GRCh38
rs952723506 Chr.4: 153581799-153581799 on Build GRCh38
rs773130619 Chr.4: 153581564-153581564 on Build GRCh38
rs528205076 Chr.4: 153581796-153581796 on Build GRCh38
rs188378248 Chr.4: 153581540-153581540 on Build GRCh38
rs770580253 Chr.4: 153581586-153581586 on Build GRCh38
rs764963956 Chr.4: 153581394-153581395 on Build GRCh38
rs150068649 Chr.4: 153581543-153581543 on Build GRCh38
rs781303404 Chr.4: 153581547-153581547 on Build GRCh38
rs530667363 Chr.4: 153581713-153581713 on Build GRCh38
rs140425563 Chr.4: 153581437-153581437 on Build GRCh38
rs769441882 Chr.4: 153581431-153581431 on Build GRCh38
rs755542243 Chr.4: 153581383-153581383 on Build GRCh38
rs560511385 Chr.4: 153581729-153581729 on Build GRCh38
rs33922047 Chr.4: 153581788-153581788 on Build GRCh38
rs775836469 Chr.4: 153581607-153581607 on Build GRCh38
rs1022903724 Chr.4: 153581748-153581748 on Build GRCh38
rs768014082 Chr.4: 153581475-153581475 on Build GRCh38
rs147278172 Chr.4: 153581340-153581340 on Build GRCh38
rs144382561 Chr.4: 153581689-153581689 on Build GRCh38
rs998562022 Chr.4: 153581664-153581664 on Build GRCh38
rs942796560 Chr.4: 153581717-153581717 on Build GRCh38
rs913457352 Chr.4: 153581657-153581657 on Build GRCh38
rs772719885 Chr.4: 153581432-153581432 on Build GRCh38
rs887005762 Chr.4: 153581761-153581761 on Build GRCh38
rs772517194 Chr.4: 153581605-153581605 on Build GRCh38
rs757558926 Chr.4: 153581533-153581533 on Build GRCh38
rs759075607 Chr.4: 153581599-153581599 on Build GRCh38
rs756455779 Chr.4: 153581523-153581523 on Build GRCh38
rs762394317 Chr.4: 153581358-153581358 on Build GRCh38
rs904366557 Chr.4: 153581325-153581325 on Build GRCh38
rs150888716 Chr.4: 153581414-153581414 on Build GRCh38
rs79899824 Chr.4: 153581589-153581589 on Build GRCh38
rs139705683 Chr.4: 153581554-153581554 on Build GRCh38
rs1043177883 Chr.4: 153581430-153581430 on Build GRCh38
rs748876104 Chr.4: 153581568-153581568 on Build GRCh38
rs764176541 Chr.4: 153581369-153581369 on Build GRCh38
rs1018225161 Chr.4: 153581342-153581342 on Build GRCh38
rs753656380 Chr.4: 153581373-153581380 on Build GRCh38
rs57000514 Chr.4: 153581403-153581403 on Build GRCh38
rs192611840 Chr.4: 153581618-153581618 on Build GRCh38
rs540907335 Chr.4: 153581378-153581378 on Build GRCh38
rs559623594 Chr.4: 153581559-153581559 on Build GRCh38
rs1016583766 Chr.4: 153581859-153581859 on Build GRCh38
rs1025522380 Chr.4: 153581516-153581516 on Build GRCh38
rs747927324 Chr.4: 153581424-153581424 on Build GRCh38
rs779145440 Chr.4: 153581368-153581368 on Build GRCh38
rs370453341 Chr.4: 153581832-153581832 on Build GRCh38
rs965700605 Chr.4: 153581345-153581345 on Build GRCh38
rs997996426 Chr.4: 153581742-153581742 on Build GRCh38
rs757262504 Chr.4: 153581412-153581412 on Build GRCh38
rs767850706 Chr.4: 153581388-153581388 on Build GRCh38
rs548587936 Chr.4: 153581716-153581716 on Build GRCh38
rs753975812 Chr.4: 153581532-153581532 on Build GRCh38
rs143221640 Chr.4: 153581407-153581407 on Build GRCh38
rs370093363 Chr.4: 153581439-153581439 on Build GRCh38
rs1023274731 Chr.4: 153581386-153581386 on Build GRCh38
rs761628964 Chr.4: 153581386-153581388 on Build GRCh38
rs375755658 Chr.4: 153581438-153581438 on Build GRCh38
rs201714497 Chr.4: 153581426-153581426 on Build GRCh38
rs910778003 Chr.4: 153581556-153581556 on Build GRCh38
rs1006964059 Chr.4: 153581469-153581469 on Build GRCh38
rs759974848 Chr.4: 153581468-153581468 on Build GRCh38
rs946027801 Chr.4: 153581422-153581422 on Build GRCh38
rs377398615 Chr.4: 153581346-153581346 on Build GRCh38
rs373560748 Chr.4: 153581524-153581524 on Build GRCh38
rs902978376 Chr.4: 153581646-153581646 on Build GRCh38
rs752034549 Chr.4: 153581381-153581381 on Build GRCh38
rs778514775 Chr.4: 153581792-153581792 on Build GRCh38
rs139356600 Chr.4: 153581503-153581503 on Build GRCh38
rs1047434429 Chr.4: 153581753-153581753 on Build GRCh38
rs372748910 Chr.4: 153581416-153581416 on Build GRCh38
rs143931322 Chr.4: 153581421-153581421 on Build GRCh38
rs998550378 Chr.4: 153581513-153581513 on Build GRCh38
rs537557329 Chr.4: 153581836-153581836 on Build GRCh38
rs1056771162 Chr.4: 153581669-153581669 on Build GRCh38
rs773144453 Chr.4: 153581318-153581318 on Build GRCh38
rs199904834 Chr.4: 153581521-153581521 on Build GRCh38
rs760679068 Chr.4: 153581368-153581369 on Build GRCh38
rs901005237 Chr.4: 153581739-153581739 on Build GRCh38
rs938859369 Chr.4: 153581619-153581619 on Build GRCh38
rs772741560 Chr.4: 153581356-153581356 on Build GRCh38
rs993488802 Chr.4: 153581620-153581620 on Build GRCh38

View Full Product Details