Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 175633498-175634077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPM6A
Entrez Gene ID
2823
Gene Name
glycoprotein M6A
Gene Aliases
GPM6, M6A
UniGene
Hs.75819
Gene Chromosome Location
Chr.4: 175632937-176002691 on Build GRCh38
Transcripts
NM_001261447.1 NM_001261448.1 NM_005277.4 NM_201591.2 NM_201592.2 NR_048571.1 XM_006714183.1 XM_011531877.1 XM_017008036.1

SNP Detail

SNP ID Chromosome Location
rs867830301 Chr.4: 175633576-175633576 on Build GRCh38
rs554308074 Chr.4: 175633610-175633610 on Build GRCh38
rs969784240 Chr.4: 175633944-175633944 on Build GRCh38
rs549730710 Chr.4: 175633969-175633969 on Build GRCh38
rs780223532 Chr.4: 175633589-175633589 on Build GRCh38
rs552821029 Chr.4: 175633676-175633676 on Build GRCh38
rs967518931 Chr.4: 175633701-175633701 on Build GRCh38
rs571986413 Chr.4: 175633769-175633769 on Build GRCh38
rs530024490 Chr.4: 175633877-175633877 on Build GRCh38
rs896053287 Chr.4: 175634025-175634025 on Build GRCh38
rs747296453 Chr.4: 175634035-175634035 on Build GRCh38
rs76400487 Chr.4: 175633697-175633697 on Build GRCh38
rs868292835 Chr.4: 175633555-175633555 on Build GRCh38
rs576157512 Chr.4: 175633600-175633600 on Build GRCh38
rs574481016 Chr.4: 175633628-175633628 on Build GRCh38
rs1015408725 Chr.4: 175633851-175633851 on Build GRCh38
rs527824659 Chr.4: 175633780-175633780 on Build GRCh38
rs911781720 Chr.4: 175633738-175633738 on Build GRCh38
rs953199905 Chr.4: 175633733-175633733 on Build GRCh38
rs1055908354 Chr.4: 175633975-175633975 on Build GRCh38
rs534676057 Chr.4: 175633533-175633533 on Build GRCh38
rs192750884 Chr.4: 175633723-175633723 on Build GRCh38
rs1009638418 Chr.4: 175633719-175633719 on Build GRCh38
rs961533523 Chr.4: 175633895-175633895 on Build GRCh38
rs1026668773 Chr.4: 175633935-175633935 on Build GRCh38
rs1034530436 Chr.4: 175633627-175633627 on Build GRCh38
rs11545194 Chr.4: 175633734-175633734 on Build GRCh38
rs1019932090 Chr.4: 175633696-175633696 on Build GRCh38
rs749399113 Chr.4: 175633602-175633602 on Build GRCh38
rs868261184 Chr.4: 175633577-175633577 on Build GRCh38
rs780796425 Chr.4: 175633930-175633930 on Build GRCh38
rs758547034 Chr.4: 175633858-175633858 on Build GRCh38
rs944621962 Chr.4: 175633747-175633747 on Build GRCh38
rs930051359 Chr.4: 175633905-175633905 on Build GRCh38
rs897218307 Chr.4: 175633543-175633543 on Build GRCh38
rs374999908 Chr.4: 175633992-175633992 on Build GRCh38
rs939608385 Chr.4: 175633561-175633561 on Build GRCh38
rs1002114295 Chr.4: 175633625-175633625 on Build GRCh38
rs545700194 Chr.4: 175633730-175633730 on Build GRCh38
rs547868268 Chr.4: 175633788-175633788 on Build GRCh38
rs555990232 Chr.4: 175633553-175633553 on Build GRCh38
rs150627381 Chr.4: 175633636-175633636 on Build GRCh38
rs926857464 Chr.4: 175633556-175633557 on Build GRCh38
rs1048436471 Chr.4: 175633921-175633921 on Build GRCh38
rs973891162 Chr.4: 175633914-175633914 on Build GRCh38
rs570036741 Chr.4: 175633974-175633974 on Build GRCh38
rs939619020 Chr.4: 175634030-175634030 on Build GRCh38
rs6820412 Chr.4: 175633823-175633823 on Build GRCh38

View Full Product Details