Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 133064766-133065209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA4
Entrez Gene ID
3308
Gene Name
heat shock protein family A (Hsp70) member 4
Gene Aliases
APG-2, HEL-S-5a, HS24/P52, HSPH2, RY, hsp70, hsp70RY
UniGene
Hs.90093
Gene Chromosome Location
Chr.5: 133051970-133105017 on Build GRCh38
Transcripts
NM_002154.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3730459 c.138T>C p.R46R Chr.5: 133065010-133065010 on Build GRCh38
1285442 c.108-1G>T p.? Chr.5: 133064979-133064979 on Build GRCh38
3918631 c.155C>T p.A52V Chr.5: 133065027-133065027 on Build GRCh38
3137754 c.128C>T p.P43L Chr.5: 133065000-133065000 on Build GRCh38
1060803 c.160A>C p.S54R Chr.5: 133065032-133065032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774294120 Chr.5: 133064937-133064937 on Build GRCh38
rs376571322 Chr.5: 133065090-133065090 on Build GRCh38
rs768558135 Chr.5: 133064935-133064935 on Build GRCh38
rs1026304717 Chr.5: 133064814-133064814 on Build GRCh38
rs1051910077 Chr.5: 133064845-133064845 on Build GRCh38
rs959621447 Chr.5: 133065133-133065134 on Build GRCh38
rs777014539 Chr.5: 133064931-133064933 on Build GRCh38
rs1047474152 Chr.5: 133065109-133065109 on Build GRCh38
rs761555302 Chr.5: 133064944-133064944 on Build GRCh38
rs568408922 Chr.5: 133065168-133065168 on Build GRCh38
rs998340591 Chr.5: 133064818-133064818 on Build GRCh38
rs762916839 Chr.5: 133065058-133065059 on Build GRCh38
rs527572952 Chr.5: 133065179-133065179 on Build GRCh38
rs752112757 Chr.5: 133065057-133065057 on Build GRCh38
rs142422741 Chr.5: 133065006-133065006 on Build GRCh38
rs957252333 Chr.5: 133065110-133065110 on Build GRCh38
rs1005556900 Chr.5: 133064899-133064899 on Build GRCh38
rs760369966 Chr.5: 133064974-133064974 on Build GRCh38
rs772939952 Chr.5: 133064973-133064973 on Build GRCh38
rs990076289 Chr.5: 133065113-133065113 on Build GRCh38
rs1003798337 Chr.5: 133065122-133065122 on Build GRCh38
rs1017312411 Chr.5: 133064901-133064901 on Build GRCh38
rs771446035 Chr.5: 133065072-133065072 on Build GRCh38
rs765985570 Chr.5: 133064987-133064987 on Build GRCh38
rs1050017770 Chr.5: 133064820-133064820 on Build GRCh38
rs1023004454 Chr.5: 133065180-133065180 on Build GRCh38
rs759136502 Chr.5: 133065033-133065033 on Build GRCh38
rs767914805 Chr.5: 133065066-133065066 on Build GRCh38
rs545872961 Chr.5: 133064850-133064850 on Build GRCh38
rs375669138 Chr.5: 133065022-133065022 on Build GRCh38
rs901361822 Chr.5: 133064794-133064794 on Build GRCh38
rs771868059 Chr.5: 133064955-133064955 on Build GRCh38
rs776470271 Chr.5: 133065049-133065049 on Build GRCh38
rs185692327 Chr.5: 133064907-133064907 on Build GRCh38
rs77097989 Chr.5: 133064863-133064863 on Build GRCh38
rs757641903 Chr.5: 133065059-133065059 on Build GRCh38
rs1024383054 Chr.5: 133065074-133065074 on Build GRCh38
rs959573535 Chr.5: 133065165-133065165 on Build GRCh38
rs764652689 Chr.5: 133065040-133065040 on Build GRCh38
rs886157986 Chr.5: 133065116-133065116 on Build GRCh38
rs548130305 Chr.5: 133065143-133065143 on Build GRCh38
rs746634878 Chr.5: 133064955-133064955 on Build GRCh38
rs202246039 Chr.5: 133065034-133065034 on Build GRCh38
rs780190708 Chr.5: 133065082-133065082 on Build GRCh38
rs770673981 Chr.5: 133065048-133065049 on Build GRCh38
rs912928619 Chr.5: 133065123-133065123 on Build GRCh38
rs756330749 Chr.5: 133065076-133065076 on Build GRCh38
rs979883787 Chr.5: 133065087-133065087 on Build GRCh38
rs750822270 Chr.5: 133065075-133065075 on Build GRCh38
rs1038706174 Chr.5: 133064919-133064919 on Build GRCh38
rs777116945 Chr.5: 133064908-133064908 on Build GRCh38

View Full Product Details