Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132682389-132682926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4
Entrez Gene ID
3565
Gene Name
interleukin 4
Gene Aliases
BCGF-1, BCGF1, BSF-1, BSF1, IL-4
UniGene
Hs.73917
Gene Chromosome Location
Chr.5: 132673986-132682678 on Build GRCh38
Transcripts
NM_000589.3 NM_172348.2 NR_134248.1

Gene Symbol
LOC105379176
Entrez Gene ID
105379176
Gene Name
uncharacterized LOC105379176
Gene Aliases
-
UniGene
Hs.73917
Gene Chromosome Location
Chr.5: 132679085-132682629 on Build GRCh38
Transcripts
NM_000589.3 NM_172348.2 NR_134248.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3365824 c.440A>G p.K147R Chr.5: 132682565-132682565 on Build GRCh38
3137487 c.436G>C p.E146Q Chr.5: 132682561-132682561 on Build GRCh38
1696197 c.412G>A p.E138K Chr.5: 132682537-132682537 on Build GRCh38
5834935 c.447delA p.K150fs*>4 Chr.5: 132682572-132682572 on Build GRCh38
5560713 c.387C>A p.N129K Chr.5: 132682512-132682512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373826113 Chr.5: 132682506-132682506 on Build GRCh38
rs781537325 Chr.5: 132682485-132682485 on Build GRCh38
rs200479203 Chr.5: 132682497-132682497 on Build GRCh38
rs1013286548 Chr.5: 132682843-132682843 on Build GRCh38
rs181279353 Chr.5: 132682852-132682852 on Build GRCh38
rs1013339964 Chr.5: 132682545-132682545 on Build GRCh38
rs2243290 Chr.5: 132682477-132682477 on Build GRCh38
rs117440960 Chr.5: 132682548-132682548 on Build GRCh38
rs368386975 Chr.5: 132682572-132682572 on Build GRCh38
rs201145789 Chr.5: 132682520-132682520 on Build GRCh38
rs550558636 Chr.5: 132682580-132682580 on Build GRCh38
rs780599702 Chr.5: 132682629-132682629 on Build GRCh38
rs376367511 Chr.5: 132682492-132682492 on Build GRCh38
rs145068648 Chr.5: 132682550-132682550 on Build GRCh38
rs1027254647 Chr.5: 132682804-132682804 on Build GRCh38
rs41403547 Chr.5: 132682422-132682422 on Build GRCh38
rs202123536 Chr.5: 132682613-132682613 on Build GRCh38
rs529787799 Chr.5: 132682772-132682772 on Build GRCh38
rs568198897 Chr.5: 132682888-132682888 on Build GRCh38
rs769754731 Chr.5: 132682899-132682899 on Build GRCh38
rs189727059 Chr.5: 132682847-132682847 on Build GRCh38
rs374355865 Chr.5: 132682530-132682530 on Build GRCh38
rs201594156 Chr.5: 132682502-132682502 on Build GRCh38
rs754478579 Chr.5: 132682468-132682468 on Build GRCh38
rs780509862 Chr.5: 132682517-132682517 on Build GRCh38
rs770754358 Chr.5: 132682616-132682619 on Build GRCh38
rs758376138 Chr.5: 132682689-132682689 on Build GRCh38
rs770304588 Chr.5: 132682513-132682513 on Build GRCh38
rs778479319 Chr.5: 132682474-132682474 on Build GRCh38
rs548288224 Chr.5: 132682831-132682831 on Build GRCh38
rs1040473188 Chr.5: 132682437-132682437 on Build GRCh38
rs2243289 Chr.5: 132682440-132682440 on Build GRCh38
rs940944257 Chr.5: 132682802-132682802 on Build GRCh38
rs922042102 Chr.5: 132682766-132682766 on Build GRCh38
rs199929962 Chr.5: 132682556-132682556 on Build GRCh38
rs896199221 Chr.5: 132682521-132682521 on Build GRCh38
rs200269241 Chr.5: 132682665-132682665 on Build GRCh38
rs956842431 Chr.5: 132682625-132682625 on Build GRCh38
rs969196869 Chr.5: 132682569-132682569 on Build GRCh38
rs1036657033 Chr.5: 132682807-132682807 on Build GRCh38
rs368607250 Chr.5: 132682866-132682866 on Build GRCh38
rs1032591381 Chr.5: 132682889-132682889 on Build GRCh38
rs776686173 Chr.5: 132682551-132682551 on Build GRCh38
rs778103717 Chr.5: 132682759-132682759 on Build GRCh38
rs201210927 Chr.5: 132682610-132682610 on Build GRCh38
rs748407182 Chr.5: 132682529-132682529 on Build GRCh38
rs913231829 Chr.5: 132682681-132682681 on Build GRCh38
rs565969000 Chr.5: 132682795-132682795 on Build GRCh38
rs757625990 Chr.5: 132682481-132682481 on Build GRCh38
rs201704672 Chr.5: 132682695-132682695 on Build GRCh38
rs371841361 Chr.5: 132682581-132682581 on Build GRCh38
rs202229913 Chr.5: 132682669-132682669 on Build GRCh38
rs149147538 Chr.5: 132682574-132682574 on Build GRCh38
rs376073510 Chr.5: 132682603-132682603 on Build GRCh38
rs200011844 Chr.5: 132682518-132682518 on Build GRCh38
rs753428723 Chr.5: 132682457-132682457 on Build GRCh38
rs201253645 Chr.5: 132682668-132682668 on Build GRCh38
rs773042624 Chr.5: 132682816-132682816 on Build GRCh38
rs774507412 Chr.5: 132682522-132682522 on Build GRCh38
rs1000670746 Chr.5: 132682872-132682872 on Build GRCh38
rs142651732 Chr.5: 132682785-132682785 on Build GRCh38
rs55857563 Chr.5: 132682447-132682447 on Build GRCh38
rs988787472 Chr.5: 132682666-132682666 on Build GRCh38
rs200428290 Chr.5: 132682692-132682692 on Build GRCh38
rs1001125550 Chr.5: 132682583-132682583 on Build GRCh38
rs113603154 Chr.5: 132682832-132682832 on Build GRCh38
rs530521308 Chr.5: 132682897-132682897 on Build GRCh38

View Full Product Details