Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125276952-125277486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDDC2
Entrez Gene ID
51020
Gene Name
HD domain containing 2
Gene Aliases
C6orf74, CGI-130, NS5ATP2, dJ167O5.2
UniGene
Hs.32826
Gene Chromosome Location
Chr.6: 125274934-125302141 on Build GRCh38
Transcripts
NM_016063.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1487194 c.495A>T p.Q165H Chr.6: 125277124-125277124 on Build GRCh38
7187486 c.452C>T p.A151V Chr.6: 125277167-125277167 on Build GRCh38
1440247 c.514G>T p.A172S Chr.6: 125277105-125277105 on Build GRCh38
6172561 c.391C>A p.Q131K Chr.6: 125277228-125277228 on Build GRCh38
1745227 c.491_492delTG p.L164fs*5 Chr.6: 125277127-125277128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756671571 Chr.6: 125277102-125277102 on Build GRCh38
rs1044693194 Chr.6: 125277307-125277307 on Build GRCh38
rs772537178 Chr.6: 125277123-125277123 on Build GRCh38
rs751352650 Chr.6: 125277224-125277224 on Build GRCh38
rs761430840 Chr.6: 125277057-125277057 on Build GRCh38
rs935121341 Chr.6: 125277235-125277235 on Build GRCh38
rs750072372 Chr.6: 125277281-125277282 on Build GRCh38
rs544448288 Chr.6: 125277103-125277103 on Build GRCh38
rs764759757 Chr.6: 125277074-125277074 on Build GRCh38
rs749295350 Chr.6: 125277107-125277107 on Build GRCh38
rs375169737 Chr.6: 125277212-125277212 on Build GRCh38
rs527742405 Chr.6: 125277348-125277348 on Build GRCh38
rs776665742 Chr.6: 125277196-125277196 on Build GRCh38
rs369638071 Chr.6: 125277226-125277226 on Build GRCh38
rs909966219 Chr.6: 125277137-125277137 on Build GRCh38
rs748123187 Chr.6: 125277292-125277292 on Build GRCh38
rs948562492 Chr.6: 125277243-125277247 on Build GRCh38
rs2243391 Chr.6: 125277054-125277054 on Build GRCh38
rs771191986 Chr.6: 125277395-125277395 on Build GRCh38
rs768451024 Chr.6: 125277175-125277175 on Build GRCh38
rs376453479 Chr.6: 125277372-125277372 on Build GRCh38
rs191484950 Chr.6: 125277319-125277319 on Build GRCh38
rs535827225 Chr.6: 125277015-125277015 on Build GRCh38
rs972729320 Chr.6: 125277186-125277186 on Build GRCh38
rs750368831 Chr.6: 125277320-125277320 on Build GRCh38
rs992131925 Chr.6: 125277386-125277386 on Build GRCh38
rs895691137 Chr.6: 125277240-125277240 on Build GRCh38
rs914601908 Chr.6: 125277144-125277144 on Build GRCh38
rs1023132489 Chr.6: 125277008-125277008 on Build GRCh38
rs73581639 Chr.6: 125277435-125277435 on Build GRCh38
rs918621867 Chr.6: 125277356-125277356 on Build GRCh38
rs750561763 Chr.6: 125277269-125277269 on Build GRCh38
rs766188818 Chr.6: 125277232-125277232 on Build GRCh38
rs34154907 Chr.6: 125277053-125277054 on Build GRCh38
rs369347497 Chr.6: 125277098-125277098 on Build GRCh38
rs546384719 Chr.6: 125277169-125277169 on Build GRCh38
rs979315084 Chr.6: 125277189-125277189 on Build GRCh38
rs1019811503 Chr.6: 125277426-125277426 on Build GRCh38
rs772670912 Chr.6: 125277218-125277218 on Build GRCh38
rs948634791 Chr.6: 125277409-125277409 on Build GRCh38
rs962794559 Chr.6: 125277331-125277331 on Build GRCh38
rs78731112 Chr.6: 125276996-125276996 on Build GRCh38
rs746770487 Chr.6: 125277276-125277276 on Build GRCh38
rs370167765 Chr.6: 125277241-125277241 on Build GRCh38
rs1026280821 Chr.6: 125277398-125277398 on Build GRCh38
rs1030323316 Chr.6: 125277315-125277315 on Build GRCh38
rs998471152 Chr.6: 125277281-125277281 on Build GRCh38
rs772348549 Chr.6: 125277126-125277126 on Build GRCh38
rs761182166 Chr.6: 125277206-125277206 on Build GRCh38
rs375928144 Chr.6: 125277260-125277260 on Build GRCh38
rs939354643 Chr.6: 125277385-125277385 on Build GRCh38
rs371610845 Chr.6: 125277116-125277116 on Build GRCh38
rs895627700 Chr.6: 125276973-125276973 on Build GRCh38
rs775252609 Chr.6: 125277162-125277162 on Build GRCh38
rs780295164 Chr.6: 125277275-125277275 on Build GRCh38
rs1017081514 Chr.6: 125277327-125277327 on Build GRCh38
rs753085694 Chr.6: 125277234-125277234 on Build GRCh38
rs941422376 Chr.6: 125277145-125277145 on Build GRCh38
rs537930760 Chr.6: 125277029-125277029 on Build GRCh38
rs796473024 Chr.6: 125277351-125277351 on Build GRCh38
rs1023290692 Chr.6: 125276986-125276986 on Build GRCh38
rs11537939 Chr.6: 125277215-125277215 on Build GRCh38
rs377730880 Chr.6: 125277293-125277293 on Build GRCh38
rs1008805619 Chr.6: 125277400-125277400 on Build GRCh38
rs891452070 Chr.6: 125277277-125277277 on Build GRCh38
rs542805186 Chr.6: 125277238-125277238 on Build GRCh38
rs2243392 Chr.6: 125277053-125277053 on Build GRCh38
rs757388724 Chr.6: 125277109-125277109 on Build GRCh38
rs755339227 Chr.6: 125277089-125277089 on Build GRCh38
rs887368582 Chr.6: 125277399-125277399 on Build GRCh38
rs780948694 Chr.6: 125277198-125277211 on Build GRCh38
rs904335942 Chr.6: 125277430-125277430 on Build GRCh38
rs768522136 Chr.6: 125277288-125277288 on Build GRCh38
rs757224074 Chr.6: 125277261-125277261 on Build GRCh38
rs186920234 Chr.6: 125277055-125277055 on Build GRCh38
rs780936045 Chr.6: 125277290-125277290 on Build GRCh38
rs560540561 Chr.6: 125277340-125277340 on Build GRCh38
rs746856033 Chr.6: 125277174-125277174 on Build GRCh38
rs779677796 Chr.6: 125276990-125276990 on Build GRCh38
rs369952602 Chr.6: 125277268-125277268 on Build GRCh38
rs2243388 Chr.6: 125277262-125277262 on Build GRCh38
rs139764635 Chr.6: 125277079-125277079 on Build GRCh38

View Full Product Details