Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132708115-132708447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5A
Entrez Gene ID
3632
Gene Name
inositol polyphosphate-5-phosphatase A
Gene Aliases
5PTASE
UniGene
Hs.523360
Gene Chromosome Location
Chr.10: 132537779-132783480 on Build GRCh38
Transcripts
NM_001321042.1 NM_005539.4 XM_017016204.1 XM_017016205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3966966 c.501C>A p.I167I Chr.10: 132708339-132708339 on Build GRCh38
4761659 c.475-2A>C p.? Chr.10: 132708311-132708311 on Build GRCh38
6249186 c.509G>T p.R170M Chr.10: 132708347-132708347 on Build GRCh38
3435853 c.507G>A p.T169T Chr.10: 132708345-132708345 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775493983 Chr.10: 132708304-132708305 on Build GRCh38
rs1035404117 Chr.10: 132708164-132708164 on Build GRCh38
rs371118880 Chr.10: 132708395-132708395 on Build GRCh38
rs756699232 Chr.10: 132708422-132708423 on Build GRCh38
rs781647578 Chr.10: 132708412-132708413 on Build GRCh38
rs898481497 Chr.10: 132708276-132708276 on Build GRCh38
rs758206139 Chr.10: 132708422-132708422 on Build GRCh38
rs986851488 Chr.10: 132708232-132708232 on Build GRCh38
rs775635584 Chr.10: 132708286-132708286 on Build GRCh38
rs765917658 Chr.10: 132708411-132708411 on Build GRCh38
rs35067177 Chr.10: 132708242-132708242 on Build GRCh38
rs767997562 Chr.10: 132708368-132708368 on Build GRCh38
rs777872170 Chr.10: 132708428-132708428 on Build GRCh38
rs376142223 Chr.10: 132708297-132708297 on Build GRCh38
rs549435844 Chr.10: 132708345-132708345 on Build GRCh38
rs371766077 Chr.10: 132708377-132708377 on Build GRCh38
rs563901854 Chr.10: 132708180-132708180 on Build GRCh38
rs766043988 Chr.10: 132708370-132708370 on Build GRCh38
rs745936272 Chr.10: 132708332-132708332 on Build GRCh38
rs565166176 Chr.10: 132708245-132708245 on Build GRCh38
rs1021319694 Chr.10: 132708392-132708392 on Build GRCh38
rs755820164 Chr.10: 132708280-132708280 on Build GRCh38
rs772101826 Chr.10: 132708285-132708285 on Build GRCh38
rs545054232 Chr.10: 132708243-132708243 on Build GRCh38
rs750746307 Chr.10: 132708421-132708422 on Build GRCh38
rs778945741 Chr.10: 132708302-132708302 on Build GRCh38
rs112466073 Chr.10: 132708209-132708209 on Build GRCh38
rs761786872 Chr.10: 132708183-132708183 on Build GRCh38
rs779617346 Chr.10: 132708289-132708289 on Build GRCh38
rs368149106 Chr.10: 132708363-132708363 on Build GRCh38
rs374793547 Chr.10: 132708400-132708400 on Build GRCh38
rs372448375 Chr.10: 132708304-132708304 on Build GRCh38
rs763394017 Chr.10: 132708341-132708341 on Build GRCh38
rs749911870 Chr.10: 132708272-132708272 on Build GRCh38
rs886953516 Chr.10: 132708342-132708342 on Build GRCh38
rs141017103 Chr.10: 132708347-132708347 on Build GRCh38
rs768900259 Chr.10: 132708394-132708394 on Build GRCh38
rs201462366 Chr.10: 132708372-132708372 on Build GRCh38
rs924148940 Chr.10: 132708192-132708192 on Build GRCh38
rs372787901 Chr.10: 132708382-132708382 on Build GRCh38
rs764941760 Chr.10: 132708418-132708418 on Build GRCh38
rs138322259 Chr.10: 132708162-132708162 on Build GRCh38
rs534701871 Chr.10: 132708284-132708284 on Build GRCh38
rs1031633128 Chr.10: 132708423-132708423 on Build GRCh38
rs575655431 Chr.10: 132708257-132708257 on Build GRCh38
rs991188907 Chr.10: 132708173-132708173 on Build GRCh38
rs375843411 Chr.10: 132708390-132708390 on Build GRCh38
rs1039706669 Chr.10: 132708273-132708273 on Build GRCh38
rs772582733 Chr.10: 132708401-132708401 on Build GRCh38
rs544375296 Chr.10: 132708258-132708258 on Build GRCh38
rs774635988 Chr.10: 132708398-132708398 on Build GRCh38
rs910066131 Chr.10: 132708249-132708249 on Build GRCh38
rs1003174752 Chr.10: 132708193-132708193 on Build GRCh38
rs1025305675 Chr.10: 132708203-132708203 on Build GRCh38
rs749942469 Chr.10: 132708369-132708369 on Build GRCh38
rs758337279 Chr.10: 132708379-132708379 on Build GRCh38
rs752436424 Chr.10: 132708419-132708419 on Build GRCh38
rs529340981 Chr.10: 132708310-132708310 on Build GRCh38
rs546762032 Chr.10: 132708152-132708152 on Build GRCh38
rs968352691 Chr.10: 132708181-132708181 on Build GRCh38
rs559627024 Chr.10: 132708384-132708384 on Build GRCh38
rs374675950 Chr.10: 132708337-132708337 on Build GRCh38
rs778626509 Chr.10: 132708376-132708376 on Build GRCh38
rs775793924 Chr.10: 132708340-132708340 on Build GRCh38
rs536359666 Chr.10: 132708163-132708163 on Build GRCh38
rs912687218 Chr.10: 132708234-132708234 on Build GRCh38
rs745955341 Chr.10: 132708383-132708383 on Build GRCh38
rs754631791 Chr.10: 132708375-132708375 on Build GRCh38
rs958361183 Chr.10: 132708167-132708167 on Build GRCh38
rs760209994 Chr.10: 132708408-132708408 on Build GRCh38
rs753370707 Chr.10: 132708417-132708417 on Build GRCh38
rs41301113 Chr.10: 132708298-132708298 on Build GRCh38
rs773661406 Chr.10: 132708404-132708404 on Build GRCh38

View Full Product Details