Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 20364117-20364639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTATIP2
Entrez Gene ID
10553
Gene Name
HIV-1 Tat interactive protein 2
Gene Aliases
CC3, SDR44U1, TIP30
UniGene
Hs.90753
Gene Chromosome Location
Chr.11: 20363685-20383783 on Build GRCh38
Transcripts
NM_001098520.1 NM_001098521.1 NM_001098522.1 NM_001098523.1 NM_006410.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
925894 c.104T>C p.L35S Chr.11: 20364341-20364341 on Build GRCh38
4032035 c.120G>A p.E40E Chr.11: 20364357-20364357 on Build GRCh38
1585651 c.206T>C p.L69S Chr.11: 20364341-20364341 on Build GRCh38
5961779 c.115C>A p.L39M Chr.11: 20364352-20364352 on Build GRCh38
1638802 c.123G>C p.Q41H Chr.11: 20364360-20364360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748247933 Chr.11: 20364466-20364466 on Build GRCh38
rs371982853 Chr.11: 20364202-20364202 on Build GRCh38
rs931427014 Chr.11: 20364618-20364618 on Build GRCh38
rs771485360 Chr.11: 20364286-20364286 on Build GRCh38
rs373770270 Chr.11: 20364467-20364467 on Build GRCh38
rs527767104 Chr.11: 20364328-20364328 on Build GRCh38
rs752464428 Chr.11: 20364393-20364393 on Build GRCh38
rs369625277 Chr.11: 20364242-20364242 on Build GRCh38
rs954472607 Chr.11: 20364222-20364222 on Build GRCh38
rs766257066 Chr.11: 20364312-20364312 on Build GRCh38
rs779311906 Chr.11: 20364428-20364428 on Build GRCh38
rs772015987 Chr.11: 20364294-20364294 on Build GRCh38
rs199846818 Chr.11: 20364233-20364233 on Build GRCh38
rs202128953 Chr.11: 20364346-20364346 on Build GRCh38
rs778437497 Chr.11: 20364265-20364265 on Build GRCh38
rs995652502 Chr.11: 20364181-20364181 on Build GRCh38
rs570755891 Chr.11: 20364500-20364500 on Build GRCh38
rs539357656 Chr.11: 20364391-20364391 on Build GRCh38
rs751151798 Chr.11: 20364313-20364313 on Build GRCh38
rs200846453 Chr.11: 20364362-20364362 on Build GRCh38
rs780254299 Chr.11: 20364449-20364449 on Build GRCh38
rs757672606 Chr.11: 20364335-20364335 on Build GRCh38
rs1030738439 Chr.11: 20364458-20364458 on Build GRCh38
rs879187362 Chr.11: 20364485-20364485 on Build GRCh38
rs762736025 Chr.11: 20364306-20364306 on Build GRCh38
rs928731103 Chr.11: 20364142-20364142 on Build GRCh38
rs775602858 Chr.11: 20364297-20364297 on Build GRCh38
rs145143466 Chr.11: 20364381-20364381 on Build GRCh38
rs773141766 Chr.11: 20364305-20364305 on Build GRCh38
rs747538096 Chr.11: 20364277-20364278 on Build GRCh38
rs574258970 Chr.11: 20364583-20364583 on Build GRCh38
rs767691558 Chr.11: 20364199-20364199 on Build GRCh38
rs746251655 Chr.11: 20364345-20364345 on Build GRCh38
rs577039737 Chr.11: 20364440-20364440 on Build GRCh38
rs868775571 Chr.11: 20364249-20364250 on Build GRCh38
rs548276280 Chr.11: 20364179-20364179 on Build GRCh38
rs977800021 Chr.11: 20364231-20364231 on Build GRCh38
rs537724443 Chr.11: 20364518-20364518 on Build GRCh38
rs533828029 Chr.11: 20364253-20364253 on Build GRCh38
rs960108674 Chr.11: 20364611-20364611 on Build GRCh38
rs759684347 Chr.11: 20364380-20364380 on Build GRCh38
rs778382489 Chr.11: 20364247-20364248 on Build GRCh38
rs957802260 Chr.11: 20364495-20364495 on Build GRCh38
rs771399529 Chr.11: 20364435-20364448 on Build GRCh38
rs781291964 Chr.11: 20364454-20364454 on Build GRCh38
rs753694777 Chr.11: 20364201-20364201 on Build GRCh38
rs747641513 Chr.11: 20364252-20364252 on Build GRCh38
rs747293118 Chr.11: 20364450-20364450 on Build GRCh38
rs750174056 Chr.11: 20364218-20364218 on Build GRCh38
rs774156790 Chr.11: 20364471-20364471 on Build GRCh38
rs566317704 Chr.11: 20364522-20364522 on Build GRCh38
rs754363466 Chr.11: 20364412-20364412 on Build GRCh38
rs1037024799 Chr.11: 20364578-20364578 on Build GRCh38
rs758313233 Chr.11: 20364141-20364141 on Build GRCh38
rs199622189 Chr.11: 20364452-20364452 on Build GRCh38
rs566345801 Chr.11: 20364221-20364221 on Build GRCh38
rs948005942 Chr.11: 20364277-20364277 on Build GRCh38
rs758207523 Chr.11: 20364226-20364226 on Build GRCh38
rs557993544 Chr.11: 20364392-20364392 on Build GRCh38
rs531502343 Chr.11: 20364171-20364171 on Build GRCh38
rs777175252 Chr.11: 20364478-20364478 on Build GRCh38
rs760767967 Chr.11: 20364299-20364299 on Build GRCh38
rs774420448 Chr.11: 20364288-20364288 on Build GRCh38
rs752192706 Chr.11: 20364319-20364319 on Build GRCh38
rs749629526 Chr.11: 20364163-20364163 on Build GRCh38
rs555960781 Chr.11: 20364557-20364557 on Build GRCh38
rs779615431 Chr.11: 20364229-20364229 on Build GRCh38
rs200473579 Chr.11: 20364385-20364385 on Build GRCh38
rs765101155 Chr.11: 20364303-20364303 on Build GRCh38
rs773123281 Chr.11: 20364373-20364373 on Build GRCh38
rs1035683252 Chr.11: 20364595-20364595 on Build GRCh38
rs1040774431 Chr.11: 20364326-20364326 on Build GRCh38
rs1046313961 Chr.11: 20364292-20364292 on Build GRCh38
rs938668315 Chr.11: 20364575-20364575 on Build GRCh38
rs775589236 Chr.11: 20364348-20364348 on Build GRCh38
rs916361723 Chr.11: 20364519-20364519 on Build GRCh38
rs747025235 Chr.11: 20364349-20364349 on Build GRCh38
rs1019736002 Chr.11: 20364223-20364223 on Build GRCh38
rs759366963 Chr.11: 20364472-20364472 on Build GRCh38
rs746148379 Chr.11: 20364293-20364293 on Build GRCh38
rs757744999 Chr.11: 20364415-20364415 on Build GRCh38
rs1048475063 Chr.11: 20364619-20364619 on Build GRCh38
rs991269966 Chr.11: 20364173-20364173 on Build GRCh38
rs749593203 Chr.11: 20364280-20364280 on Build GRCh38
rs191533610 Chr.11: 20364327-20364327 on Build GRCh38
rs900902008 Chr.11: 20364337-20364337 on Build GRCh38
rs918664460 Chr.11: 20364597-20364597 on Build GRCh38
rs750743670 Chr.11: 20364439-20364439 on Build GRCh38
rs779372390 Chr.11: 20364339-20364339 on Build GRCh38
rs866764307 Chr.11: 20364279-20364279 on Build GRCh38
rs751294106 Chr.11: 20364232-20364232 on Build GRCh38
rs754617086 Chr.11: 20364315-20364315 on Build GRCh38
rs774580732 Chr.11: 20364237-20364237 on Build GRCh38
rs145549108 Chr.11: 20364396-20364396 on Build GRCh38
rs763744772 Chr.11: 20364403-20364403 on Build GRCh38
rs768795320 Chr.11: 20364357-20364357 on Build GRCh38
rs780793051 Chr.11: 20364248-20364248 on Build GRCh38
rs982409418 Chr.11: 20364546-20364546 on Build GRCh38
rs767095366 Chr.11: 20364316-20364316 on Build GRCh38
rs765193068 Chr.11: 20364203-20364203 on Build GRCh38

View Full Product Details