Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97930433-97930908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEMGN
Entrez Gene ID
55363
Gene Name
hemogen
Gene Aliases
CT155, EDAG, EDAG-1, NDR
UniGene
Hs.176626
Gene Chromosome Location
Chr.9: 97926791-97944915 on Build GRCh38
Transcripts
NM_018437.4 NM_197978.2 XM_005252086.1 XM_011518845.2 XM_011518846.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2793229 c.640C>T p.H214Y Chr.9: 97930755-97930755 on Build GRCh38
4498478 c.520C>T p.P174S Chr.9: 97930875-97930875 on Build GRCh38
1103066 c.918G>A p.E306E Chr.9: 97930477-97930477 on Build GRCh38
5954531 c.547C>A p.L183I Chr.9: 97930848-97930848 on Build GRCh38
1103067 c.915T>C p.A305A Chr.9: 97930480-97930480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746162223 Chr.9: 97930741-97930741 on Build GRCh38
rs576790299 Chr.9: 97930809-97930809 on Build GRCh38
rs778937138 Chr.9: 97930814-97930814 on Build GRCh38
rs768261992 Chr.9: 97930771-97930771 on Build GRCh38
rs766236906 Chr.9: 97930677-97930677 on Build GRCh38
rs376645183 Chr.9: 97930837-97930837 on Build GRCh38
rs150614678 Chr.9: 97930658-97930658 on Build GRCh38
rs770329844 Chr.9: 97930671-97930671 on Build GRCh38
rs1004251163 Chr.9: 97930715-97930717 on Build GRCh38
rs371151700 Chr.9: 97930796-97930796 on Build GRCh38
rs140700506 Chr.9: 97930559-97930559 on Build GRCh38
rs767025188 Chr.9: 97930460-97930460 on Build GRCh38
rs139808473 Chr.9: 97930804-97930804 on Build GRCh38
rs368946756 Chr.9: 97930498-97930498 on Build GRCh38
rs953425759 Chr.9: 97930815-97930815 on Build GRCh38
rs574361292 Chr.9: 97930754-97930754 on Build GRCh38
rs377743188 Chr.9: 97930721-97930721 on Build GRCh38
rs772763536 Chr.9: 97930597-97930597 on Build GRCh38
rs554987978 Chr.9: 97930800-97930800 on Build GRCh38
rs552975476 Chr.9: 97930680-97930680 on Build GRCh38
rs751475385 Chr.9: 97930803-97930803 on Build GRCh38
rs763785625 Chr.9: 97930486-97930486 on Build GRCh38
rs758332990 Chr.9: 97930516-97930516 on Build GRCh38
rs764155775 Chr.9: 97930806-97930806 on Build GRCh38
rs191644480 Chr.9: 97930836-97930836 on Build GRCh38
rs779323105 Chr.9: 97930730-97930730 on Build GRCh38
rs996698063 Chr.9: 97930735-97930735 on Build GRCh38
rs948723546 Chr.9: 97930857-97930857 on Build GRCh38
rs781231718 Chr.9: 97930520-97930520 on Build GRCh38
rs775207073 Chr.9: 97930466-97930466 on Build GRCh38
rs141818371 Chr.9: 97930578-97930578 on Build GRCh38
rs201689350 Chr.9: 97930799-97930799 on Build GRCh38
rs761510736 Chr.9: 97930785-97930785 on Build GRCh38
rs760200705 Chr.9: 97930631-97930631 on Build GRCh38
rs754061719 Chr.9: 97930722-97930722 on Build GRCh38
rs750692896 Chr.9: 97930823-97930823 on Build GRCh38
rs776067752 Chr.9: 97930784-97930784 on Build GRCh38
rs747543860 Chr.9: 97930761-97930761 on Build GRCh38
rs976201261 Chr.9: 97930756-97930756 on Build GRCh38
rs750240939 Chr.9: 97930659-97930659 on Build GRCh38
rs990530163 Chr.9: 97930824-97930824 on Build GRCh38
rs756190428 Chr.9: 97930526-97930526 on Build GRCh38
rs147210952 Chr.9: 97930530-97930530 on Build GRCh38
rs753549435 Chr.9: 97930492-97930492 on Build GRCh38
rs914595266 Chr.9: 97930830-97930830 on Build GRCh38
rs956159427 Chr.9: 97930822-97930822 on Build GRCh38
rs777930808 Chr.9: 97930529-97930529 on Build GRCh38
rs755598168 Chr.9: 97930879-97930879 on Build GRCh38
rs750340722 Chr.9: 97930508-97930508 on Build GRCh38
rs907351528 Chr.9: 97930476-97930476 on Build GRCh38
rs955198864 Chr.9: 97930782-97930782 on Build GRCh38
rs760391229 Chr.9: 97930473-97930473 on Build GRCh38
rs756135648 Chr.9: 97930710-97930710 on Build GRCh38
rs780671478 Chr.9: 97930757-97930757 on Build GRCh38
rs769155522 Chr.9: 97930481-97930481 on Build GRCh38
rs772918303 Chr.9: 97930795-97930795 on Build GRCh38
rs374570190 Chr.9: 97930643-97930643 on Build GRCh38
rs547930508 Chr.9: 97930634-97930634 on Build GRCh38
rs776155696 Chr.9: 97930630-97930630 on Build GRCh38
rs781594044 Chr.9: 97930632-97930632 on Build GRCh38
rs896163783 Chr.9: 97930540-97930540 on Build GRCh38
rs746383608 Chr.9: 97930585-97930585 on Build GRCh38
rs986563473 Chr.9: 97930829-97930829 on Build GRCh38
rs757039108 Chr.9: 97930497-97930497 on Build GRCh38
rs748164340 Chr.9: 97930523-97930523 on Build GRCh38
rs75267104 Chr.9: 97930728-97930728 on Build GRCh38
rs375684699 Chr.9: 97930810-97930810 on Build GRCh38
rs747490832 Chr.9: 97930854-97930854 on Build GRCh38
rs772573235 Chr.9: 97930746-97930746 on Build GRCh38
rs762816603 Chr.9: 97930672-97930672 on Build GRCh38
rs757490390 Chr.9: 97930726-97930726 on Build GRCh38

View Full Product Details