Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 137001070-137001641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL20RA
Entrez Gene ID
53832
Gene Name
interleukin 20 receptor subunit alpha
Gene Aliases
CRF2-8, IL-20R-alpha, IL-20R1, IL-20RA
UniGene
Hs.445868
Gene Chromosome Location
Chr.6: 136999971-137045366 on Build GRCh38
Transcripts
NM_001278722.1 NM_001278723.1 NM_001278724.1 NM_014432.3 XM_006715506.2 XM_011535904.2 XM_017010954.1 XM_017010955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3621015 c.1607C>T p.T536I Chr.6: 137001613-137001613 on Build GRCh38
5822709 c.1643A>T p.Y548F Chr.6: 137001577-137001577 on Build GRCh38
5418715 c.1634G>A p.W545* Chr.6: 137001586-137001586 on Build GRCh38
230105 c.1606A>G p.T536A Chr.6: 137001614-137001614 on Build GRCh38
232534 c.1654G>A p.E552K Chr.6: 137001566-137001566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984237004 Chr.6: 137001265-137001265 on Build GRCh38
rs752577982 Chr.6: 137001611-137001611 on Build GRCh38
rs188274860 Chr.6: 137001325-137001325 on Build GRCh38
rs761996780 Chr.6: 137001527-137001527 on Build GRCh38
rs911024786 Chr.6: 137001318-137001318 on Build GRCh38
rs948140924 Chr.6: 137001483-137001483 on Build GRCh38
rs750673622 Chr.6: 137001569-137001569 on Build GRCh38
rs761253102 Chr.6: 137001509-137001509 on Build GRCh38
rs999139075 Chr.6: 137001151-137001151 on Build GRCh38
rs1023037232 Chr.6: 137001183-137001183 on Build GRCh38
rs977375997 Chr.6: 137001292-137001292 on Build GRCh38
rs1056437607 Chr.6: 137001406-137001406 on Build GRCh38
rs778404293 Chr.6: 137001122-137001122 on Build GRCh38
rs998178969 Chr.6: 137001598-137001598 on Build GRCh38
rs768902576 Chr.6: 137001523-137001523 on Build GRCh38
rs578012285 Chr.6: 137001115-137001115 on Build GRCh38
rs771261881 Chr.6: 137001365-137001365 on Build GRCh38
rs988492968 Chr.6: 137001242-137001242 on Build GRCh38
rs186439336 Chr.6: 137001583-137001583 on Build GRCh38
rs76578405 Chr.6: 137001289-137001289 on Build GRCh38
rs941183463 Chr.6: 137001332-137001332 on Build GRCh38
rs902812812 Chr.6: 137001434-137001434 on Build GRCh38
rs957201661 Chr.6: 137001217-137001217 on Build GRCh38
rs780827570 Chr.6: 137001606-137001606 on Build GRCh38
rs371502611 Chr.6: 137001497-137001497 on Build GRCh38
rs1029776627 Chr.6: 137001284-137001284 on Build GRCh38
rs180800340 Chr.6: 137001381-137001381 on Build GRCh38
rs973899371 Chr.6: 137001335-137001335 on Build GRCh38
rs1012755413 Chr.6: 137001102-137001102 on Build GRCh38
rs568906775 Chr.6: 137001287-137001287 on Build GRCh38
rs568663769 Chr.6: 137001363-137001363 on Build GRCh38
rs143965193 Chr.6: 137001154-137001154 on Build GRCh38
rs918499073 Chr.6: 137001369-137001369 on Build GRCh38
rs1001089450 Chr.6: 137001170-137001170 on Build GRCh38
rs141167070 Chr.6: 137001491-137001491 on Build GRCh38
rs534146931 Chr.6: 137001366-137001366 on Build GRCh38
rs751386481 Chr.6: 137001586-137001586 on Build GRCh38
rs1042814990 Chr.6: 137001403-137001403 on Build GRCh38
rs896559017 Chr.6: 137001099-137001099 on Build GRCh38
rs148602084 Chr.6: 137001171-137001171 on Build GRCh38
rs562468304 Chr.6: 137001173-137001173 on Build GRCh38
rs552155403 Chr.6: 137001238-137001238 on Build GRCh38
rs117482843 Chr.6: 137001114-137001114 on Build GRCh38
rs765481430 Chr.6: 137001551-137001551 on Build GRCh38
rs985193609 Chr.6: 137001301-137001301 on Build GRCh38
rs755010486 Chr.6: 137001592-137001592 on Build GRCh38
rs749774468 Chr.6: 137001342-137001342 on Build GRCh38
rs904041895 Chr.6: 137001144-137001144 on Build GRCh38
rs896482819 Chr.6: 137001450-137001450 on Build GRCh38
rs1015989183 Chr.6: 137001268-137001268 on Build GRCh38
rs777106543 Chr.6: 137001526-137001526 on Build GRCh38
rs1001017592 Chr.6: 137001553-137001553 on Build GRCh38
rs774888035 Chr.6: 137001508-137001508 on Build GRCh38
rs915421628 Chr.6: 137001380-137001380 on Build GRCh38
rs762969669 Chr.6: 137001576-137001576 on Build GRCh38
rs952802189 Chr.6: 137001255-137001255 on Build GRCh38

View Full Product Details