Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 2718664-2719232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNV2
Entrez Gene ID
169522
Gene Name
potassium voltage-gated channel modifier subfamily V member 2
Gene Aliases
KV11.1, Kv8.2, RCD3B
UniGene
Hs.622675
Gene Chromosome Location
Chr.9: 2717526-2730037 on Build GRCh38
Transcripts
NM_133497.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3732145 c.1026C>A p.A342A Chr.9: 2718765-2718765 on Build GRCh38
6691959 c.1272T>C p.T424T Chr.9: 2719011-2719011 on Build GRCh38
1107972 c.1094C>T p.T365M Chr.9: 2718833-2718833 on Build GRCh38
753701 c.1221C>A p.C407* Chr.9: 2718960-2718960 on Build GRCh38
5893374 c.1260G>A p.M420I Chr.9: 2718999-2718999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201033142 Chr.9: 2718922-2718922 on Build GRCh38
rs886063821 Chr.9: 2718791-2718792 on Build GRCh38
rs750359924 Chr.9: 2718792-2718792 on Build GRCh38
rs777069016 Chr.9: 2718731-2718731 on Build GRCh38
rs543556674 Chr.9: 2718995-2718995 on Build GRCh38
rs761948684 Chr.9: 2719189-2719189 on Build GRCh38
rs972364567 Chr.9: 2719059-2719059 on Build GRCh38
rs568147806 Chr.9: 2718862-2718862 on Build GRCh38
rs1044937878 Chr.9: 2718793-2718793 on Build GRCh38
rs759430990 Chr.9: 2718879-2718879 on Build GRCh38
rs749220459 Chr.9: 2719013-2719013 on Build GRCh38
rs757270560 Chr.9: 2718712-2718712 on Build GRCh38
rs750338987 Chr.9: 2719133-2719133 on Build GRCh38
rs752172249 Chr.9: 2718744-2718745 on Build GRCh38
rs750012815 Chr.9: 2719067-2719067 on Build GRCh38
rs770442556 Chr.9: 2718913-2718913 on Build GRCh38
rs921097273 Chr.9: 2718911-2718911 on Build GRCh38
rs368533172 Chr.9: 2719115-2719115 on Build GRCh38
rs201201478 Chr.9: 2719110-2719110 on Build GRCh38
rs761283518 Chr.9: 2718695-2718695 on Build GRCh38
rs980513372 Chr.9: 2718709-2718709 on Build GRCh38
rs766985581 Chr.9: 2718824-2718824 on Build GRCh38
rs148096197 Chr.9: 2718880-2718880 on Build GRCh38
rs138924201 Chr.9: 2718925-2718925 on Build GRCh38
rs768011155 Chr.9: 2718693-2718693 on Build GRCh38
rs762423786 Chr.9: 2719107-2719107 on Build GRCh38
rs754528788 Chr.9: 2719121-2719121 on Build GRCh38
rs906005731 Chr.9: 2719024-2719024 on Build GRCh38
rs1042566787 Chr.9: 2719198-2719198 on Build GRCh38
rs143664586 Chr.9: 2718868-2718868 on Build GRCh38
rs141789289 Chr.9: 2718765-2718765 on Build GRCh38
rs142744007 Chr.9: 2718822-2718822 on Build GRCh38
rs374070004 Chr.9: 2718861-2718861 on Build GRCh38
rs777405471 Chr.9: 2718704-2718704 on Build GRCh38
rs373961607 Chr.9: 2719033-2719033 on Build GRCh38
rs139767162 Chr.9: 2719094-2719094 on Build GRCh38
rs754275640 Chr.9: 2718697-2718697 on Build GRCh38
rs986350598 Chr.9: 2718728-2718729 on Build GRCh38
rs746629468 Chr.9: 2719163-2719163 on Build GRCh38
rs755848294 Chr.9: 2719075-2719075 on Build GRCh38
rs748084580 Chr.9: 2719096-2719099 on Build GRCh38
rs769944276 Chr.9: 2719140-2719140 on Build GRCh38
rs760904700 Chr.9: 2718964-2718964 on Build GRCh38
rs763381795 Chr.9: 2718819-2718819 on Build GRCh38
rs370789466 Chr.9: 2719090-2719090 on Build GRCh38
rs374028926 Chr.9: 2719143-2719143 on Build GRCh38
rs764218863 Chr.9: 2719102-2719102 on Build GRCh38
rs780804898 Chr.9: 2718713-2718713 on Build GRCh38
rs769505203 Chr.9: 2718872-2718880 on Build GRCh38
rs904754294 Chr.9: 2718939-2718939 on Build GRCh38
rs113766372 Chr.9: 2719182-2719182 on Build GRCh38
rs753171063 Chr.9: 2718973-2718973 on Build GRCh38
rs769184629 Chr.9: 2719004-2719004 on Build GRCh38
rs780756183 Chr.9: 2718834-2718836 on Build GRCh38
rs750825755 Chr.9: 2719111-2719111 on Build GRCh38
rs200018497 Chr.9: 2718884-2718884 on Build GRCh38
rs141881396 Chr.9: 2718887-2718887 on Build GRCh38
rs764608030 Chr.9: 2719095-2719095 on Build GRCh38
rs376858051 Chr.9: 2718710-2718710 on Build GRCh38
rs75645675 Chr.9: 2718802-2718802 on Build GRCh38
rs574582499 Chr.9: 2718968-2718968 on Build GRCh38
rs777263450 Chr.9: 2718777-2718777 on Build GRCh38
rs147385726 Chr.9: 2718914-2718914 on Build GRCh38
rs764587081 Chr.9: 2719058-2719058 on Build GRCh38
rs150231850 Chr.9: 2718905-2718905 on Build GRCh38
rs968160228 Chr.9: 2719056-2719056 on Build GRCh38
rs960100570 Chr.9: 2718843-2718843 on Build GRCh38
rs761404248 Chr.9: 2718786-2718786 on Build GRCh38
rs530163400 Chr.9: 2718779-2718779 on Build GRCh38
rs143382624 Chr.9: 2719083-2719083 on Build GRCh38
rs376016220 Chr.9: 2718906-2718906 on Build GRCh38
rs372668634 Chr.9: 2718703-2718703 on Build GRCh38
rs778638987 Chr.9: 2718844-2718844 on Build GRCh38
rs35221706 Chr.9: 2719111-2719111 on Build GRCh38
rs745605626 Chr.9: 2718759-2718759 on Build GRCh38
rs748514014 Chr.9: 2718901-2718901 on Build GRCh38
rs1000777419 Chr.9: 2719057-2719057 on Build GRCh38
rs745697896 Chr.9: 2718715-2718715 on Build GRCh38
rs751864649 Chr.9: 2719062-2719062 on Build GRCh38
rs1030493328 Chr.9: 2718821-2718821 on Build GRCh38
rs745375996 Chr.9: 2718947-2718947 on Build GRCh38
rs1028221432 Chr.9: 2719209-2719209 on Build GRCh38
rs979304433 Chr.9: 2718742-2718742 on Build GRCh38
rs768136198 Chr.9: 2718820-2718820 on Build GRCh38
rs765927725 Chr.9: 2718878-2718878 on Build GRCh38
rs534563578 Chr.9: 2718848-2718848 on Build GRCh38
rs527647651 Chr.9: 2718733-2718733 on Build GRCh38
rs201425989 Chr.9: 2718782-2718782 on Build GRCh38
rs757550503 Chr.9: 2719064-2719064 on Build GRCh38
rs780711475 Chr.9: 2718945-2718945 on Build GRCh38
rs970151348 Chr.9: 2719020-2719020 on Build GRCh38
rs748180390 Chr.9: 2719087-2719087 on Build GRCh38
rs267602205 Chr.9: 2718831-2718831 on Build GRCh38
rs745592530 Chr.9: 2718996-2718996 on Build GRCh38
rs768545361 Chr.9: 2718956-2718956 on Build GRCh38
rs903486029 Chr.9: 2719201-2719201 on Build GRCh38
rs776710119 Chr.9: 2719035-2719035 on Build GRCh38
rs765119911 Chr.9: 2718918-2718918 on Build GRCh38
rs141378856 Chr.9: 2718696-2718696 on Build GRCh38
rs199886720 Chr.9: 2718825-2718825 on Build GRCh38
rs537049308 Chr.9: 2719125-2719125 on Build GRCh38
rs376306786 Chr.9: 2719042-2719042 on Build GRCh38
rs774569926 Chr.9: 2718723-2718723 on Build GRCh38
rs761237693 Chr.9: 2718933-2718933 on Build GRCh38
rs1008177536 Chr.9: 2719213-2719213 on Build GRCh38
rs761443523 Chr.9: 2718823-2718823 on Build GRCh38
rs774716111 Chr.9: 2719104-2719104 on Build GRCh38
rs775678049 Chr.9: 2719141-2719141 on Build GRCh38
rs796086240 Chr.9: 2719202-2719203 on Build GRCh38
rs773122223 Chr.9: 2718773-2718773 on Build GRCh38
rs748906559 Chr.9: 2718880-2718881 on Build GRCh38
rs374211938 Chr.9: 2718811-2718811 on Build GRCh38
rs750883269 Chr.9: 2718694-2718694 on Build GRCh38
rs946520647 Chr.9: 2718738-2718738 on Build GRCh38
rs968707552 Chr.9: 2718750-2718750 on Build GRCh38
rs773388753 Chr.9: 2718729-2718729 on Build GRCh38
rs745311059 Chr.9: 2718871-2718872 on Build GRCh38
rs751106123 Chr.9: 2718748-2718748 on Build GRCh38
rs779755195 Chr.9: 2719005-2719005 on Build GRCh38
rs750673481 Chr.9: 2718924-2718924 on Build GRCh38
rs763052137 Chr.9: 2718955-2718955 on Build GRCh38
rs770448872 Chr.9: 2718810-2718810 on Build GRCh38
rs568111360 Chr.9: 2719112-2719112 on Build GRCh38
rs200767774 Chr.9: 2718789-2718789 on Build GRCh38
rs770511296 Chr.9: 2718942-2718942 on Build GRCh38
rs779290388 Chr.9: 2718897-2718897 on Build GRCh38
rs370412027 Chr.9: 2718874-2718874 on Build GRCh38
rs201361335 Chr.9: 2719019-2719019 on Build GRCh38
rs778096918 Chr.9: 2719197-2719197 on Build GRCh38
rs202139237 Chr.9: 2718916-2718916 on Build GRCh38
rs752390776 Chr.9: 2718737-2718737 on Build GRCh38
rs372410707 Chr.9: 2718903-2718903 on Build GRCh38
rs370525145 Chr.9: 2718993-2718993 on Build GRCh38
rs200444639 Chr.9: 2718935-2718935 on Build GRCh38
rs765751812 Chr.9: 2718734-2718734 on Build GRCh38
rs559765711 Chr.9: 2719080-2719080 on Build GRCh38
rs757516625 Chr.9: 2718883-2718883 on Build GRCh38
rs771848113 Chr.9: 2719117-2719117 on Build GRCh38
rs781669034 Chr.9: 2719065-2719065 on Build GRCh38
rs753695734 Chr.9: 2718798-2718798 on Build GRCh38
rs775290203 Chr.9: 2718919-2718919 on Build GRCh38
rs758106979 Chr.9: 2718740-2718740 on Build GRCh38
rs763962122 Chr.9: 2718912-2718912 on Build GRCh38
rs1036473671 Chr.9: 2718806-2718806 on Build GRCh38
rs748708161 Chr.9: 2718932-2718932 on Build GRCh38
rs547033951 Chr.9: 2718833-2718833 on Build GRCh38
rs774289518 Chr.9: 2718753-2718753 on Build GRCh38
rs139581747 Chr.9: 2719047-2719047 on Build GRCh38
rs761886297 Chr.9: 2718685-2718685 on Build GRCh38
rs781116632 Chr.9: 2718754-2718754 on Build GRCh38
rs747253127 Chr.9: 2718804-2718804 on Build GRCh38
rs774340434 Chr.9: 2718957-2718957 on Build GRCh38
rs199497822 Chr.9: 2718965-2718965 on Build GRCh38
rs776176542 Chr.9: 2718896-2718896 on Build GRCh38
rs765420295 Chr.9: 2718972-2718972 on Build GRCh38
rs754637111 Chr.9: 2718700-2718700 on Build GRCh38
rs781374665 Chr.9: 2718860-2718860 on Build GRCh38
rs774378296 Chr.9: 2718684-2718684 on Build GRCh38
rs780069100 Chr.9: 2718891-2718891 on Build GRCh38
rs182424197 Chr.9: 2719211-2719211 on Build GRCh38
rs118098044 Chr.9: 2719204-2719204 on Build GRCh38
rs142146344 Chr.9: 2719021-2719021 on Build GRCh38
rs772042385 Chr.9: 2718727-2718727 on Build GRCh38
rs538237334 Chr.9: 2719077-2719077 on Build GRCh38
rs778061758 Chr.9: 2718978-2718978 on Build GRCh38
rs776795213 Chr.9: 2719120-2719120 on Build GRCh38
rs535399096 Chr.9: 2719208-2719208 on Build GRCh38
rs763838984 Chr.9: 2718780-2718780 on Build GRCh38
rs977445103 Chr.9: 2718837-2718837 on Build GRCh38
rs931321136 Chr.9: 2719116-2719116 on Build GRCh38
rs1001901052 Chr.9: 2718841-2718841 on Build GRCh38
rs367972852 Chr.9: 2718930-2718930 on Build GRCh38
rs763035119 Chr.9: 2719098-2719098 on Build GRCh38
rs960007958 Chr.9: 2718989-2718989 on Build GRCh38
rs763492761 Chr.9: 2718741-2718741 on Build GRCh38
rs199829137 Chr.9: 2718768-2718768 on Build GRCh38
rs758953080 Chr.9: 2718975-2718975 on Build GRCh38
rs747113494 Chr.9: 2718774-2718774 on Build GRCh38
rs778977288 Chr.9: 2719086-2719086 on Build GRCh38
rs1020602079 Chr.9: 2719055-2719055 on Build GRCh38
rs768486552 Chr.9: 2718937-2718937 on Build GRCh38
rs200266207 Chr.9: 2718830-2718830 on Build GRCh38
rs767518507 Chr.9: 2718870-2718870 on Build GRCh38
rs780625453 Chr.9: 2718867-2718867 on Build GRCh38
rs775556815 Chr.9: 2718952-2718952 on Build GRCh38
rs111792867 Chr.9: 2718994-2718994 on Build GRCh38
rs7860945 Chr.9: 2718717-2718717 on Build GRCh38
rs779297718 Chr.9: 2718931-2718931 on Build GRCh38
rs553161580 Chr.9: 2719191-2719191 on Build GRCh38
rs773104305 Chr.9: 2719091-2719091 on Build GRCh38
rs1017035625 Chr.9: 2719207-2719207 on Build GRCh38
rs749775089 Chr.9: 2718895-2718895 on Build GRCh38
rs369528480 Chr.9: 2719137-2719137 on Build GRCh38
rs774116702 Chr.9: 2719041-2719041 on Build GRCh38
rs746352514 Chr.9: 2718808-2718808 on Build GRCh38
rs1051219107 Chr.9: 2718770-2718770 on Build GRCh38
rs779641908 Chr.9: 2718878-2718879 on Build GRCh38
rs563513486 Chr.9: 2719001-2719001 on Build GRCh38
rs377129306 Chr.9: 2718838-2718838 on Build GRCh38
rs760186581 Chr.9: 2718778-2718778 on Build GRCh38
rs746641535 Chr.9: 2718940-2718940 on Build GRCh38
rs945146175 Chr.9: 2719195-2719195 on Build GRCh38
rs748841744 Chr.9: 2719082-2719082 on Build GRCh38
rs889720803 Chr.9: 2719212-2719212 on Build GRCh38
rs566426805 Chr.9: 2718826-2718826 on Build GRCh38
rs770032221 Chr.9: 2719093-2719093 on Build GRCh38
rs201564009 Chr.9: 2718857-2718857 on Build GRCh38
rs786205064 Chr.9: 2718755-2718763 on Build GRCh38
rs762855201 Chr.9: 2718921-2718921 on Build GRCh38
rs771741178 Chr.9: 2718869-2718869 on Build GRCh38
rs971928763 Chr.9: 2719209-2719210 on Build GRCh38
rs977830892 Chr.9: 2718762-2718762 on Build GRCh38
rs769715394 Chr.9: 2718948-2718948 on Build GRCh38
rs570543476 Chr.9: 2719161-2719161 on Build GRCh38
rs759838750 Chr.9: 2719038-2719038 on Build GRCh38
rs771129642 Chr.9: 2718807-2718807 on Build GRCh38
rs752928296 Chr.9: 2718871-2718871 on Build GRCh38
rs775957845 Chr.9: 2718816-2718816 on Build GRCh38
rs545829113 Chr.9: 2718730-2718730 on Build GRCh38
rs746028705 Chr.9: 2719092-2719092 on Build GRCh38
rs563806792 Chr.9: 2719014-2719014 on Build GRCh38
rs779892023 Chr.9: 2718760-2718760 on Build GRCh38
rs369069912 Chr.9: 2718796-2718796 on Build GRCh38
rs771957198 Chr.9: 2719029-2719029 on Build GRCh38
rs147822789 Chr.9: 2718687-2718687 on Build GRCh38
rs766136806 Chr.9: 2719113-2719113 on Build GRCh38

View Full Product Details