Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33339240-33339726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK3
Entrez Gene ID
10114
Gene Name
homeodomain interacting protein kinase 3
Gene Aliases
DYRK6, FIST3, PKY, YAK1
UniGene
Hs.201918
Gene Chromosome Location
Chr.11: 33256672-33357023 on Build GRCh38
Transcripts
NM_001048200.2 NM_001278162.1 NM_001278163.1 NM_005734.4 XM_005252729.2 XM_017017076.1 XM_017017077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
926477 c.1489G>T p.E497* Chr.11: 33339410-33339410 on Build GRCh38
7351813 c.1429-1G>T p.? Chr.11: 33339349-33339349 on Build GRCh38
6202626 c.1548A>C p.P516P Chr.11: 33339469-33339469 on Build GRCh38
6244508 c.1480G>T p.D494Y Chr.11: 33339401-33339401 on Build GRCh38
7078566 c.1591C>G p.L531V Chr.11: 33339512-33339512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006552172 Chr.11: 33339317-33339317 on Build GRCh38
rs5790925 Chr.11: 33339341-33339341 on Build GRCh38
rs748108573 Chr.11: 33339445-33339445 on Build GRCh38
rs889442845 Chr.11: 33339304-33339304 on Build GRCh38
rs1033447016 Chr.11: 33339528-33339528 on Build GRCh38
rs765714177 Chr.11: 33339533-33339533 on Build GRCh38
rs201753105 Chr.11: 33339432-33339432 on Build GRCh38
rs16924164 Chr.11: 33339287-33339287 on Build GRCh38
rs758883625 Chr.11: 33339300-33339301 on Build GRCh38
rs779894414 Chr.11: 33339425-33339425 on Build GRCh38
rs775798510 Chr.11: 33339363-33339363 on Build GRCh38
rs776927541 Chr.11: 33339513-33339513 on Build GRCh38
rs754831592 Chr.11: 33339436-33339436 on Build GRCh38
rs376796358 Chr.11: 33339301-33339301 on Build GRCh38
rs796151099 Chr.11: 33339344-33339344 on Build GRCh38
rs776944505 Chr.11: 33339298-33339298 on Build GRCh38
rs888402216 Chr.11: 33339265-33339265 on Build GRCh38
rs749268746 Chr.11: 33339346-33339346 on Build GRCh38
rs759737822 Chr.11: 33339361-33339361 on Build GRCh38
rs774357374 Chr.11: 33339358-33339358 on Build GRCh38
rs11032229 Chr.11: 33339420-33339420 on Build GRCh38
rs551342205 Chr.11: 33339701-33339701 on Build GRCh38
rs757579453 Chr.11: 33339546-33339550 on Build GRCh38
rs769673994 Chr.11: 33339447-33339447 on Build GRCh38
rs956254148 Chr.11: 33339567-33339567 on Build GRCh38
rs201420846 Chr.11: 33339567-33339568 on Build GRCh38
rs771047290 Chr.11: 33339353-33339353 on Build GRCh38
rs751260598 Chr.11: 33339568-33339568 on Build GRCh38
rs941259347 Chr.11: 33339269-33339269 on Build GRCh38
rs577981222 Chr.11: 33339314-33339314 on Build GRCh38
rs111384072 Chr.11: 33339646-33339646 on Build GRCh38
rs749923424 Chr.11: 33339549-33339549 on Build GRCh38
rs769942432 Chr.11: 33339568-33339569 on Build GRCh38
rs746053997 Chr.11: 33339568-33339568 on Build GRCh38
rs914238918 Chr.11: 33339412-33339412 on Build GRCh38
rs371957082 Chr.11: 33339477-33339477 on Build GRCh38
rs1045996807 Chr.11: 33339350-33339350 on Build GRCh38
rs998325916 Chr.11: 33339297-33339297 on Build GRCh38
rs749427477 Chr.11: 33339582-33339583 on Build GRCh38
rs201838261 Chr.11: 33339344-33339344 on Build GRCh38
rs758003431 Chr.11: 33339552-33339552 on Build GRCh38
rs765378445 Chr.11: 33339676-33339676 on Build GRCh38
rs540576523 Chr.11: 33339387-33339387 on Build GRCh38
rs764461793 Chr.11: 33339382-33339382 on Build GRCh38
rs778313269 Chr.11: 33339314-33339314 on Build GRCh38
rs759869903 Chr.11: 33339612-33339612 on Build GRCh38
rs948696490 Chr.11: 33339610-33339610 on Build GRCh38
rs780923607 Chr.11: 33339439-33339439 on Build GRCh38
rs760975918 Chr.11: 33339485-33339485 on Build GRCh38
rs200260579 Chr.11: 33339449-33339449 on Build GRCh38
rs1021920560 Chr.11: 33339319-33339319 on Build GRCh38
rs538568531 Chr.11: 33339562-33339562 on Build GRCh38
rs896999960 Chr.11: 33339295-33339295 on Build GRCh38
rs368066220 Chr.11: 33339372-33339372 on Build GRCh38
rs754743645 Chr.11: 33339577-33339577 on Build GRCh38
rs188152932 Chr.11: 33339497-33339497 on Build GRCh38
rs764800823 Chr.11: 33339391-33339391 on Build GRCh38
rs894663166 Chr.11: 33339328-33339328 on Build GRCh38
rs998774722 Chr.11: 33339501-33339501 on Build GRCh38
rs928988242 Chr.11: 33339283-33339283 on Build GRCh38
rs780248602 Chr.11: 33339583-33339583 on Build GRCh38
rs531571577 Chr.11: 33339682-33339682 on Build GRCh38
rs965904402 Chr.11: 33339601-33339601 on Build GRCh38
rs371398763 Chr.11: 33339299-33339299 on Build GRCh38
rs376578336 Chr.11: 33339302-33339302 on Build GRCh38
rs776651408 Chr.11: 33339334-33339334 on Build GRCh38
rs200686050 Chr.11: 33339340-33339340 on Build GRCh38
rs909891639 Chr.11: 33339616-33339616 on Build GRCh38
rs373644583 Chr.11: 33339362-33339362 on Build GRCh38
rs781024031 Chr.11: 33339585-33339585 on Build GRCh38
rs749959602 Chr.11: 33339413-33339413 on Build GRCh38
rs756659597 Chr.11: 33339390-33339390 on Build GRCh38
rs772244370 Chr.11: 33339464-33339464 on Build GRCh38
rs762368820 Chr.11: 33339532-33339532 on Build GRCh38
rs201133345 Chr.11: 33339450-33339450 on Build GRCh38
rs558054049 Chr.11: 33339303-33339303 on Build GRCh38
rs941229549 Chr.11: 33339618-33339618 on Build GRCh38
rs917372760 Chr.11: 33339594-33339594 on Build GRCh38

View Full Product Details