Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 156511988-156512552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPD2
Entrez Gene ID
2820
Gene Name
glycerol-3-phosphate dehydrogenase 2
Gene Aliases
GDH2, GPDM, mGPDH
UniGene
Hs.512382
Gene Chromosome Location
Chr.2: 156435290-156586403 on Build GRCh38
Transcripts
NM_000408.4 NM_001083112.2 XM_005246469.2 XM_011510977.2 XM_011510978.2 XM_017003830.1 XM_017003831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6901129 c.400-1G>A p.? Chr.2: 156512219-156512219 on Build GRCh38
3837089 c.410T>G p.V137G Chr.2: 156512230-156512230 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs989608231 Chr.2: 156512049-156512049 on Build GRCh38
rs757179675 Chr.2: 156512213-156512213 on Build GRCh38
rs757233126 Chr.2: 156512316-156512316 on Build GRCh38
rs766456077 Chr.2: 156512278-156512278 on Build GRCh38
rs969725120 Chr.2: 156512458-156512458 on Build GRCh38
rs768207129 Chr.2: 156512241-156512241 on Build GRCh38
rs199809700 Chr.2: 156512277-156512277 on Build GRCh38
rs977593447 Chr.2: 156512029-156512029 on Build GRCh38
rs748685338 Chr.2: 156512358-156512358 on Build GRCh38
rs200713191 Chr.2: 156512255-156512255 on Build GRCh38
rs913955943 Chr.2: 156512097-156512097 on Build GRCh38
rs763184741 Chr.2: 156512261-156512261 on Build GRCh38
rs778833866 Chr.2: 156512227-156512227 on Build GRCh38
rs775064429 Chr.2: 156512094-156512094 on Build GRCh38
rs968552697 Chr.2: 156512028-156512028 on Build GRCh38
rs776175414 Chr.2: 156512352-156512352 on Build GRCh38
rs770238795 Chr.2: 156512359-156512359 on Build GRCh38
rs1007780478 Chr.2: 156512398-156512398 on Build GRCh38
rs201559833 Chr.2: 156512251-156512251 on Build GRCh38
rs369349969 Chr.2: 156512364-156512364 on Build GRCh38
rs762285348 Chr.2: 156512248-156512248 on Build GRCh38
rs887959518 Chr.2: 156512162-156512162 on Build GRCh38
rs1036924319 Chr.2: 156512220-156512220 on Build GRCh38
rs746433826 Chr.2: 156512240-156512240 on Build GRCh38
rs114815323 Chr.2: 156512312-156512312 on Build GRCh38
rs201153701 Chr.2: 156512246-156512246 on Build GRCh38
rs141392427 Chr.2: 156512228-156512228 on Build GRCh38
rs1045290679 Chr.2: 156512155-156512155 on Build GRCh38
rs190634369 Chr.2: 156512341-156512341 on Build GRCh38
rs763379883 Chr.2: 156512144-156512144 on Build GRCh38
rs374101429 Chr.2: 156512365-156512365 on Build GRCh38
rs772465746 Chr.2: 156512171-156512173 on Build GRCh38
rs768097688 Chr.2: 156512349-156512349 on Build GRCh38
rs531051841 Chr.2: 156512250-156512250 on Build GRCh38
rs958253927 Chr.2: 156512468-156512468 on Build GRCh38
rs542361341 Chr.2: 156512116-156512116 on Build GRCh38
rs992541375 Chr.2: 156512392-156512392 on Build GRCh38
rs751664830 Chr.2: 156512294-156512294 on Build GRCh38
rs751544316 Chr.2: 156512175-156512175 on Build GRCh38
rs764326515 Chr.2: 156512172-156512172 on Build GRCh38
rs563535638 Chr.2: 156512136-156512136 on Build GRCh38
rs200768161 Chr.2: 156512257-156512257 on Build GRCh38
rs1011488078 Chr.2: 156512496-156512496 on Build GRCh38
rs753879898 Chr.2: 156512315-156512315 on Build GRCh38
rs779637238 Chr.2: 156512335-156512335 on Build GRCh38
rs749247587 Chr.2: 156512212-156512212 on Build GRCh38
rs745596992 Chr.2: 156512324-156512324 on Build GRCh38
rs766512303 Chr.2: 156512173-156512173 on Build GRCh38
rs777060448 Chr.2: 156512245-156512245 on Build GRCh38
rs775077056 Chr.2: 156512238-156512238 on Build GRCh38
rs755083234 Chr.2: 156512305-156512305 on Build GRCh38
rs777507245 Chr.2: 156512178-156512178 on Build GRCh38
rs756093375 Chr.2: 156512177-156512177 on Build GRCh38
rs771797900 Chr.2: 156512236-156512236 on Build GRCh38
rs376581357 Chr.2: 156512332-156512332 on Build GRCh38
rs547094265 Chr.2: 156512369-156512369 on Build GRCh38
rs762200156 Chr.2: 156512402-156512402 on Build GRCh38
rs924382667 Chr.2: 156512052-156512052 on Build GRCh38
rs938400431 Chr.2: 156512067-156512067 on Build GRCh38
rs565728219 Chr.2: 156512370-156512370 on Build GRCh38
rs369276284 Chr.2: 156512322-156512322 on Build GRCh38

View Full Product Details