Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19859476-19860054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPRC5B
Entrez Gene ID
51704
Gene Name
G protein-coupled receptor class C group 5 member B
Gene Aliases
RAIG-2, RAIG2
UniGene
Hs.148685
Gene Chromosome Location
Chr.16: 19858971-19886045 on Build GRCh38
Transcripts
NM_001304771.1 NM_016235.2 XM_006721051.2 XM_006721052.2

SNP Detail

SNP ID Chromosome Location
rs61661380 Chr.16: 19859594-19859594 on Build GRCh38
rs185176462 Chr.16: 19859807-19859807 on Build GRCh38
rs1053801361 Chr.16: 19859789-19859789 on Build GRCh38
rs949355487 Chr.16: 19859717-19859717 on Build GRCh38
rs868773648 Chr.16: 19860018-19860018 on Build GRCh38
rs1018382239 Chr.16: 19859675-19859675 on Build GRCh38
rs974562545 Chr.16: 19859720-19859720 on Build GRCh38
rs959706877 Chr.16: 19860011-19860011 on Build GRCh38
rs1000344869 Chr.16: 19859536-19859536 on Build GRCh38
rs893974593 Chr.16: 19859513-19859513 on Build GRCh38
rs545999123 Chr.16: 19859593-19859593 on Build GRCh38
rs1006687508 Chr.16: 19859662-19859662 on Build GRCh38
rs564837212 Chr.16: 19859552-19859552 on Build GRCh38
rs779243876 Chr.16: 19859984-19859984 on Build GRCh38
rs996321573 Chr.16: 19859551-19859551 on Build GRCh38
rs990643752 Chr.16: 19859678-19859678 on Build GRCh38
rs990958944 Chr.16: 19859985-19859985 on Build GRCh38
rs1015175845 Chr.16: 19859918-19859918 on Build GRCh38
rs935300728 Chr.16: 19859784-19859784 on Build GRCh38
rs1050643466 Chr.16: 19859827-19859827 on Build GRCh38
rs905577291 Chr.16: 19859522-19859523 on Build GRCh38
rs970529185 Chr.16: 19860017-19860017 on Build GRCh38
rs533635563 Chr.16: 19859655-19859655 on Build GRCh38
rs1018148708 Chr.16: 19859958-19859964 on Build GRCh38
rs951447375 Chr.16: 19859753-19859753 on Build GRCh38
rs918145798 Chr.16: 19859777-19859777 on Build GRCh38
rs775798162 Chr.16: 19859610-19859610 on Build GRCh38
rs550420148 Chr.16: 19859928-19859928 on Build GRCh38
rs772151812 Chr.16: 19859965-19859965 on Build GRCh38
rs996762551 Chr.16: 19859809-19859809 on Build GRCh38
rs992383841 Chr.16: 19859776-19859776 on Build GRCh38
rs1009192164 Chr.16: 19859891-19859891 on Build GRCh38
rs764042283 Chr.16: 19859888-19859888 on Build GRCh38
rs918620517 Chr.16: 19859730-19859730 on Build GRCh38
rs952523782 Chr.16: 19859968-19859968 on Build GRCh38
rs575316605 Chr.16: 19859587-19859587 on Build GRCh38
rs891011237 Chr.16: 19859929-19859929 on Build GRCh38
rs878875729 Chr.16: 19859885-19859885 on Build GRCh38
rs142574439 Chr.16: 19859644-19859644 on Build GRCh38
rs761197392 Chr.16: 19859947-19859947 on Build GRCh38
rs1006807146 Chr.16: 19859948-19859948 on Build GRCh38
rs541440724 Chr.16: 19859697-19859697 on Build GRCh38
rs948226453 Chr.16: 19859781-19859781 on Build GRCh38
rs962845729 Chr.16: 19859967-19859967 on Build GRCh38
rs923979789 Chr.16: 19859780-19859780 on Build GRCh38
rs138089050 Chr.16: 19859566-19859566 on Build GRCh38
rs139556727 Chr.16: 19860006-19860006 on Build GRCh38
rs954954790 Chr.16: 19859553-19859553 on Build GRCh38
rs150554245 Chr.16: 19859768-19859768 on Build GRCh38
rs961037274 Chr.16: 19859927-19859927 on Build GRCh38
rs765550764 Chr.16: 19859565-19859565 on Build GRCh38

View Full Product Details