Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97929869-97930454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEMGN
Entrez Gene ID
55363
Gene Name
hemogen
Gene Aliases
CT155, EDAG, EDAG-1, NDR
UniGene
Hs.176626
Gene Chromosome Location
Chr.9: 97926791-97944915 on Build GRCh38
Transcripts
NM_018437.4 NM_197978.2 XM_005252086.1 XM_011518845.2 XM_011518846.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4519910 c.1033G>A p.E345K Chr.9: 97930362-97930362 on Build GRCh38
311749 c.1225T>C p.S409P Chr.9: 97930170-97930170 on Build GRCh38
4564601 c.1299_1300CC>TT p.Q434* Chr.9: 97930095-97930096 on Build GRCh38
6114624 c.1281C>G p.H427Q Chr.9: 97930114-97930114 on Build GRCh38
3432691 c.1261G>A p.E421K Chr.9: 97930134-97930134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536606309 Chr.9: 97930284-97930284 on Build GRCh38
rs79474766 Chr.9: 97929955-97929956 on Build GRCh38
rs146429876 Chr.9: 97930052-97930052 on Build GRCh38
rs770110338 Chr.9: 97930324-97930324 on Build GRCh38
rs761007719 Chr.9: 97930072-97930072 on Build GRCh38
rs778394985 Chr.9: 97929994-97929994 on Build GRCh38
rs766851402 Chr.9: 97930350-97930350 on Build GRCh38
rs901077192 Chr.9: 97929901-97929901 on Build GRCh38
rs367714883 Chr.9: 97930186-97930186 on Build GRCh38
rs149135993 Chr.9: 97929955-97929955 on Build GRCh38
rs904915318 Chr.9: 97929944-97929944 on Build GRCh38
rs570241773 Chr.9: 97930401-97930401 on Build GRCh38
rs201694228 Chr.9: 97930356-97930356 on Build GRCh38
rs760803544 Chr.9: 97930214-97930214 on Build GRCh38
rs2436 Chr.9: 97930148-97930148 on Build GRCh38
rs749261670 Chr.9: 97930190-97930190 on Build GRCh38
rs1023148387 Chr.9: 97929939-97929939 on Build GRCh38
rs776525041 Chr.9: 97930017-97930017 on Build GRCh38
rs752376629 Chr.9: 97930386-97930386 on Build GRCh38
rs773628693 Chr.9: 97930341-97930341 on Build GRCh38
rs772599187 Chr.9: 97930046-97930046 on Build GRCh38
rs150918405 Chr.9: 97930391-97930391 on Build GRCh38
rs760601222 Chr.9: 97930381-97930382 on Build GRCh38
rs775680618 Chr.9: 97930203-97930203 on Build GRCh38
rs757800981 Chr.9: 97929989-97929989 on Build GRCh38
rs770055325 Chr.9: 97930429-97930429 on Build GRCh38
rs150117068 Chr.9: 97930163-97930163 on Build GRCh38
rs138098420 Chr.9: 97930348-97930348 on Build GRCh38
rs759043789 Chr.9: 97930361-97930361 on Build GRCh38
rs763976396 Chr.9: 97930396-97930396 on Build GRCh38
rs752528459 Chr.9: 97930103-97930103 on Build GRCh38
rs768985827 Chr.9: 97930215-97930215 on Build GRCh38
rs35223978 Chr.9: 97930048-97930048 on Build GRCh38
rs756056267 Chr.9: 97930240-97930240 on Build GRCh38
rs2435 Chr.9: 97930112-97930112 on Build GRCh38
rs991092279 Chr.9: 97930256-97930256 on Build GRCh38
rs78169548 Chr.9: 97929957-97929957 on Build GRCh38
rs202170428 Chr.9: 97930426-97930426 on Build GRCh38
rs375380681 Chr.9: 97930302-97930302 on Build GRCh38
rs1016714176 Chr.9: 97930096-97930096 on Build GRCh38
rs547815344 Chr.9: 97929920-97929920 on Build GRCh38
rs951106705 Chr.9: 97930022-97930022 on Build GRCh38
rs140657626 Chr.9: 97930055-97930055 on Build GRCh38
rs369622151 Chr.9: 97930311-97930311 on Build GRCh38
rs569904796 Chr.9: 97930318-97930318 on Build GRCh38
rs776735078 Chr.9: 97930237-97930237 on Build GRCh38
rs369459043 Chr.9: 97930410-97930410 on Build GRCh38
rs779661691 Chr.9: 97930114-97930114 on Build GRCh38
rs780835769 Chr.9: 97930185-97930185 on Build GRCh38
rs750904964 Chr.9: 97930244-97930244 on Build GRCh38
rs764334865 Chr.9: 97930092-97930092 on Build GRCh38
rs746694096 Chr.9: 97930012-97930012 on Build GRCh38
rs201229826 Chr.9: 97930232-97930232 on Build GRCh38
rs149270294 Chr.9: 97930293-97930293 on Build GRCh38
rs769910210 Chr.9: 97930038-97930038 on Build GRCh38
rs768310083 Chr.9: 97930014-97930014 on Build GRCh38
rs748415690 Chr.9: 97930323-97930323 on Build GRCh38
rs762297706 Chr.9: 97930101-97930101 on Build GRCh38
rs757852013 Chr.9: 97930107-97930107 on Build GRCh38
rs765624587 Chr.9: 97930242-97930242 on Build GRCh38
rs975186685 Chr.9: 97930146-97930146 on Build GRCh38
rs750936346 Chr.9: 97930104-97930104 on Build GRCh38
rs757921129 Chr.9: 97930000-97930000 on Build GRCh38
rs777811108 Chr.9: 97930187-97930187 on Build GRCh38
rs968205258 Chr.9: 97930270-97930270 on Build GRCh38
rs138552503 Chr.9: 97930281-97930281 on Build GRCh38
rs757019453 Chr.9: 97930405-97930405 on Build GRCh38
rs776446278 Chr.9: 97930393-97930397 on Build GRCh38
rs758753533 Chr.9: 97930315-97930315 on Build GRCh38
rs745826816 Chr.9: 97930411-97930411 on Build GRCh38
rs202184558 Chr.9: 97930260-97930260 on Build GRCh38
rs750515468 Chr.9: 97930300-97930300 on Build GRCh38
rs745444718 Chr.9: 97929998-97929998 on Build GRCh38
rs1054465754 Chr.9: 97930333-97930333 on Build GRCh38
rs772258055 Chr.9: 97930202-97930202 on Build GRCh38
rs754723328 Chr.9: 97930176-97930176 on Build GRCh38
rs748030198 Chr.9: 97930028-97930028 on Build GRCh38
rs749208146 Chr.9: 97930308-97930308 on Build GRCh38
rs774427335 Chr.9: 97930125-97930125 on Build GRCh38
rs1013052384 Chr.9: 97929930-97929930 on Build GRCh38
rs779617495 Chr.9: 97930007-97930007 on Build GRCh38
rs149708018 Chr.9: 97929954-97929954 on Build GRCh38
rs35527067 Chr.9: 97930366-97930366 on Build GRCh38
rs770967896 Chr.9: 97930309-97930309 on Build GRCh38
rs146990988 Chr.9: 97930316-97930316 on Build GRCh38
rs200368619 Chr.9: 97930043-97930043 on Build GRCh38
rs770793656 Chr.9: 97930384-97930385 on Build GRCh38
rs752427462 Chr.9: 97930290-97930290 on Build GRCh38
rs372858440 Chr.9: 97930417-97930417 on Build GRCh38
rs754299125 Chr.9: 97930098-97930098 on Build GRCh38
rs778904356 Chr.9: 97930285-97930285 on Build GRCh38

View Full Product Details