Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114096058-114096636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF12
Entrez Gene ID
113220
Gene Name
kinesin family member 12
Gene Aliases
-
UniGene
Hs.28149
Gene Chromosome Location
Chr.9: 114091623-114100099 on Build GRCh38
Transcripts
NM_138424.1 XM_005251683.4 XM_006716947.2 XM_011518190.2 XM_011518191.2 XM_011518192.2 XM_011518193.2 XM_017014240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4879490 c.837G>T p.L279L Chr.9: 114096094-114096094 on Build GRCh38
6536826 c.393G>C p.L131L Chr.9: 114096139-114096139 on Build GRCh38
7416248 c.298C>A p.L100I Chr.9: 114096413-114096413 on Build GRCh38
4392466 c.443T>C p.L148P Chr.9: 114096089-114096089 on Build GRCh38
7224029 c.632-3C>T p.? Chr.9: 114096481-114096481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898556545 Chr.9: 114096533-114096533 on Build GRCh38
rs369433012 Chr.9: 114096117-114096117 on Build GRCh38
rs774296266 Chr.9: 114096399-114096399 on Build GRCh38
rs369824310 Chr.9: 114096246-114096246 on Build GRCh38
rs748426864 Chr.9: 114096379-114096379 on Build GRCh38
rs1030971963 Chr.9: 114096104-114096104 on Build GRCh38
rs958592096 Chr.9: 114096274-114096274 on Build GRCh38
rs759008470 Chr.9: 114096405-114096405 on Build GRCh38
rs543250349 Chr.9: 114096609-114096609 on Build GRCh38
rs557804117 Chr.9: 114096272-114096272 on Build GRCh38
rs778073032 Chr.9: 114096489-114096489 on Build GRCh38
rs747909415 Chr.9: 114096245-114096245 on Build GRCh38
rs751637318 Chr.9: 114096364-114096364 on Build GRCh38
rs905737489 Chr.9: 114096540-114096540 on Build GRCh38
rs754145219 Chr.9: 114096206-114096206 on Build GRCh38
rs1004424893 Chr.9: 114096509-114096509 on Build GRCh38
rs763811977 Chr.9: 114096426-114096426 on Build GRCh38
rs891282842 Chr.9: 114096497-114096497 on Build GRCh38
rs751214754 Chr.9: 114096191-114096191 on Build GRCh38
rs747088229 Chr.9: 114096230-114096230 on Build GRCh38
rs188880158 Chr.9: 114096279-114096279 on Build GRCh38
rs758491443 Chr.9: 114096221-114096221 on Build GRCh38
rs918224378 Chr.9: 114096297-114096297 on Build GRCh38
rs745913192 Chr.9: 114096099-114096099 on Build GRCh38
rs145670325 Chr.9: 114096469-114096469 on Build GRCh38
rs763396255 Chr.9: 114096428-114096428 on Build GRCh38
rs755137105 Chr.9: 114096373-114096373 on Build GRCh38
rs959779204 Chr.9: 114096295-114096295 on Build GRCh38
rs765859441 Chr.9: 114096183-114096183 on Build GRCh38
rs751412095 Chr.9: 114096296-114096296 on Build GRCh38
rs781529941 Chr.9: 114096377-114096377 on Build GRCh38
rs757588159 Chr.9: 114096353-114096353 on Build GRCh38
rs779303654 Chr.9: 114096098-114096098 on Build GRCh38
rs942859870 Chr.9: 114096354-114096354 on Build GRCh38
rs779436408 Chr.9: 114096212-114096212 on Build GRCh38
rs771610677 Chr.9: 114096408-114096408 on Build GRCh38
rs770719763 Chr.9: 114096397-114096397 on Build GRCh38
rs374176491 Chr.9: 114096228-114096228 on Build GRCh38
rs893790745 Chr.9: 114096453-114096453 on Build GRCh38
rs754695309 Chr.9: 114096199-114096199 on Build GRCh38
rs35480554 Chr.9: 114096446-114096446 on Build GRCh38
rs780623448 Chr.9: 114096362-114096362 on Build GRCh38
rs748990609 Chr.9: 114096510-114096510 on Build GRCh38
rs770228178 Chr.9: 114096112-114096112 on Build GRCh38
rs773786026 Chr.9: 114096337-114096337 on Build GRCh38
rs762491064 Chr.9: 114096179-114096179 on Build GRCh38
rs749115456 Chr.9: 114096395-114096395 on Build GRCh38
rs775102998 Chr.9: 114096413-114096413 on Build GRCh38
rs747292364 Chr.9: 114096111-114096111 on Build GRCh38
rs35142355 Chr.9: 114096104-114096105 on Build GRCh38
rs761711856 Chr.9: 114096115-114096115 on Build GRCh38
rs149715619 Chr.9: 114096092-114096092 on Build GRCh38
rs955569797 Chr.9: 114096079-114096079 on Build GRCh38
rs750693411 Chr.9: 114096427-114096427 on Build GRCh38
rs762860143 Chr.9: 114096248-114096248 on Build GRCh38
rs752149678 Chr.9: 114096437-114096437 on Build GRCh38
rs757914815 Chr.9: 114096448-114096449 on Build GRCh38
rs766752937 Chr.9: 114096432-114096432 on Build GRCh38
rs777012024 Chr.9: 114096241-114096241 on Build GRCh38
rs752967226 Chr.9: 114096466-114096466 on Build GRCh38
rs758762647 Chr.9: 114096357-114096357 on Build GRCh38
rs931584699 Chr.9: 114096333-114096333 on Build GRCh38
rs767586205 Chr.9: 114096438-114096438 on Build GRCh38
rs772075259 Chr.9: 114096101-114096101 on Build GRCh38
rs769621798 Chr.9: 114096393-114096393 on Build GRCh38
rs6478062 Chr.9: 114096345-114096345 on Build GRCh38
rs760310016 Chr.9: 114096416-114096416 on Build GRCh38
rs995567431 Chr.9: 114096539-114096539 on Build GRCh38
rs1035496464 Chr.9: 114096605-114096605 on Build GRCh38
rs750936933 Chr.9: 114096356-114096356 on Build GRCh38
rs977570676 Chr.9: 114096391-114096391 on Build GRCh38
rs778172314 Chr.9: 114096082-114096082 on Build GRCh38
rs775758437 Chr.9: 114096106-114096106 on Build GRCh38
rs757696767 Chr.9: 114096094-114096094 on Build GRCh38
rs781476794 Chr.9: 114096449-114096449 on Build GRCh38
rs369984638 Chr.9: 114096355-114096355 on Build GRCh38
rs184997854 Chr.9: 114096231-114096231 on Build GRCh38
rs765912638 Chr.9: 114096258-114096258 on Build GRCh38
rs926638974 Chr.9: 114096260-114096260 on Build GRCh38
rs773050639 Chr.9: 114096134-114096134 on Build GRCh38
rs757643289 Chr.9: 114096209-114096209 on Build GRCh38
rs984319440 Chr.9: 114096344-114096344 on Build GRCh38
rs746402976 Chr.9: 114096215-114096215 on Build GRCh38
rs767109936 Chr.9: 114096342-114096342 on Build GRCh38
rs767165166 Chr.9: 114096201-114096201 on Build GRCh38
rs529548284 Chr.9: 114096599-114096599 on Build GRCh38
rs752330031 Chr.9: 114096384-114096385 on Build GRCh38
rs564040906 Chr.9: 114096394-114096394 on Build GRCh38
rs750300616 Chr.9: 114096580-114096580 on Build GRCh38
rs759063430 Chr.9: 114096339-114096339 on Build GRCh38

View Full Product Details