Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 175812026-175812578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPM6A
Entrez Gene ID
2823
Gene Name
glycoprotein M6A
Gene Aliases
GPM6, M6A
UniGene
Hs.75819
Gene Chromosome Location
Chr.4: 175632937-176002691 on Build GRCh38
Transcripts
NM_001261447.1 NM_005277.4 NM_201591.2 NM_201592.2 NR_048571.1 XM_011531877.1 NR_125901.1

Gene Symbol
LOC101928590
Entrez Gene ID
101928590
Gene Name
uncharacterized LOC101928590
Gene Aliases
-
UniGene
Hs.599383
Gene Chromosome Location
Chr.4: 175790307-175812189 on Build GRCh38
Transcripts
NM_001261447.1 NM_005277.4 NM_201591.2 NM_201592.2 NR_048571.1 XM_011531877.1 NR_125901.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
176821 c.9G>T p.E3D Chr.4: 175812219-175812219 on Build GRCh38
1237149 c.15G>T p.M5I Chr.4: 175812213-175812213 on Build GRCh38
3696357 c.24A>G p.G8G Chr.4: 175812204-175812204 on Build GRCh38
4429662 c.7G>T p.E3* Chr.4: 175812221-175812221 on Build GRCh38
5883980 c.33A>C p.Q11H Chr.4: 175812195-175812195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748348508 Chr.4: 175812297-175812297 on Build GRCh38
rs11732859 Chr.4: 175812126-175812126 on Build GRCh38
rs1031274299 Chr.4: 175812320-175812320 on Build GRCh38
rs11943437 Chr.4: 175812408-175812408 on Build GRCh38
rs543299110 Chr.4: 175812151-175812151 on Build GRCh38
rs934086402 Chr.4: 175812093-175812093 on Build GRCh38
rs747853754 Chr.4: 175812275-175812275 on Build GRCh38
rs779771658 Chr.4: 175812233-175812233 on Build GRCh38
rs976058417 Chr.4: 175812340-175812340 on Build GRCh38
rs878967127 Chr.4: 175812304-175812304 on Build GRCh38
rs1049505859 Chr.4: 175812186-175812186 on Build GRCh38
rs925074973 Chr.4: 175812118-175812118 on Build GRCh38
rs750402106 Chr.4: 175812143-175812143 on Build GRCh38
rs894747598 Chr.4: 175812145-175812145 on Build GRCh38
rs11934159 Chr.4: 175812381-175812381 on Build GRCh38
rs992139396 Chr.4: 175812486-175812486 on Build GRCh38
rs922580819 Chr.4: 175812059-175812059 on Build GRCh38
rs11732930 Chr.4: 175812238-175812238 on Build GRCh38
rs998601431 Chr.4: 175812181-175812181 on Build GRCh38
rs796982772 Chr.4: 175812299-175812300 on Build GRCh38
rs1005728687 Chr.4: 175812119-175812119 on Build GRCh38
rs752899483 Chr.4: 175812166-175812166 on Build GRCh38
rs745963666 Chr.4: 175812461-175812461 on Build GRCh38
rs945264014 Chr.4: 175812049-175812049 on Build GRCh38
rs1033705846 Chr.4: 175812451-175812451 on Build GRCh38
rs1017460716 Chr.4: 175812324-175812324 on Build GRCh38
rs534772446 Chr.4: 175812077-175812077 on Build GRCh38
rs1038549981 Chr.4: 175812311-175812311 on Build GRCh38
rs892900869 Chr.4: 175812321-175812321 on Build GRCh38
rs1038906775 Chr.4: 175812138-175812138 on Build GRCh38
rs974546003 Chr.4: 175812546-175812546 on Build GRCh38
rs964897454 Chr.4: 175812325-175812325 on Build GRCh38
rs370231380 Chr.4: 175812274-175812274 on Build GRCh38
rs762568746 Chr.4: 175812299-175812299 on Build GRCh38
rs779248511 Chr.4: 175812187-175812187 on Build GRCh38
rs887935022 Chr.4: 175812428-175812428 on Build GRCh38
rs774528465 Chr.4: 175812214-175812214 on Build GRCh38
rs113129787 Chr.4: 175812301-175812301 on Build GRCh38
rs908632540 Chr.4: 175812308-175812308 on Build GRCh38
rs906911897 Chr.4: 175812315-175812315 on Build GRCh38
rs1013558438 Chr.4: 175812157-175812157 on Build GRCh38
rs771294727 Chr.4: 175812286-175812286 on Build GRCh38
rs1008144624 Chr.4: 175812302-175812302 on Build GRCh38
rs922635270 Chr.4: 175812305-175812305 on Build GRCh38
rs201180260 Chr.4: 175812301-175812302 on Build GRCh38
rs1036669248 Chr.4: 175812412-175812412 on Build GRCh38
rs62338073 Chr.4: 175812299-175812299 on Build GRCh38
rs943674567 Chr.4: 175812278-175812278 on Build GRCh38
rs941484860 Chr.4: 175812466-175812466 on Build GRCh38
rs561449470 Chr.4: 175812549-175812549 on Build GRCh38
rs528170008 Chr.4: 175812417-175812417 on Build GRCh38
rs933969788 Chr.4: 175812306-175812306 on Build GRCh38
rs768360312 Chr.4: 175812256-175812256 on Build GRCh38
rs545497960 Chr.4: 175812361-175812361 on Build GRCh38
rs182265357 Chr.4: 175812391-175812391 on Build GRCh38
rs375299132 Chr.4: 175812297-175812299 on Build GRCh38
rs765030076 Chr.4: 175812141-175812141 on Build GRCh38
rs34321288 Chr.4: 175812347-175812347 on Build GRCh38
rs1030259340 Chr.4: 175812384-175812384 on Build GRCh38
rs998485209 Chr.4: 175812316-175812316 on Build GRCh38
rs746428584 Chr.4: 175812203-175812203 on Build GRCh38
rs773444755 Chr.4: 175812206-175812207 on Build GRCh38
rs367587263 Chr.4: 175812174-175812174 on Build GRCh38
rs1046403469 Chr.4: 175812314-175812314 on Build GRCh38
rs1011728176 Chr.4: 175812322-175812322 on Build GRCh38
rs554341666 Chr.4: 175812092-175812092 on Build GRCh38
rs1002283886 Chr.4: 175812430-175812430 on Build GRCh38
rs70962418 Chr.4: 175812340-175812341 on Build GRCh38
rs941421443 Chr.4: 175812310-175812310 on Build GRCh38
rs758961367 Chr.4: 175812300-175812301 on Build GRCh38
rs894623734 Chr.4: 175812312-175812312 on Build GRCh38
rs541985196 Chr.4: 175812490-175812490 on Build GRCh38
rs769899164 Chr.4: 175812300-175812300 on Build GRCh38
rs868550867 Chr.4: 175812460-175812460 on Build GRCh38
rs982802166 Chr.4: 175812457-175812457 on Build GRCh38
rs916120862 Chr.4: 175812555-175812555 on Build GRCh38
rs574334319 Chr.4: 175812099-175812099 on Build GRCh38
rs139008815 Chr.4: 175812283-175812283 on Build GRCh38
rs767681711 Chr.4: 175812161-175812161 on Build GRCh38
rs910347992 Chr.4: 175812282-175812282 on Build GRCh38
rs949023338 Chr.4: 175812313-175812313 on Build GRCh38
rs746657097 Chr.4: 175812254-175812254 on Build GRCh38
rs979292672 Chr.4: 175812087-175812087 on Build GRCh38
rs75902123 Chr.4: 175812250-175812250 on Build GRCh38
rs1027828532 Chr.4: 175812427-175812427 on Build GRCh38
rs982744724 Chr.4: 175812307-175812307 on Build GRCh38
rs772307406 Chr.4: 175812440-175812457 on Build GRCh38
rs371268534 Chr.4: 175812175-175812175 on Build GRCh38
rs898093474 Chr.4: 175812415-175812415 on Build GRCh38
rs749856489 Chr.4: 175812104-175812104 on Build GRCh38
rs370293687 Chr.4: 175812532-175812532 on Build GRCh38
rs762981651 Chr.4: 175812269-175812269 on Build GRCh38
rs776439674 Chr.4: 175812259-175812259 on Build GRCh38
rs878964996 Chr.4: 175812299-175812302 on Build GRCh38
rs879251110 Chr.4: 175812303-175812303 on Build GRCh38

View Full Product Details