Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 153676870-153677304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIA1
Entrez Gene ID
2890
Gene Name
glutamate ionotropic receptor AMPA type subunit 1
Gene Aliases
GLUH1, GLUR1, GLURA, GluA1, HBGR1
UniGene
Hs.519693
Gene Chromosome Location
Chr.5: 153489615-153813873 on Build GRCh38
Transcripts
NM_000827.3 NM_001114183.1 NM_001258019.1 NM_001258020.1 NM_001258021.1 NM_001258022.1 NM_001258023.1 NR_047578.1 XM_011537635.2 XM_017009392.1 XM_017009393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7079555 c.930G>C p.Q310H Chr.5: 153677062-153677062 on Build GRCh38
449152 c.873G>A p.A291A Chr.5: 153677005-153677005 on Build GRCh38
737144 c.961G>C p.G321R Chr.5: 153677093-153677093 on Build GRCh38
3975344 c.1001G>A p.G334E Chr.5: 153677133-153677133 on Build GRCh38
3245304 c.1018G>T p.A340S Chr.5: 153677150-153677150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747812130 Chr.5: 153677003-153677003 on Build GRCh38
rs113653577 Chr.5: 153676946-153676946 on Build GRCh38
rs753904191 Chr.5: 153676986-153676986 on Build GRCh38
rs767745327 Chr.5: 153676949-153676949 on Build GRCh38
rs1019760547 Chr.5: 153677270-153677270 on Build GRCh38
rs908373142 Chr.5: 153676948-153676948 on Build GRCh38
rs754969941 Chr.5: 153677205-153677205 on Build GRCh38
rs759960256 Chr.5: 153677053-153677053 on Build GRCh38
rs563194672 Chr.5: 153677054-153677054 on Build GRCh38
rs965140568 Chr.5: 153677276-153677276 on Build GRCh38
rs1033154959 Chr.5: 153676940-153676940 on Build GRCh38
rs977225720 Chr.5: 153676932-153676932 on Build GRCh38
rs774967854 Chr.5: 153676986-153676986 on Build GRCh38
rs200078037 Chr.5: 153677002-153677002 on Build GRCh38
rs368839980 Chr.5: 153676988-153676988 on Build GRCh38
rs778853422 Chr.5: 153677081-153677081 on Build GRCh38
rs746887407 Chr.5: 153676989-153676989 on Build GRCh38
rs770035156 Chr.5: 153677275-153677275 on Build GRCh38
rs894096028 Chr.5: 153677058-153677058 on Build GRCh38
rs967607219 Chr.5: 153676898-153676898 on Build GRCh38
rs770544535 Chr.5: 153677134-153677134 on Build GRCh38
rs374559725 Chr.5: 153677174-153677174 on Build GRCh38
rs769455691 Chr.5: 153677107-153677107 on Build GRCh38
rs768560984 Chr.5: 153676999-153676999 on Build GRCh38
rs376405457 Chr.5: 153677200-153677200 on Build GRCh38
rs998626508 Chr.5: 153677216-153677216 on Build GRCh38
rs778012531 Chr.5: 153676972-153676972 on Build GRCh38
rs752606801 Chr.5: 153677212-153677212 on Build GRCh38
rs572816757 Chr.5: 153676890-153676890 on Build GRCh38
rs899755269 Chr.5: 153677246-153677246 on Build GRCh38
rs772640709 Chr.5: 153677167-153677167 on Build GRCh38
rs557600394 Chr.5: 153676938-153676938 on Build GRCh38
rs758300485 Chr.5: 153676987-153676987 on Build GRCh38
rs867097904 Chr.5: 153677039-153677039 on Build GRCh38
rs768979604 Chr.5: 153677079-153677079 on Build GRCh38
rs761926140 Chr.5: 153677194-153677194 on Build GRCh38
rs776029254 Chr.5: 153677169-153677169 on Build GRCh38
rs746403999 Chr.5: 153677148-153677148 on Build GRCh38
rs1055119824 Chr.5: 153677049-153677049 on Build GRCh38
rs142974388 Chr.5: 153677127-153677127 on Build GRCh38
rs774966581 Chr.5: 153677047-153677047 on Build GRCh38
rs533521208 Chr.5: 153676997-153676997 on Build GRCh38
rs767390298 Chr.5: 153677209-153677209 on Build GRCh38
rs372429427 Chr.5: 153677029-153677029 on Build GRCh38
rs188078784 Chr.5: 153676963-153676963 on Build GRCh38
rs1039694504 Chr.5: 153677203-153677203 on Build GRCh38
rs756395599 Chr.5: 153676970-153676970 on Build GRCh38
rs957076667 Chr.5: 153676954-153676954 on Build GRCh38
rs769255856 Chr.5: 153676968-153676971 on Build GRCh38
rs764372850 Chr.5: 153677078-153677078 on Build GRCh38
rs866330059 Chr.5: 153677095-153677095 on Build GRCh38
rs760982517 Chr.5: 153677055-153677055 on Build GRCh38
rs1048294193 Chr.5: 153676939-153676939 on Build GRCh38
rs762474670 Chr.5: 153677021-153677021 on Build GRCh38
rs146697277 Chr.5: 153677086-153677086 on Build GRCh38
rs774584155 Chr.5: 153677118-153677118 on Build GRCh38
rs745977119 Chr.5: 153677120-153677120 on Build GRCh38
rs754814414 Chr.5: 153677082-153677082 on Build GRCh38
rs114632468 Chr.5: 153677014-153677014 on Build GRCh38
rs750402597 Chr.5: 153677196-153677196 on Build GRCh38
rs888505854 Chr.5: 153677180-153677180 on Build GRCh38
rs773723701 Chr.5: 153677140-153677140 on Build GRCh38
rs764532419 Chr.5: 153677181-153677181 on Build GRCh38
rs749706041 Chr.5: 153677005-153677005 on Build GRCh38

View Full Product Details