Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21481950-21482533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNE
Entrez Gene ID
338376
Gene Name
interferon epsilon
Gene Aliases
IFN-E, IFNE1, IFNT1, INFE1, PRO655
UniGene
Hs.682604
Gene Chromosome Location
Chr.9: 21480839-21482313 on Build GRCh38
Transcripts
NM_176891.4 NR_027054.1

Gene Symbol
MIR31HG
Entrez Gene ID
554202
Gene Name
MIR31 host gene
Gene Aliases
-
UniGene
Hs.458096
Gene Chromosome Location
Chr.9: 21454268-21559698 on Build GRCh38
Transcripts
NM_176891.4 NR_027054.1

SNP Detail

SNP ID Chromosome Location
rs996186433 Chr.9: 21482223-21482223 on Build GRCh38
rs140795364 Chr.9: 21482186-21482186 on Build GRCh38
rs933487098 Chr.9: 21482134-21482134 on Build GRCh38
rs772796412 Chr.9: 21482321-21482321 on Build GRCh38
rs1010039652 Chr.9: 21482013-21482013 on Build GRCh38
rs1029279727 Chr.9: 21482333-21482333 on Build GRCh38
rs10811549 Chr.9: 21482352-21482352 on Build GRCh38
rs1001436179 Chr.9: 21481980-21481980 on Build GRCh38
rs3028326 Chr.9: 21482241-21482243 on Build GRCh38
rs974731721 Chr.9: 21482420-21482420 on Build GRCh38
rs748911051 Chr.9: 21482229-21482229 on Build GRCh38
rs780758192 Chr.9: 21482502-21482502 on Build GRCh38
rs397819775 Chr.9: 21482127-21482128 on Build GRCh38
rs771634754 Chr.9: 21482418-21482418 on Build GRCh38
rs540477357 Chr.9: 21482002-21482002 on Build GRCh38
rs1056027918 Chr.9: 21482156-21482156 on Build GRCh38
rs1008252831 Chr.9: 21482253-21482253 on Build GRCh38
rs2039380 Chr.9: 21482182-21482182 on Build GRCh38
rs979369387 Chr.9: 21482099-21482099 on Build GRCh38
rs966734233 Chr.9: 21482039-21482039 on Build GRCh38
rs923872569 Chr.9: 21482344-21482344 on Build GRCh38
rs917574533 Chr.9: 21482393-21482393 on Build GRCh38
rs941800891 Chr.9: 21482473-21482473 on Build GRCh38
rs370999269 Chr.9: 21482433-21482433 on Build GRCh38
rs907630712 Chr.9: 21482151-21482151 on Build GRCh38
rs371686766 Chr.9: 21482243-21482243 on Build GRCh38
rs895697444 Chr.9: 21482377-21482377 on Build GRCh38
rs34814436 Chr.9: 21482126-21482127 on Build GRCh38
rs979785458 Chr.9: 21482342-21482342 on Build GRCh38
rs761927191 Chr.9: 21482150-21482150 on Build GRCh38
rs905526052 Chr.9: 21482244-21482244 on Build GRCh38
rs551226982 Chr.9: 21482258-21482258 on Build GRCh38
rs1019726880 Chr.9: 21481987-21481987 on Build GRCh38
rs758924360 Chr.9: 21482400-21482400 on Build GRCh38
rs975513397 Chr.9: 21482330-21482330 on Build GRCh38
rs75140720 Chr.9: 21482410-21482410 on Build GRCh38
rs182964858 Chr.9: 21482078-21482078 on Build GRCh38
rs113164481 Chr.9: 21482476-21482476 on Build GRCh38
rs548554615 Chr.9: 21482180-21482180 on Build GRCh38
rs925267883 Chr.9: 21482089-21482089 on Build GRCh38
rs144694040 Chr.9: 21482401-21482401 on Build GRCh38
rs1021473144 Chr.9: 21482022-21482022 on Build GRCh38
rs34533918 Chr.9: 21482240-21482242 on Build GRCh38
rs775976504 Chr.9: 21482009-21482009 on Build GRCh38
rs964130052 Chr.9: 21482294-21482294 on Build GRCh38
rs908019193 Chr.9: 21482462-21482462 on Build GRCh38
rs947772952 Chr.9: 21482206-21482206 on Build GRCh38
rs1045675485 Chr.9: 21482213-21482213 on Build GRCh38
rs1037396008 Chr.9: 21482235-21482235 on Build GRCh38
rs397894161 Chr.9: 21482243-21482245 on Build GRCh38
rs1035570045 Chr.9: 21481981-21481981 on Build GRCh38
rs201968293 Chr.9: 21482124-21482125 on Build GRCh38
rs967888338 Chr.9: 21482334-21482334 on Build GRCh38
rs185302379 Chr.9: 21482398-21482398 on Build GRCh38
rs545444801 Chr.9: 21482442-21482442 on Build GRCh38
rs923513387 Chr.9: 21482133-21482133 on Build GRCh38
rs10125074 Chr.9: 21482090-21482090 on Build GRCh38
rs373745100 Chr.9: 21482245-21482245 on Build GRCh38
rs571770831 Chr.9: 21482335-21482335 on Build GRCh38
rs545953852 Chr.9: 21482492-21482492 on Build GRCh38
rs1020101227 Chr.9: 21482399-21482399 on Build GRCh38
rs1047867184 Chr.9: 21482135-21482135 on Build GRCh38
rs974956761 Chr.9: 21482103-21482103 on Build GRCh38
rs755339651 Chr.9: 21482454-21482454 on Build GRCh38
rs954955056 Chr.9: 21482449-21482449 on Build GRCh38
rs964690163 Chr.9: 21482413-21482413 on Build GRCh38
rs974981781 Chr.9: 21482065-21482065 on Build GRCh38

View Full Product Details