Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32138399-32138966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPD1L
Entrez Gene ID
23171
Gene Name
glycerol-3-phosphate dehydrogenase 1-like
Gene Aliases
GPD1-L
UniGene
Hs.82432
Gene Chromosome Location
Chr.3: 32106511-32168715 on Build GRCh38
Transcripts
NM_015141.3 XM_006713068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7167370 c.309C>G p.I103M Chr.3: 32138670-32138670 on Build GRCh38
6097488 c.335C>T p.P112L Chr.3: 32138696-32138696 on Build GRCh38
5160748 c.366+8G>A p.? Chr.3: 32138735-32138735 on Build GRCh38
7155492 c.247G>A p.E83K Chr.3: 32138608-32138608 on Build GRCh38
6544301 c.262G>T p.A88S Chr.3: 32138623-32138623 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774974692 Chr.3: 32138667-32138667 on Build GRCh38
rs779901962 Chr.3: 32138771-32138771 on Build GRCh38
rs991849621 Chr.3: 32138740-32138740 on Build GRCh38
rs751700398 Chr.3: 32138721-32138721 on Build GRCh38
rs935769817 Chr.3: 32138840-32138840 on Build GRCh38
rs924413141 Chr.3: 32138834-32138834 on Build GRCh38
rs764507257 Chr.3: 32138587-32138587 on Build GRCh38
rs774085519 Chr.3: 32138548-32138548 on Build GRCh38
rs997573890 Chr.3: 32138922-32138922 on Build GRCh38
rs750846725 Chr.3: 32138598-32138598 on Build GRCh38
rs72552292 Chr.3: 32138608-32138608 on Build GRCh38
rs568004238 Chr.3: 32138734-32138734 on Build GRCh38
rs745770005 Chr.3: 32138654-32138654 on Build GRCh38
rs770277485 Chr.3: 32138685-32138685 on Build GRCh38
rs756016432 Chr.3: 32138540-32138540 on Build GRCh38
rs767879744 Chr.3: 32138738-32138738 on Build GRCh38
rs771610382 Chr.3: 32138659-32138659 on Build GRCh38
rs150334706 Chr.3: 32138868-32138868 on Build GRCh38
rs112709405 Chr.3: 32138473-32138473 on Build GRCh38
rs374305150 Chr.3: 32138681-32138681 on Build GRCh38
rs750440738 Chr.3: 32138752-32138752 on Build GRCh38
rs769167911 Chr.3: 32138928-32138928 on Build GRCh38
rs532757147 Chr.3: 32138842-32138842 on Build GRCh38
rs1007778712 Chr.3: 32138881-32138881 on Build GRCh38
rs949859402 Chr.3: 32138518-32138518 on Build GRCh38
rs1017232651 Chr.3: 32138671-32138671 on Build GRCh38
rs977211295 Chr.3: 32138519-32138519 on Build GRCh38
rs778927711 Chr.3: 32138639-32138639 on Build GRCh38
rs765805642 Chr.3: 32138597-32138597 on Build GRCh38
rs368838349 Chr.3: 32138622-32138622 on Build GRCh38
rs377378878 Chr.3: 32138607-32138607 on Build GRCh38
rs116185919 Chr.3: 32138545-32138545 on Build GRCh38
rs113776126 Chr.3: 32138809-32138809 on Build GRCh38
rs901549626 Chr.3: 32138905-32138905 on Build GRCh38
rs111570801 Chr.3: 32138878-32138878 on Build GRCh38
rs776108577 Chr.3: 32138692-32138692 on Build GRCh38
rs768304305 Chr.3: 32138690-32138690 on Build GRCh38
rs569738824 Chr.3: 32138861-32138861 on Build GRCh38
rs372917584 Chr.3: 32138761-32138761 on Build GRCh38
rs760287995 Chr.3: 32138585-32138585 on Build GRCh38
rs746721040 Chr.3: 32138688-32138688 on Build GRCh38
rs576121998 Chr.3: 32138753-32138753 on Build GRCh38
rs962057022 Chr.3: 32138444-32138444 on Build GRCh38
rs1050672695 Chr.3: 32138917-32138917 on Build GRCh38
rs915985296 Chr.3: 32138535-32138535 on Build GRCh38
rs776114036 Chr.3: 32138741-32138741 on Build GRCh38
rs768277076 Chr.3: 32138882-32138884 on Build GRCh38
rs979205112 Chr.3: 32138795-32138795 on Build GRCh38
rs527946229 Chr.3: 32138423-32138423 on Build GRCh38
rs932325334 Chr.3: 32138885-32138885 on Build GRCh38
rs1032101167 Chr.3: 32138936-32138936 on Build GRCh38
rs936017308 Chr.3: 32138496-32138496 on Build GRCh38
rs149167213 Chr.3: 32138706-32138706 on Build GRCh38
rs141876129 Chr.3: 32138835-32138836 on Build GRCh38
rs762018216 Chr.3: 32138593-32138593 on Build GRCh38
rs758316011 Chr.3: 32138764-32138764 on Build GRCh38
rs757123438 Chr.3: 32138745-32138745 on Build GRCh38
rs924341488 Chr.3: 32138528-32138528 on Build GRCh38
rs1036500693 Chr.3: 32138933-32138933 on Build GRCh38
rs747068747 Chr.3: 32138489-32138489 on Build GRCh38
rs888002540 Chr.3: 32138647-32138647 on Build GRCh38
rs762262112 Chr.3: 32138695-32138695 on Build GRCh38
rs1045837412 Chr.3: 32138848-32138848 on Build GRCh38
rs909609625 Chr.3: 32138841-32138841 on Build GRCh38
rs199693644 Chr.3: 32138513-32138513 on Build GRCh38
rs771522425 Chr.3: 32138562-32138562 on Build GRCh38
rs768331840 Chr.3: 32138641-32138641 on Build GRCh38
rs751900212 Chr.3: 32138613-32138613 on Build GRCh38
rs932226827 Chr.3: 32138564-32138564 on Build GRCh38
rs991905342 Chr.3: 32138826-32138826 on Build GRCh38
rs897968981 Chr.3: 32138934-32138934 on Build GRCh38
rs72546650 Chr.3: 32138735-32138735 on Build GRCh38
rs1009017741 Chr.3: 32138616-32138616 on Build GRCh38
rs775170888 Chr.3: 32138580-32138580 on Build GRCh38
rs879420060 Chr.3: 32138799-32138799 on Build GRCh38
rs777069543 Chr.3: 32138592-32138592 on Build GRCh38
rs10627424 Chr.3: 32138468-32138469 on Build GRCh38
rs944904483 Chr.3: 32138930-32138930 on Build GRCh38
rs140207438 Chr.3: 32138430-32138430 on Build GRCh38
rs181959523 Chr.3: 32138833-32138833 on Build GRCh38
rs563810749 Chr.3: 32138509-32138509 on Build GRCh38
rs543054841 Chr.3: 32138561-32138561 on Build GRCh38
rs766627054 Chr.3: 32138717-32138717 on Build GRCh38
rs878855004 Chr.3: 32138606-32138606 on Build GRCh38
rs763553263 Chr.3: 32138710-32138710 on Build GRCh38
rs551149293 Chr.3: 32138858-32138858 on Build GRCh38
rs1049236893 Chr.3: 32138870-32138870 on Build GRCh38
rs371030358 Chr.3: 32138746-32138746 on Build GRCh38
rs991383513 Chr.3: 32138494-32138494 on Build GRCh38
rs770912333 Chr.3: 32138547-32138547 on Build GRCh38
rs777708856 Chr.3: 32138542-32138542 on Build GRCh38
rs1039687230 Chr.3: 32138881-32138881 on Build GRCh38
rs890689154 Chr.3: 32138919-32138919 on Build GRCh38
rs781503701 Chr.3: 32138621-32138621 on Build GRCh38
rs765497074 Chr.3: 32138705-32138705 on Build GRCh38
rs374241561 Chr.3: 32138478-32138478 on Build GRCh38
rs755240955 Chr.3: 32138618-32138618 on Build GRCh38
rs1007252272 Chr.3: 32138652-32138652 on Build GRCh38
rs528792876 Chr.3: 32138625-32138625 on Build GRCh38
rs776329795 Chr.3: 32138735-32138751 on Build GRCh38
rs979356980 Chr.3: 32138463-32138463 on Build GRCh38
rs971245412 Chr.3: 32138804-32138804 on Build GRCh38
rs888033588 Chr.3: 32138873-32138873 on Build GRCh38
rs994902269 Chr.3: 32138693-32138693 on Build GRCh38

View Full Product Details