Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 226735280-226735843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRS1
Entrez Gene ID
3667
Gene Name
insulin receptor substrate 1
Gene Aliases
HIRS-1
UniGene
Hs.471508
Gene Chromosome Location
Chr.2: 226731317-226798790 on Build GRCh38
Transcripts
NM_005544.2

SNP Detail

SNP ID Chromosome Location
rs13306464 Chr.2: 226735589-226735589 on Build GRCh38
rs944453852 Chr.2: 226735523-226735523 on Build GRCh38
rs931610146 Chr.2: 226735545-226735545 on Build GRCh38
rs1014540970 Chr.2: 226735353-226735353 on Build GRCh38
rs188099215 Chr.2: 226735647-226735647 on Build GRCh38
rs985471748 Chr.2: 226735475-226735475 on Build GRCh38
rs547979511 Chr.2: 226735560-226735560 on Build GRCh38
rs139228892 Chr.2: 226735408-226735408 on Build GRCh38
rs537809110 Chr.2: 226735743-226735743 on Build GRCh38
rs751718728 Chr.2: 226735365-226735366 on Build GRCh38
rs967726691 Chr.2: 226735407-226735407 on Build GRCh38
rs999197333 Chr.2: 226735335-226735335 on Build GRCh38
rs1049343329 Chr.2: 226735757-226735757 on Build GRCh38
rs569816731 Chr.2: 226735466-226735466 on Build GRCh38
rs906292933 Chr.2: 226735670-226735670 on Build GRCh38
rs934958781 Chr.2: 226735692-226735692 on Build GRCh38
rs760611394 Chr.2: 226735648-226735648 on Build GRCh38
rs918815850 Chr.2: 226735539-226735539 on Build GRCh38
rs919447160 Chr.2: 226735590-226735590 on Build GRCh38
rs183768589 Chr.2: 226735613-226735613 on Build GRCh38
rs930773793 Chr.2: 226735747-226735747 on Build GRCh38
rs559973247 Chr.2: 226735584-226735584 on Build GRCh38
rs896559143 Chr.2: 226735315-226735315 on Build GRCh38
rs965322167 Chr.2: 226735483-226735483 on Build GRCh38
rs1031304274 Chr.2: 226735383-226735383 on Build GRCh38
rs893709585 Chr.2: 226735753-226735753 on Build GRCh38
rs1012036209 Chr.2: 226735311-226735311 on Build GRCh38
rs748250973 Chr.2: 226735504-226735504 on Build GRCh38
rs1052128155 Chr.2: 226735728-226735728 on Build GRCh38
rs756354711 Chr.2: 226735698-226735698 on Build GRCh38
rs911670691 Chr.2: 226735472-226735472 on Build GRCh38
rs562327159 Chr.2: 226735363-226735363 on Build GRCh38
rs1032852416 Chr.2: 226735432-226735432 on Build GRCh38
rs916788438 Chr.2: 226735581-226735581 on Build GRCh38
rs144208373 Chr.2: 226735802-226735802 on Build GRCh38
rs972671348 Chr.2: 226735569-226735569 on Build GRCh38
rs549756409 Chr.2: 226735397-226735397 on Build GRCh38
rs34286846 Chr.2: 226735484-226735485 on Build GRCh38
rs1020096498 Chr.2: 226735355-226735355 on Build GRCh38
rs1046144446 Chr.2: 226735617-226735617 on Build GRCh38
rs570459331 Chr.2: 226735712-226735712 on Build GRCh38
rs948204321 Chr.2: 226735610-226735614 on Build GRCh38
rs1019666704 Chr.2: 226735480-226735480 on Build GRCh38
rs953059591 Chr.2: 226735456-226735456 on Build GRCh38

View Full Product Details