Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 99254623-99255222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR18
Entrez Gene ID
2841
Gene Name
G protein-coupled receptor 18
Gene Aliases
-
UniGene
Hs.741589
Gene Chromosome Location
Chr.13: 99254713-99262498 on Build GRCh38
Transcripts
NM_001098200.1 NM_005292.3 XM_006719946.3 NM_001144072.1 NM_177967.3 NR_026644.1 XM_006719948.3 XM_011521082.2 XM_011521083.2 XM_011521084.2

Gene Symbol
UBAC2
Entrez Gene ID
337867
Gene Name
UBA domain containing 2
Gene Aliases
PHGDHL1
UniGene
Hs.508545
Gene Chromosome Location
Chr.13: 99200425-99386499 on Build GRCh38
Transcripts
NM_001098200.1 NM_005292.3 XM_006719946.3 NM_001144072.1 NM_177967.3 NR_026644.1 XM_006719948.3 XM_011521082.2 XM_011521083.2 XM_011521084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4049028 c.931C>T p.R311C Chr.13: 99254942-99254942 on Build GRCh38
6674003 c.727G>A p.V243I Chr.13: 99255146-99255146 on Build GRCh38
6674004 c.864C>T p.I288I Chr.13: 99255009-99255009 on Build GRCh38
6674001 c.932G>A p.R311H Chr.13: 99254941-99254941 on Build GRCh38
6674000 c.834C>T p.S278S Chr.13: 99255039-99255039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374003672 Chr.13: 99254674-99254674 on Build GRCh38
rs759164125 Chr.13: 99254918-99254918 on Build GRCh38
rs758115946 Chr.13: 99254832-99254836 on Build GRCh38
rs966118456 Chr.13: 99254754-99254754 on Build GRCh38
rs766783804 Chr.13: 99254997-99254997 on Build GRCh38
rs750126175 Chr.13: 99254827-99254830 on Build GRCh38
rs780622512 Chr.13: 99254850-99254850 on Build GRCh38
rs146455146 Chr.13: 99254819-99254819 on Build GRCh38
rs555756794 Chr.13: 99254896-99254896 on Build GRCh38
rs1051577 Chr.13: 99255044-99255044 on Build GRCh38
rs758993071 Chr.13: 99254833-99254833 on Build GRCh38
rs574195979 Chr.13: 99254973-99254973 on Build GRCh38
rs750387704 Chr.13: 99255172-99255172 on Build GRCh38
rs780958175 Chr.13: 99255059-99255059 on Build GRCh38
rs752017507 Chr.13: 99254840-99254840 on Build GRCh38
rs1007690420 Chr.13: 99255056-99255056 on Build GRCh38
rs77539420 Chr.13: 99254737-99254737 on Build GRCh38
rs780161923 Chr.13: 99254885-99254885 on Build GRCh38
rs371765611 Chr.13: 99254738-99254738 on Build GRCh38
rs768365665 Chr.13: 99255112-99255112 on Build GRCh38
rs764766723 Chr.13: 99254835-99254835 on Build GRCh38
rs773712081 Chr.13: 99254990-99254990 on Build GRCh38
rs372086125 Chr.13: 99254899-99254899 on Build GRCh38
rs764458382 Chr.13: 99255147-99255147 on Build GRCh38
rs148795491 Chr.13: 99255104-99255104 on Build GRCh38
rs780396822 Chr.13: 99254954-99254954 on Build GRCh38
rs746959412 Chr.13: 99255037-99255037 on Build GRCh38
rs567962939 Chr.13: 99254898-99254898 on Build GRCh38
rs769289500 Chr.13: 99254933-99254934 on Build GRCh38
rs532887631 Chr.13: 99254789-99254789 on Build GRCh38
rs768620730 Chr.13: 99254892-99254892 on Build GRCh38
rs1002470610 Chr.13: 99254653-99254653 on Build GRCh38
rs372413471 Chr.13: 99255132-99255132 on Build GRCh38
rs369995186 Chr.13: 99255182-99255182 on Build GRCh38
rs757282650 Chr.13: 99255040-99255040 on Build GRCh38
rs758638842 Chr.13: 99255092-99255092 on Build GRCh38
rs762471452 Chr.13: 99254987-99254987 on Build GRCh38
rs754481541 Chr.13: 99254759-99254759 on Build GRCh38
rs893213158 Chr.13: 99254955-99254955 on Build GRCh38
rs781659489 Chr.13: 99254864-99254864 on Build GRCh38
rs540457420 Chr.13: 99255077-99255077 on Build GRCh38
rs745721043 Chr.13: 99255063-99255063 on Build GRCh38
rs773963673 Chr.13: 99255116-99255116 on Build GRCh38
rs777880406 Chr.13: 99254966-99254966 on Build GRCh38
rs781694906 Chr.13: 99255074-99255075 on Build GRCh38
rs763554698 Chr.13: 99254989-99254989 on Build GRCh38
rs140769802 Chr.13: 99255146-99255146 on Build GRCh38
rs747319192 Chr.13: 99254925-99254925 on Build GRCh38
rs201584591 Chr.13: 99255082-99255082 on Build GRCh38
rs142437132 Chr.13: 99255106-99255106 on Build GRCh38
rs61736137 Chr.13: 99254986-99254986 on Build GRCh38
rs74498945 Chr.13: 99254817-99254817 on Build GRCh38
rs981933685 Chr.13: 99254742-99254742 on Build GRCh38
rs755906704 Chr.13: 99255103-99255103 on Build GRCh38
rs150932589 Chr.13: 99255136-99255136 on Build GRCh38
rs765605928 Chr.13: 99255015-99255015 on Build GRCh38
rs776181586 Chr.13: 99254912-99254912 on Build GRCh38
rs748873391 Chr.13: 99255071-99255071 on Build GRCh38
rs767096993 Chr.13: 99254941-99254941 on Build GRCh38
rs752922229 Chr.13: 99255022-99255022 on Build GRCh38
rs968649799 Chr.13: 99254729-99254729 on Build GRCh38
rs1031301958 Chr.13: 99254667-99254667 on Build GRCh38
rs759244925 Chr.13: 99254982-99254982 on Build GRCh38
rs750822967 Chr.13: 99254873-99254873 on Build GRCh38
rs773252636 Chr.13: 99254902-99254902 on Build GRCh38
rs202049006 Chr.13: 99254908-99254908 on Build GRCh38
rs59722139 Chr.13: 99254964-99254964 on Build GRCh38
rs762670401 Chr.13: 99254700-99254700 on Build GRCh38
rs1004013686 Chr.13: 99255009-99255009 on Build GRCh38
rs773837678 Chr.13: 99255028-99255028 on Build GRCh38
rs755286750 Chr.13: 99254962-99254962 on Build GRCh38
rs140120491 Chr.13: 99255061-99255061 on Build GRCh38
rs771435262 Chr.13: 99255125-99255125 on Build GRCh38
rs777935684 Chr.13: 99255034-99255034 on Build GRCh38
rs761185561 Chr.13: 99254996-99254996 on Build GRCh38
rs572780975 Chr.13: 99254712-99254712 on Build GRCh38
rs775154268 Chr.13: 99255070-99255070 on Build GRCh38
rs1002833847 Chr.13: 99255153-99255153 on Build GRCh38
rs765661875 Chr.13: 99255084-99255084 on Build GRCh38
rs903298208 Chr.13: 99254657-99254660 on Build GRCh38
rs150612287 Chr.13: 99254926-99254926 on Build GRCh38
rs3742130 Chr.13: 99255087-99255087 on Build GRCh38
rs769523403 Chr.13: 99254985-99254985 on Build GRCh38
rs140955383 Chr.13: 99254961-99254961 on Build GRCh38
rs1052716348 Chr.13: 99254967-99254967 on Build GRCh38
rs773906701 Chr.13: 99255078-99255078 on Build GRCh38
rs762415931 Chr.13: 99254928-99254928 on Build GRCh38
rs1033077432 Chr.13: 99254703-99254703 on Build GRCh38
rs60331560 Chr.13: 99254678-99254678 on Build GRCh38
rs766951793 Chr.13: 99255079-99255079 on Build GRCh38
rs779000601 Chr.13: 99254963-99254963 on Build GRCh38
rs374416624 Chr.13: 99254942-99254942 on Build GRCh38
rs758585811 Chr.13: 99255033-99255033 on Build GRCh38
rs770714276 Chr.13: 99254911-99254911 on Build GRCh38
rs199762287 Chr.13: 99255140-99255140 on Build GRCh38
rs769423816 Chr.13: 99255067-99255067 on Build GRCh38
rs761980710 Chr.13: 99255166-99255166 on Build GRCh38
rs777229237 Chr.13: 99255127-99255127 on Build GRCh38
rs757646007 Chr.13: 99255117-99255117 on Build GRCh38
rs768167198 Chr.13: 99255075-99255075 on Build GRCh38
rs147474658 Chr.13: 99254970-99254970 on Build GRCh38
rs755267322 Chr.13: 99255011-99255011 on Build GRCh38
rs756448167 Chr.13: 99254878-99254878 on Build GRCh38
rs376966351 Chr.13: 99255165-99255165 on Build GRCh38
rs202154292 Chr.13: 99254887-99254887 on Build GRCh38
rs143074994 Chr.13: 99255096-99255096 on Build GRCh38
rs984822888 Chr.13: 99255121-99255121 on Build GRCh38
rs756466021 Chr.13: 99254944-99254944 on Build GRCh38
rs367603189 Chr.13: 99254969-99254969 on Build GRCh38
rs767428640 Chr.13: 99255171-99255171 on Build GRCh38
rs1051247626 Chr.13: 99254654-99254654 on Build GRCh38
rs754297101 Chr.13: 99255008-99255008 on Build GRCh38
rs1049474617 Chr.13: 99254824-99254824 on Build GRCh38
rs757815333 Chr.13: 99254859-99254859 on Build GRCh38
rs193218266 Chr.13: 99255097-99255097 on Build GRCh38

View Full Product Details