Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84122101-84122699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSDL1
Entrez Gene ID
83693
Gene Name
hydroxysteroid dehydrogenase like 1
Gene Aliases
SDR12C3
UniGene
Hs.555992
Gene Chromosome Location
Chr.16: 84122139-84145195 on Build GRCh38
Transcripts
NM_001146051.1 NM_031463.4 XM_005256189.2

SNP Detail

SNP ID Chromosome Location
rs1007996096 Chr.16: 84122202-84122202 on Build GRCh38
rs1045796489 Chr.16: 84122323-84122323 on Build GRCh38
rs752580900 Chr.16: 84122473-84122473 on Build GRCh38
rs1019364473 Chr.16: 84122516-84122516 on Build GRCh38
rs189704956 Chr.16: 84122331-84122331 on Build GRCh38
rs961190165 Chr.16: 84122540-84122540 on Build GRCh38
rs765814387 Chr.16: 84122265-84122265 on Build GRCh38
rs184063793 Chr.16: 84122296-84122296 on Build GRCh38
rs980651634 Chr.16: 84122637-84122637 on Build GRCh38
rs147520975 Chr.16: 84122667-84122667 on Build GRCh38
rs764184693 Chr.16: 84122650-84122650 on Build GRCh38
rs536997350 Chr.16: 84122447-84122447 on Build GRCh38
rs953764515 Chr.16: 84122161-84122161 on Build GRCh38
rs1011679581 Chr.16: 84122217-84122218 on Build GRCh38
rs1017457317 Chr.16: 84122389-84122389 on Build GRCh38
rs146600846 Chr.16: 84122379-84122379 on Build GRCh38
rs540043846 Chr.16: 84122577-84122577 on Build GRCh38
rs550710280 Chr.16: 84122154-84122154 on Build GRCh38
rs1011628671 Chr.16: 84122371-84122371 on Build GRCh38
rs903677351 Chr.16: 84122374-84122374 on Build GRCh38
rs948899015 Chr.16: 84122624-84122624 on Build GRCh38
rs374319864 Chr.16: 84122251-84122265 on Build GRCh38
rs916040098 Chr.16: 84122576-84122576 on Build GRCh38
rs191129238 Chr.16: 84122269-84122269 on Build GRCh38
rs1026679145 Chr.16: 84122564-84122564 on Build GRCh38
rs928721916 Chr.16: 84122642-84122642 on Build GRCh38
rs550615496 Chr.16: 84122340-84122340 on Build GRCh38
rs111273677 Chr.16: 84122332-84122332 on Build GRCh38
rs533242242 Chr.16: 84122677-84122677 on Build GRCh38
rs764961432 Chr.16: 84122349-84122349 on Build GRCh38
rs572386990 Chr.16: 84122368-84122368 on Build GRCh38
rs1049562889 Chr.16: 84122424-84122424 on Build GRCh38
rs553318131 Chr.16: 84122589-84122589 on Build GRCh38
rs1053078119 Chr.16: 84122365-84122365 on Build GRCh38
rs1007833684 Chr.16: 84122381-84122381 on Build GRCh38
rs751462541 Chr.16: 84122657-84122657 on Build GRCh38
rs554069616 Chr.16: 84122238-84122238 on Build GRCh38
rs972457987 Chr.16: 84122252-84122252 on Build GRCh38
rs893025832 Chr.16: 84122360-84122360 on Build GRCh38
rs1021756835 Chr.16: 84122372-84122372 on Build GRCh38
rs773699812 Chr.16: 84122229-84122229 on Build GRCh38
rs994218881 Chr.16: 84122551-84122551 on Build GRCh38
rs891859828 Chr.16: 84122201-84122201 on Build GRCh38
rs531441713 Chr.16: 84122363-84122364 on Build GRCh38
rs948936197 Chr.16: 84122344-84122344 on Build GRCh38
rs984499447 Chr.16: 84122548-84122548 on Build GRCh38
rs975980502 Chr.16: 84122453-84122453 on Build GRCh38
rs149584051 Chr.16: 84122289-84122289 on Build GRCh38
rs948782296 Chr.16: 84122309-84122309 on Build GRCh38
rs916092371 Chr.16: 84122293-84122293 on Build GRCh38
rs543083932 Chr.16: 84122237-84122251 on Build GRCh38
rs76974140 Chr.16: 84122377-84122377 on Build GRCh38
rs776168957 Chr.16: 84122320-84122320 on Build GRCh38
rs186800236 Chr.16: 84122237-84122237 on Build GRCh38
rs914592927 Chr.16: 84122652-84122652 on Build GRCh38
rs1022101373 Chr.16: 84122218-84122218 on Build GRCh38
rs879081793 Chr.16: 84122364-84122364 on Build GRCh38
rs546051694 Chr.16: 84122399-84122399 on Build GRCh38
rs369362104 Chr.16: 84122390-84122390 on Build GRCh38
rs139198253 Chr.16: 84122463-84122463 on Build GRCh38
rs878881380 Chr.16: 84122317-84122317 on Build GRCh38
rs536339953 Chr.16: 84122486-84122486 on Build GRCh38
rs376448102 Chr.16: 84122622-84122622 on Build GRCh38
rs75622403 Chr.16: 84122375-84122375 on Build GRCh38
rs1045084067 Chr.16: 84122128-84122128 on Build GRCh38
rs553134617 Chr.16: 84122645-84122645 on Build GRCh38
rs981610930 Chr.16: 84122641-84122641 on Build GRCh38
rs534946643 Chr.16: 84122359-84122359 on Build GRCh38
rs984509125 Chr.16: 84122330-84122330 on Build GRCh38
rs575239126 Chr.16: 84122653-84122653 on Build GRCh38
rs985077820 Chr.16: 84122571-84122571 on Build GRCh38
rs780191507 Chr.16: 84122200-84122200 on Build GRCh38
rs952366679 Chr.16: 84122567-84122567 on Build GRCh38
rs11448771 Chr.16: 84122364-84122365 on Build GRCh38
rs1031741196 Chr.16: 84122176-84122176 on Build GRCh38
rs570416874 Chr.16: 84122519-84122519 on Build GRCh38
rs143223859 Chr.16: 84122647-84122647 on Build GRCh38
rs758263068 Chr.16: 84122534-84122534 on Build GRCh38
rs542157285 Chr.16: 84122643-84122643 on Build GRCh38
rs1058444 Chr.16: 84122675-84122675 on Build GRCh38
rs1011823201 Chr.16: 84122153-84122153 on Build GRCh38
rs573084984 Chr.16: 84122612-84122612 on Build GRCh38

View Full Product Details