Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83970596-83971160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPK
Entrez Gene ID
3190
Gene Name
heterogeneous nuclear ribonucleoprotein K
Gene Aliases
AUKS, CSBP, HNRPK, TUNP
UniGene
Hs.522257
Gene Chromosome Location
Chr.9: 83968083-83980782 on Build GRCh38
Transcripts
NM_001318186.1 NM_001318187.1 NM_001318188.1 NM_002140.4 NM_031262.3 NM_031263.3 XM_005251960.2 XM_005251963.3 XM_005251965.2 XM_011518616.1 XM_017014668.1 XM_017014669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5816466 c.1154G>T p.G385V Chr.9: 83970774-83970774 on Build GRCh38
6268155 c.1129C>A p.R377S Chr.9: 83970799-83970799 on Build GRCh38
6272503 c.1098C>A p.G366G Chr.9: 83970907-83970907 on Build GRCh38
5899578 c.1129C>T p.R377C Chr.9: 83970799-83970799 on Build GRCh38
5712507 c.1114T>G p.S372A Chr.9: 83970814-83970814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980772629 Chr.9: 83970680-83970680 on Build GRCh38
rs750511980 Chr.9: 83971009-83971009 on Build GRCh38
rs760749429 Chr.9: 83970883-83970887 on Build GRCh38
rs755597367 Chr.9: 83970716-83970716 on Build GRCh38
rs1046783823 Chr.9: 83971065-83971065 on Build GRCh38
rs942829364 Chr.9: 83970781-83970781 on Build GRCh38
rs777424249 Chr.9: 83970717-83970717 on Build GRCh38
rs751523268 Chr.9: 83970709-83970709 on Build GRCh38
rs746255704 Chr.9: 83970922-83970922 on Build GRCh38
rs199855906 Chr.9: 83970839-83970839 on Build GRCh38
rs181141777 Chr.9: 83970788-83970788 on Build GRCh38
rs754012600 Chr.9: 83970921-83970924 on Build GRCh38
rs747681079 Chr.9: 83970715-83970715 on Build GRCh38
rs767301453 Chr.9: 83970954-83970954 on Build GRCh38
rs757027678 Chr.9: 83970848-83970848 on Build GRCh38
rs555489265 Chr.9: 83971065-83971066 on Build GRCh38
rs535382871 Chr.9: 83971115-83971115 on Build GRCh38
rs971594875 Chr.9: 83970674-83970674 on Build GRCh38
rs907991268 Chr.9: 83970795-83970795 on Build GRCh38
rs775667809 Chr.9: 83970995-83970996 on Build GRCh38
rs535402341 Chr.9: 83971051-83971051 on Build GRCh38
rs767555294 Chr.9: 83970965-83970965 on Build GRCh38
rs748898288 Chr.9: 83970720-83970720 on Build GRCh38
rs760830318 Chr.9: 83970686-83970686 on Build GRCh38
rs3214660 Chr.9: 83970643-83970644 on Build GRCh38
rs751370155 Chr.9: 83970852-83970852 on Build GRCh38
rs542204841 Chr.9: 83970981-83970981 on Build GRCh38
rs201503271 Chr.9: 83970726-83970726 on Build GRCh38
rs755404355 Chr.9: 83970927-83970927 on Build GRCh38
rs373830271 Chr.9: 83970934-83970934 on Build GRCh38
rs539096532 Chr.9: 83970860-83970860 on Build GRCh38
rs535426539 Chr.9: 83970673-83970673 on Build GRCh38
rs755668461 Chr.9: 83970951-83970951 on Build GRCh38
rs1034339468 Chr.9: 83970681-83970681 on Build GRCh38
rs955411944 Chr.9: 83971034-83971034 on Build GRCh38
rs774517631 Chr.9: 83970768-83970768 on Build GRCh38
rs138398623 Chr.9: 83970950-83970950 on Build GRCh38
rs775885766 Chr.9: 83970930-83970930 on Build GRCh38
rs533667672 Chr.9: 83970998-83970998 on Build GRCh38
rs779676856 Chr.9: 83970703-83970703 on Build GRCh38
rs942969486 Chr.9: 83971042-83971042 on Build GRCh38
rs759269585 Chr.9: 83970952-83970952 on Build GRCh38
rs758293796 Chr.9: 83970702-83970702 on Build GRCh38
rs772519430 Chr.9: 83970789-83970789 on Build GRCh38
rs767805806 Chr.9: 83970723-83970724 on Build GRCh38
rs575068026 Chr.9: 83970966-83970966 on Build GRCh38
rs922441667 Chr.9: 83970812-83970812 on Build GRCh38
rs952164009 Chr.9: 83970682-83970682 on Build GRCh38
rs775395655 Chr.9: 83970696-83970697 on Build GRCh38
rs763018954 Chr.9: 83970634-83970634 on Build GRCh38
rs143712714 Chr.9: 83970809-83970809 on Build GRCh38
rs761971516 Chr.9: 83970937-83970937 on Build GRCh38
rs1054128827 Chr.9: 83970868-83970868 on Build GRCh38
rs764223631 Chr.9: 83970810-83970810 on Build GRCh38
rs150482206 Chr.9: 83970881-83970881 on Build GRCh38
rs755988059 Chr.9: 83970955-83970955 on Build GRCh38
rs184361719 Chr.9: 83970621-83970621 on Build GRCh38
rs754814694 Chr.9: 83970710-83970710 on Build GRCh38
rs1014941728 Chr.9: 83970989-83970989 on Build GRCh38
rs747482479 Chr.9: 83970931-83970931 on Build GRCh38
rs952077638 Chr.9: 83970969-83970969 on Build GRCh38
rs776032208 Chr.9: 83970792-83970792 on Build GRCh38
rs779441775 Chr.9: 83970964-83970965 on Build GRCh38
rs762893180 Chr.9: 83970718-83970723 on Build GRCh38
rs375704482 Chr.9: 83970802-83970802 on Build GRCh38
rs371386785 Chr.9: 83970728-83970728 on Build GRCh38
rs549757398 Chr.9: 83971048-83971048 on Build GRCh38
rs779773668 Chr.9: 83971109-83971109 on Build GRCh38
rs369684059 Chr.9: 83970892-83970892 on Build GRCh38
rs756956721 Chr.9: 83970904-83970904 on Build GRCh38
rs568410384 Chr.9: 83971063-83971063 on Build GRCh38
rs1021377727 Chr.9: 83970960-83970960 on Build GRCh38
rs755650634 Chr.9: 83970880-83970880 on Build GRCh38
rs766355814 Chr.9: 83970885-83970886 on Build GRCh38
rs188765983 Chr.9: 83970697-83970697 on Build GRCh38
rs933899928 Chr.9: 83970837-83970837 on Build GRCh38
rs922410404 Chr.9: 83971027-83971027 on Build GRCh38
rs750677424 Chr.9: 83970724-83970725 on Build GRCh38
rs759644134 Chr.9: 83970774-83970776 on Build GRCh38
rs766209942 Chr.9: 83970843-83970843 on Build GRCh38
rs781177925 Chr.9: 83970714-83970714 on Build GRCh38
rs11140312 Chr.9: 83970861-83970861 on Build GRCh38
rs911167301 Chr.9: 83971067-83971067 on Build GRCh38
rs772457802 Chr.9: 83970925-83970925 on Build GRCh38
rs1006188137 Chr.9: 83970973-83970973 on Build GRCh38
rs397687683 Chr.9: 83970646-83970647 on Build GRCh38
rs1010396279 Chr.9: 83970638-83970638 on Build GRCh38
rs1035049912 Chr.9: 83970967-83970967 on Build GRCh38
rs751422995 Chr.9: 83970719-83970719 on Build GRCh38
rs1039987289 Chr.9: 83971043-83971043 on Build GRCh38
rs573127876 Chr.9: 83971035-83971035 on Build GRCh38
rs189442295 Chr.9: 83971013-83971013 on Build GRCh38
rs764001747 Chr.9: 83970964-83970964 on Build GRCh38
rs560688238 Chr.9: 83970997-83970997 on Build GRCh38
rs140741394 Chr.9: 83970668-83970668 on Build GRCh38
rs748953133 Chr.9: 83970888-83970888 on Build GRCh38
rs771289643 Chr.9: 83970745-83970745 on Build GRCh38
rs968957166 Chr.9: 83970962-83970962 on Build GRCh38
rs184462197 Chr.9: 83970856-83970856 on Build GRCh38
rs148820425 Chr.9: 83970999-83970999 on Build GRCh38
rs920067416 Chr.9: 83971045-83971045 on Build GRCh38
rs777028296 Chr.9: 83970835-83970835 on Build GRCh38

View Full Product Details