Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194397757-194398356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GP5
Entrez Gene ID
2814
Gene Name
glycoprotein V platelet
Gene Aliases
CD42d, GPV
UniGene
Hs.73734
Gene Chromosome Location
Chr.3: 194394821-194399266 on Build GRCh38
Transcripts
NM_004488.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6261179 c.153C>A p.N51K Chr.3: 194398130-194398130 on Build GRCh38
729890 c.140G>T p.G47V Chr.3: 194398143-194398143 on Build GRCh38
3591570 c.243C>T p.I81I Chr.3: 194398040-194398040 on Build GRCh38
4625048 c.43C>T p.R15C Chr.3: 194398240-194398240 on Build GRCh38
5560540 c.180G>T p.M60I Chr.3: 194398103-194398103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777081348 Chr.3: 194397871-194397871 on Build GRCh38
rs763248310 Chr.3: 194398065-194398065 on Build GRCh38
rs1043497790 Chr.3: 194398160-194398160 on Build GRCh38
rs140994443 Chr.3: 194397970-194397970 on Build GRCh38
rs781760228 Chr.3: 194398040-194398040 on Build GRCh38
rs144911830 Chr.3: 194398299-194398299 on Build GRCh38
rs1032873836 Chr.3: 194398037-194398037 on Build GRCh38
rs779182714 Chr.3: 194398218-194398218 on Build GRCh38
rs747049345 Chr.3: 194398038-194398038 on Build GRCh38
rs549784939 Chr.3: 194398253-194398253 on Build GRCh38
rs918146384 Chr.3: 194398108-194398108 on Build GRCh38
rs772732114 Chr.3: 194397999-194397999 on Build GRCh38
rs897065551 Chr.3: 194397885-194397885 on Build GRCh38
rs762597397 Chr.3: 194398002-194398002 on Build GRCh38
rs556746198 Chr.3: 194398159-194398159 on Build GRCh38
rs751033170 Chr.3: 194397908-194397908 on Build GRCh38
rs148635904 Chr.3: 194398175-194398175 on Build GRCh38
rs531464548 Chr.3: 194397840-194397840 on Build GRCh38
rs941308233 Chr.3: 194397854-194397854 on Build GRCh38
rs750629635 Chr.3: 194398170-194398170 on Build GRCh38
rs757961631 Chr.3: 194397806-194397806 on Build GRCh38
rs759843994 Chr.3: 194398190-194398190 on Build GRCh38
rs753151808 Chr.3: 194398224-194398224 on Build GRCh38
rs752814508 Chr.3: 194398305-194398305 on Build GRCh38
rs771091355 Chr.3: 194397835-194397835 on Build GRCh38
rs540642755 Chr.3: 194397989-194397989 on Build GRCh38
rs767073211 Chr.3: 194398096-194398096 on Build GRCh38
rs147926792 Chr.3: 194397841-194397841 on Build GRCh38
rs868414360 Chr.3: 194398068-194398068 on Build GRCh38
rs201748538 Chr.3: 194398009-194398009 on Build GRCh38
rs200212020 Chr.3: 194398087-194398087 on Build GRCh38
rs759111164 Chr.3: 194397840-194397841 on Build GRCh38
rs968979971 Chr.3: 194398031-194398031 on Build GRCh38
rs760127909 Chr.3: 194398170-194398170 on Build GRCh38
rs147093306 Chr.3: 194397895-194397895 on Build GRCh38
rs760595151 Chr.3: 194398101-194398101 on Build GRCh38
rs71316299 Chr.3: 194397836-194397836 on Build GRCh38
rs999144567 Chr.3: 194397963-194397963 on Build GRCh38
rs768103652 Chr.3: 194398258-194398258 on Build GRCh38
rs773982652 Chr.3: 194397803-194397803 on Build GRCh38
rs752780195 Chr.3: 194398194-194398194 on Build GRCh38
rs770577099 Chr.3: 194398066-194398066 on Build GRCh38
rs779830095 Chr.3: 194397809-194397809 on Build GRCh38
rs770204579 Chr.3: 194398045-194398045 on Build GRCh38
rs148391579 Chr.3: 194398318-194398318 on Build GRCh38
rs765569501 Chr.3: 194398252-194398253 on Build GRCh38
rs749857036 Chr.3: 194397800-194397800 on Build GRCh38
rs145597337 Chr.3: 194398112-194398112 on Build GRCh38
rs1045271957 Chr.3: 194397826-194397826 on Build GRCh38
rs757582911 Chr.3: 194398249-194398249 on Build GRCh38
rs755849586 Chr.3: 194398014-194398014 on Build GRCh38
rs773566428 Chr.3: 194398051-194398051 on Build GRCh38
rs140404736 Chr.3: 194397966-194397966 on Build GRCh38
rs755100270 Chr.3: 194398116-194398116 on Build GRCh38
rs777692043 Chr.3: 194398028-194398028 on Build GRCh38
rs746160624 Chr.3: 194397959-194397959 on Build GRCh38
rs150190099 Chr.3: 194398010-194398010 on Build GRCh38
rs541888716 Chr.3: 194398069-194398069 on Build GRCh38
rs138140981 Chr.3: 194398114-194398114 on Build GRCh38
rs768901298 Chr.3: 194398039-194398039 on Build GRCh38
rs141284024 Chr.3: 194398199-194398199 on Build GRCh38
rs773979922 Chr.3: 194398146-194398146 on Build GRCh38
rs371868475 Chr.3: 194397796-194397796 on Build GRCh38
rs754760971 Chr.3: 194397819-194397819 on Build GRCh38
rs774925410 Chr.3: 194398162-194398162 on Build GRCh38
rs529238019 Chr.3: 194398279-194398279 on Build GRCh38
rs773579447 Chr.3: 194398276-194398277 on Build GRCh38
rs760822849 Chr.3: 194398325-194398325 on Build GRCh38
rs764055488 Chr.3: 194398182-194398182 on Build GRCh38
rs771529109 Chr.3: 194397979-194397979 on Build GRCh38
rs778320454 Chr.3: 194398208-194398208 on Build GRCh38
rs147680603 Chr.3: 194397938-194397938 on Build GRCh38
rs576801443 Chr.3: 194397890-194397890 on Build GRCh38
rs775175208 Chr.3: 194398169-194398170 on Build GRCh38
rs748727646 Chr.3: 194398041-194398041 on Build GRCh38
rs761651431 Chr.3: 194398186-194398186 on Build GRCh38
rs560286404 Chr.3: 194398127-194398127 on Build GRCh38
rs199581779 Chr.3: 194397968-194397968 on Build GRCh38
rs776477645 Chr.3: 194398184-194398184 on Build GRCh38
rs1037886809 Chr.3: 194397868-194397868 on Build GRCh38
rs149317860 Chr.3: 194398034-194398034 on Build GRCh38
rs369205730 Chr.3: 194398070-194398070 on Build GRCh38
rs747021716 Chr.3: 194397816-194397816 on Build GRCh38
rs768062632 Chr.3: 194397787-194397787 on Build GRCh38
rs749818356 Chr.3: 194398214-194398214 on Build GRCh38
rs745587507 Chr.3: 194398155-194398155 on Build GRCh38
rs747949677 Chr.3: 194397832-194397832 on Build GRCh38
rs773089110 Chr.3: 194398271-194398271 on Build GRCh38
rs34799505 Chr.3: 194398012-194398012 on Build GRCh38
rs752627528 Chr.3: 194398098-194398098 on Build GRCh38
rs188238078 Chr.3: 194398151-194398151 on Build GRCh38
rs1021969382 Chr.3: 194397950-194397950 on Build GRCh38
rs765186322 Chr.3: 194398187-194398187 on Build GRCh38
rs746246856 Chr.3: 194398237-194398237 on Build GRCh38
rs373377330 Chr.3: 194398193-194398193 on Build GRCh38
rs749631647 Chr.3: 194398144-194398144 on Build GRCh38
rs780895842 Chr.3: 194397825-194397825 on Build GRCh38
rs750136044 Chr.3: 194397888-194397888 on Build GRCh38
rs761333814 Chr.3: 194397851-194397851 on Build GRCh38
rs755943185 Chr.3: 194397942-194397942 on Build GRCh38
rs752760235 Chr.3: 194397793-194397793 on Build GRCh38
rs894949889 Chr.3: 194397928-194397928 on Build GRCh38
rs752449361 Chr.3: 194397939-194397939 on Build GRCh38
rs770248104 Chr.3: 194397993-194397993 on Build GRCh38
rs558020104 Chr.3: 194397974-194397974 on Build GRCh38
rs769240414 Chr.3: 194397972-194397972 on Build GRCh38
rs941378698 Chr.3: 194398094-194398094 on Build GRCh38
rs202032002 Chr.3: 194398217-194398217 on Build GRCh38
rs564091697 Chr.3: 194398196-194398196 on Build GRCh38
rs142512027 Chr.3: 194398139-194398139 on Build GRCh38
rs771665773 Chr.3: 194398243-194398243 on Build GRCh38
rs765977841 Chr.3: 194397906-194397906 on Build GRCh38
rs775848940 Chr.3: 194397842-194397842 on Build GRCh38
rs776621159 Chr.3: 194398266-194398266 on Build GRCh38
rs189097872 Chr.3: 194398005-194398005 on Build GRCh38
rs760383737 Chr.3: 194398176-194398176 on Build GRCh38
rs1056758273 Chr.3: 194398125-194398125 on Build GRCh38
rs758590525 Chr.3: 194398035-194398035 on Build GRCh38
rs780165992 Chr.3: 194398036-194398036 on Build GRCh38
rs766939396 Chr.3: 194398317-194398318 on Build GRCh38
rs781350355 Chr.3: 194398120-194398120 on Build GRCh38
rs772042244 Chr.3: 194397965-194397965 on Build GRCh38
rs754850636 Chr.3: 194397933-194397933 on Build GRCh38
rs762241412 Chr.3: 194397986-194397986 on Build GRCh38
rs932705672 Chr.3: 194397783-194397783 on Build GRCh38
rs764705809 Chr.3: 194397866-194397866 on Build GRCh38
rs568045079 Chr.3: 194398268-194398268 on Build GRCh38
rs949499114 Chr.3: 194397821-194397821 on Build GRCh38
rs186532879 Chr.3: 194398149-194398149 on Build GRCh38
rs370921872 Chr.3: 194398102-194398102 on Build GRCh38
rs771718752 Chr.3: 194398161-194398161 on Build GRCh38
rs370787336 Chr.3: 194398291-194398291 on Build GRCh38
rs764761485 Chr.3: 194397887-194397887 on Build GRCh38
rs778980398 Chr.3: 194397953-194397953 on Build GRCh38
rs773697033 Chr.3: 194397808-194397808 on Build GRCh38
rs1223989 Chr.3: 194398145-194398145 on Build GRCh38
rs890346723 Chr.3: 194398229-194398229 on Build GRCh38
rs753623237 Chr.3: 194398103-194398103 on Build GRCh38
rs73889857 Chr.3: 194397884-194397884 on Build GRCh38

View Full Product Details