Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184388155-184388753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF2
Entrez Gene ID
3660
Gene Name
interferon regulatory factor 2
Gene Aliases
IRF-2
UniGene
Hs.654566
Gene Chromosome Location
Chr.4: 184387722-184474572 on Build GRCh38
Transcripts
NM_002199.3

SNP Detail

SNP ID Chromosome Location
rs548401942 Chr.4: 184388437-184388437 on Build GRCh38
rs201154426 Chr.4: 184388210-184388210 on Build GRCh38
rs751259533 Chr.4: 184388217-184388221 on Build GRCh38
rs921933010 Chr.4: 184388436-184388436 on Build GRCh38
rs76927106 Chr.4: 184388293-184388293 on Build GRCh38
rs1011598866 Chr.4: 184388261-184388261 on Build GRCh38
rs898341542 Chr.4: 184388651-184388651 on Build GRCh38
rs541614799 Chr.4: 184388521-184388521 on Build GRCh38
rs116764376 Chr.4: 184388451-184388451 on Build GRCh38
rs1052585472 Chr.4: 184388178-184388178 on Build GRCh38
rs765953997 Chr.4: 184388711-184388711 on Build GRCh38
rs980325673 Chr.4: 184388285-184388285 on Build GRCh38
rs751322015 Chr.4: 184388714-184388714 on Build GRCh38
rs752436102 Chr.4: 184388728-184388728 on Build GRCh38
rs762602819 Chr.4: 184388710-184388710 on Build GRCh38
rs148973928 Chr.4: 184388207-184388211 on Build GRCh38
rs915421100 Chr.4: 184388492-184388492 on Build GRCh38
rs141294569 Chr.4: 184388466-184388466 on Build GRCh38
rs946958580 Chr.4: 184388529-184388529 on Build GRCh38
rs752622719 Chr.4: 184388594-184388594 on Build GRCh38
rs1049343732 Chr.4: 184388303-184388303 on Build GRCh38
rs996737566 Chr.4: 184388672-184388672 on Build GRCh38
rs777994873 Chr.4: 184388641-184388641 on Build GRCh38
rs772717277 Chr.4: 184388180-184388180 on Build GRCh38
rs901952140 Chr.4: 184388404-184388404 on Build GRCh38
rs904816439 Chr.4: 184388544-184388544 on Build GRCh38
rs992205773 Chr.4: 184388251-184388251 on Build GRCh38
rs956810568 Chr.4: 184388473-184388473 on Build GRCh38
rs753595210 Chr.4: 184388371-184388371 on Build GRCh38
rs567574848 Chr.4: 184388365-184388365 on Build GRCh38
rs201813587 Chr.4: 184388211-184388211 on Build GRCh38
rs372445068 Chr.4: 184388227-184388246 on Build GRCh38
rs755895188 Chr.4: 184388734-184388734 on Build GRCh38
rs1004047786 Chr.4: 184388337-184388337 on Build GRCh38
rs548079732 Chr.4: 184388279-184388279 on Build GRCh38
rs375222027 Chr.4: 184388454-184388454 on Build GRCh38
rs1018922933 Chr.4: 184388727-184388727 on Build GRCh38
rs749252921 Chr.4: 184388433-184388433 on Build GRCh38
rs969201074 Chr.4: 184388592-184388593 on Build GRCh38
rs148659404 Chr.4: 184388428-184388428 on Build GRCh38
rs562593345 Chr.4: 184388481-184388481 on Build GRCh38
rs760304289 Chr.4: 184388301-184388301 on Build GRCh38
rs938992855 Chr.4: 184388561-184388561 on Build GRCh38
rs547540554 Chr.4: 184388616-184388616 on Build GRCh38
rs951145871 Chr.4: 184388324-184388324 on Build GRCh38
rs545272743 Chr.4: 184388569-184388569 on Build GRCh38
rs911967908 Chr.4: 184388459-184388459 on Build GRCh38
rs758271782 Chr.4: 184388623-184388623 on Build GRCh38
rs3044523 Chr.4: 184388247-184388247 on Build GRCh38
rs890665418 Chr.4: 184388232-184388232 on Build GRCh38
rs1044720566 Chr.4: 184388286-184388286 on Build GRCh38
rs886841610 Chr.4: 184388619-184388619 on Build GRCh38
rs952200740 Chr.4: 184388697-184388697 on Build GRCh38
rs371757121 Chr.4: 184388716-184388716 on Build GRCh38
rs568181114 Chr.4: 184388537-184388537 on Build GRCh38
rs1056184733 Chr.4: 184388608-184388608 on Build GRCh38
rs374067305 Chr.4: 184388242-184388246 on Build GRCh38
rs757026700 Chr.4: 184388207-184388221 on Build GRCh38
rs917692414 Chr.4: 184388278-184388278 on Build GRCh38
rs886658925 Chr.4: 184388309-184388309 on Build GRCh38
rs975748985 Chr.4: 184388432-184388432 on Build GRCh38
rs940717849 Chr.4: 184388325-184388325 on Build GRCh38
rs1034769569 Chr.4: 184388316-184388316 on Build GRCh38
rs375994580 Chr.4: 184388726-184388726 on Build GRCh38
rs766066714 Chr.4: 184388335-184388335 on Build GRCh38
rs181297181 Chr.4: 184388483-184388483 on Build GRCh38
rs991860788 Chr.4: 184388527-184388527 on Build GRCh38
rs527758522 Chr.4: 184388595-184388595 on Build GRCh38

View Full Product Details