Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35870961-35871386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL7R
Entrez Gene ID
3575
Gene Name
interleukin 7 receptor
Gene Aliases
CD127, CDW127, IL-7R-alpha, IL7RA, ILRA
UniGene
Hs.591742
Gene Chromosome Location
Chr.5: 35856875-35879603 on Build GRCh38
Transcripts
NM_002185.3 NR_120485.1 XM_005248299.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1437174 c.481A>C p.K161Q Chr.5: 35871157-35871157 on Build GRCh38
6171022 c.510C>A p.R170R Chr.5: 35871186-35871186 on Build GRCh38
403958 c.400G>T p.D134Y Chr.5: 35871076-35871076 on Build GRCh38
3429410 c.445A>G p.T149A Chr.5: 35871121-35871121 on Build GRCh38
3393542 c.427G>T p.A143S Chr.5: 35871103-35871103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758320927 Chr.5: 35871213-35871213 on Build GRCh38
rs1494555 Chr.5: 35871088-35871088 on Build GRCh38
rs776941426 Chr.5: 35871172-35871172 on Build GRCh38
rs145210284 Chr.5: 35871184-35871184 on Build GRCh38
rs587778403 Chr.5: 35871114-35871114 on Build GRCh38
rs200373233 Chr.5: 35871095-35871095 on Build GRCh38
rs199697302 Chr.5: 35871177-35871177 on Build GRCh38
rs104893894 Chr.5: 35871070-35871070 on Build GRCh38
rs755779539 Chr.5: 35871155-35871155 on Build GRCh38
rs770431510 Chr.5: 35870999-35870999 on Build GRCh38
rs372676633 Chr.5: 35871103-35871103 on Build GRCh38
rs769080350 Chr.5: 35871248-35871248 on Build GRCh38
rs763573007 Chr.5: 35871047-35871047 on Build GRCh38
rs777878144 Chr.5: 35871214-35871214 on Build GRCh38
rs143661008 Chr.5: 35871195-35871195 on Build GRCh38
rs772130250 Chr.5: 35871091-35871091 on Build GRCh38
rs751109965 Chr.5: 35871056-35871056 on Build GRCh38
rs556527549 Chr.5: 35871008-35871008 on Build GRCh38
rs767009659 Chr.5: 35871119-35871119 on Build GRCh38
rs201940568 Chr.5: 35871122-35871122 on Build GRCh38
rs780061831 Chr.5: 35871078-35871078 on Build GRCh38
rs112368558 Chr.5: 35871144-35871144 on Build GRCh38
rs200332345 Chr.5: 35871118-35871118 on Build GRCh38
rs914777454 Chr.5: 35871246-35871246 on Build GRCh38
rs759917852 Chr.5: 35871179-35871179 on Build GRCh38
rs9282751 Chr.5: 35871361-35871361 on Build GRCh38
rs771957502 Chr.5: 35871167-35871167 on Build GRCh38
rs778642997 Chr.5: 35871211-35871211 on Build GRCh38
rs912980937 Chr.5: 35871038-35871038 on Build GRCh38
rs199654814 Chr.5: 35871191-35871191 on Build GRCh38
rs1037925725 Chr.5: 35871341-35871341 on Build GRCh38
rs192655893 Chr.5: 35871291-35871291 on Build GRCh38
rs747236451 Chr.5: 35871170-35871170 on Build GRCh38
rs139221577 Chr.5: 35871142-35871142 on Build GRCh38
rs375000958 Chr.5: 35871254-35871254 on Build GRCh38
rs531875566 Chr.5: 35871188-35871188 on Build GRCh38
rs200751605 Chr.5: 35871212-35871212 on Build GRCh38
rs767818326 Chr.5: 35871011-35871011 on Build GRCh38
rs958201184 Chr.5: 35871266-35871266 on Build GRCh38
rs756821231 Chr.5: 35871061-35871061 on Build GRCh38
rs750711583 Chr.5: 35871012-35871012 on Build GRCh38
rs773491081 Chr.5: 35871094-35871094 on Build GRCh38
rs200233367 Chr.5: 35871157-35871157 on Build GRCh38
rs375672744 Chr.5: 35871196-35871196 on Build GRCh38
rs780917994 Chr.5: 35871062-35871062 on Build GRCh38
rs746163082 Chr.5: 35871220-35871220 on Build GRCh38
rs200034351 Chr.5: 35871039-35871039 on Build GRCh38
rs567465471 Chr.5: 35870992-35870992 on Build GRCh38
rs749131336 Chr.5: 35871138-35871138 on Build GRCh38
rs747091532 Chr.5: 35871216-35871216 on Build GRCh38
rs766807796 Chr.5: 35871028-35871028 on Build GRCh38
rs940498435 Chr.5: 35871326-35871326 on Build GRCh38
rs772191954 Chr.5: 35871166-35871166 on Build GRCh38
rs1020401719 Chr.5: 35871228-35871228 on Build GRCh38
rs148306532 Chr.5: 35871360-35871360 on Build GRCh38
rs201790329 Chr.5: 35871109-35871109 on Build GRCh38
rs766507230 Chr.5: 35871355-35871355 on Build GRCh38
rs967019465 Chr.5: 35871251-35871251 on Build GRCh38
rs199727195 Chr.5: 35871136-35871136 on Build GRCh38
rs745590108 Chr.5: 35871067-35871067 on Build GRCh38
rs536233382 Chr.5: 35871005-35871005 on Build GRCh38
rs376798079 Chr.5: 35871019-35871019 on Build GRCh38
rs376971243 Chr.5: 35871045-35871045 on Build GRCh38
rs907814647 Chr.5: 35871312-35871312 on Build GRCh38
rs149833809 Chr.5: 35871204-35871204 on Build GRCh38
rs201980969 Chr.5: 35871225-35871225 on Build GRCh38
rs760005945 Chr.5: 35871115-35871115 on Build GRCh38
rs1041285489 Chr.5: 35871321-35871321 on Build GRCh38
rs112805866 Chr.5: 35871132-35871132 on Build GRCh38
rs749945425 Chr.5: 35871193-35871193 on Build GRCh38
rs754827305 Chr.5: 35871210-35871210 on Build GRCh38
rs201088820 Chr.5: 35871260-35871260 on Build GRCh38
rs765765854 Chr.5: 35871116-35871116 on Build GRCh38
rs985994233 Chr.5: 35871272-35871272 on Build GRCh38
rs768760071 Chr.5: 35871081-35871081 on Build GRCh38
rs200772681 Chr.5: 35871100-35871100 on Build GRCh38
rs755541920 Chr.5: 35871042-35871042 on Build GRCh38
rs747698244 Chr.5: 35871115-35871115 on Build GRCh38
rs911496330 Chr.5: 35871280-35871280 on Build GRCh38
rs754341555 Chr.5: 35871029-35871029 on Build GRCh38
rs9282750 Chr.5: 35871303-35871303 on Build GRCh38
rs200625485 Chr.5: 35871217-35871217 on Build GRCh38
rs545307253 Chr.5: 35871087-35871087 on Build GRCh38
rs200135239 Chr.5: 35871232-35871232 on Build GRCh38
rs199779006 Chr.5: 35871080-35871080 on Build GRCh38
rs201012821 Chr.5: 35871083-35871083 on Build GRCh38
rs779013920 Chr.5: 35871152-35871152 on Build GRCh38
rs2228141 Chr.5: 35871171-35871171 on Build GRCh38
rs753635194 Chr.5: 35871209-35871209 on Build GRCh38
rs149161098 Chr.5: 35871185-35871185 on Build GRCh38
rs750985822 Chr.5: 35871117-35871117 on Build GRCh38
rs547622474 Chr.5: 35871311-35871311 on Build GRCh38
rs748111926 Chr.5: 35871153-35871153 on Build GRCh38

View Full Product Details