Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204198843-204199379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLT1A
Entrez Gene ID
127845
Gene Name
golgi transport 1A
Gene Aliases
CGI-141, GOT1, HMFN1187, YMR292W
UniGene
Hs.532401
Gene Chromosome Location
Chr.1: 204198158-204214092 on Build GRCh38
Transcripts
NM_198447.1 XM_017000314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7424843 c.319G>A p.G107S Chr.1: 204199236-204199236 on Build GRCh38
4027222 c.328G>T p.G110C Chr.1: 204199227-204199227 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs960899456 Chr.1: 204199226-204199226 on Build GRCh38
rs868308808 Chr.1: 204199196-204199196 on Build GRCh38
rs771376509 Chr.1: 204199235-204199235 on Build GRCh38
rs1052722047 Chr.1: 204199252-204199252 on Build GRCh38
rs992422692 Chr.1: 204198980-204198980 on Build GRCh38
rs959933800 Chr.1: 204199078-204199078 on Build GRCh38
rs747306254 Chr.1: 204199222-204199222 on Build GRCh38
rs998469650 Chr.1: 204198941-204198941 on Build GRCh38
rs762580397 Chr.1: 204199279-204199279 on Build GRCh38
rs891508715 Chr.1: 204198967-204198967 on Build GRCh38
rs972166168 Chr.1: 204199326-204199326 on Build GRCh38
rs374794242 Chr.1: 204199243-204199243 on Build GRCh38
rs868477938 Chr.1: 204199237-204199237 on Build GRCh38
rs755045787 Chr.1: 204199188-204199188 on Build GRCh38
rs924142582 Chr.1: 204198946-204198946 on Build GRCh38
rs933819470 Chr.1: 204199351-204199351 on Build GRCh38
rs182073445 Chr.1: 204199031-204199031 on Build GRCh38
rs889722727 Chr.1: 204199130-204199130 on Build GRCh38
rs763625199 Chr.1: 204199341-204199341 on Build GRCh38
rs918582712 Chr.1: 204199142-204199143 on Build GRCh38
rs1034652218 Chr.1: 204199068-204199068 on Build GRCh38
rs1049115552 Chr.1: 204199010-204199010 on Build GRCh38
rs549774182 Chr.1: 204199278-204199278 on Build GRCh38
rs989828607 Chr.1: 204198951-204198951 on Build GRCh38
rs763806051 Chr.1: 204199295-204199295 on Build GRCh38
rs896983529 Chr.1: 204199040-204199040 on Build GRCh38
rs750432619 Chr.1: 204199187-204199187 on Build GRCh38
rs139644560 Chr.1: 204199201-204199201 on Build GRCh38
rs751268397 Chr.1: 204199163-204199163 on Build GRCh38
rs761359357 Chr.1: 204198916-204198916 on Build GRCh38
rs563422306 Chr.1: 204198994-204198994 on Build GRCh38
rs922791073 Chr.1: 204199219-204199219 on Build GRCh38
rs961208236 Chr.1: 204198959-204198959 on Build GRCh38
rs984761692 Chr.1: 204199301-204199301 on Build GRCh38
rs967787938 Chr.1: 204198909-204198909 on Build GRCh38
rs953729099 Chr.1: 204198883-204198883 on Build GRCh38
rs758572801 Chr.1: 204199209-204199209 on Build GRCh38
rs764653078 Chr.1: 204199250-204199250 on Build GRCh38
rs1051007842 Chr.1: 204199042-204199042 on Build GRCh38
rs998460371 Chr.1: 204199041-204199041 on Build GRCh38
rs199661237 Chr.1: 204199214-204199214 on Build GRCh38
rs1036826091 Chr.1: 204199032-204199032 on Build GRCh38
rs756533626 Chr.1: 204199320-204199320 on Build GRCh38
rs940710935 Chr.1: 204199321-204199321 on Build GRCh38
rs80046452 Chr.1: 204199141-204199141 on Build GRCh38
rs775360660 Chr.1: 204198898-204198898 on Build GRCh38
rs190061218 Chr.1: 204199277-204199277 on Build GRCh38
rs909273370 Chr.1: 204199018-204199018 on Build GRCh38
rs909969026 Chr.1: 204199181-204199181 on Build GRCh38
rs761496640 Chr.1: 204199310-204199310 on Build GRCh38
rs569756844 Chr.1: 204199329-204199329 on Build GRCh38
rs762709101 Chr.1: 204198901-204198901 on Build GRCh38
rs757111163 Chr.1: 204199167-204199167 on Build GRCh38
rs186896172 Chr.1: 204199168-204199168 on Build GRCh38
rs774099021 Chr.1: 204199308-204199308 on Build GRCh38
rs746457288 Chr.1: 204199242-204199242 on Build GRCh38
rs764902073 Chr.1: 204199135-204199135 on Build GRCh38
rs142169045 Chr.1: 204199037-204199039 on Build GRCh38
rs775280860 Chr.1: 204199136-204199136 on Build GRCh38
rs776995034 Chr.1: 204199236-204199236 on Build GRCh38
rs576792415 Chr.1: 204199001-204199001 on Build GRCh38
rs536677981 Chr.1: 204198908-204198908 on Build GRCh38
rs761192690 Chr.1: 204199234-204199234 on Build GRCh38
rs999235345 Chr.1: 204199199-204199199 on Build GRCh38
rs774074123 Chr.1: 204198914-204198914 on Build GRCh38
rs559277204 Chr.1: 204199138-204199138 on Build GRCh38
rs543463371 Chr.1: 204198903-204198903 on Build GRCh38
rs916820617 Chr.1: 204199269-204199269 on Build GRCh38
rs144909438 Chr.1: 204199195-204199195 on Build GRCh38
rs557046850 Chr.1: 204198985-204198985 on Build GRCh38

View Full Product Details