Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33426580-33427133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNGR2
Entrez Gene ID
3460
Gene Name
interferon gamma receptor 2 (interferon gamma transducer 1)
Gene Aliases
AF-1, IFGR2, IFNGT1, IMD28
UniGene
Hs.634632
Gene Chromosome Location
Chr.21: 33402896-33437521 on Build GRCh38
Transcripts
NM_005534.3 XM_005260969.2 XM_011529553.1 XM_011529554.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3735611 c.463C>T p.L155F Chr.21: 33426934-33426934 on Build GRCh38
7329299 c.537C>A p.Y179* Chr.21: 33427008-33427008 on Build GRCh38
161667 c.461C>T p.S154F Chr.21: 33426932-33426932 on Build GRCh38
220996 c.466A>C p.I156L Chr.21: 33426937-33426937 on Build GRCh38
3991906 c.429A>T p.P143P Chr.21: 33426900-33426900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974364301 Chr.21: 33426821-33426821 on Build GRCh38
rs121913210 Chr.21: 33426999-33426999 on Build GRCh38
rs931746838 Chr.21: 33426722-33426722 on Build GRCh38
rs1010193498 Chr.21: 33426840-33426840 on Build GRCh38
rs900415224 Chr.21: 33426771-33426771 on Build GRCh38
rs192350781 Chr.21: 33426674-33426674 on Build GRCh38
rs375419913 Chr.21: 33426985-33426985 on Build GRCh38
rs892472880 Chr.21: 33426837-33426837 on Build GRCh38
rs902362680 Chr.21: 33426922-33426922 on Build GRCh38
rs121913208 Chr.21: 33426937-33426937 on Build GRCh38
rs367663086 Chr.21: 33426855-33426855 on Build GRCh38
rs190049250 Chr.21: 33427062-33427062 on Build GRCh38
rs915360407 Chr.21: 33426681-33426681 on Build GRCh38
rs751606664 Chr.21: 33426916-33426916 on Build GRCh38
rs552291982 Chr.21: 33426967-33426967 on Build GRCh38
rs978481972 Chr.21: 33426699-33426699 on Build GRCh38
rs773394548 Chr.21: 33426843-33426871 on Build GRCh38
rs949113484 Chr.21: 33426709-33426709 on Build GRCh38
rs1054781077 Chr.21: 33426683-33426683 on Build GRCh38
rs969332445 Chr.21: 33426609-33426609 on Build GRCh38
rs930508115 Chr.21: 33426794-33426794 on Build GRCh38
rs112107702 Chr.21: 33426716-33426716 on Build GRCh38
rs74315444 Chr.21: 33426974-33426974 on Build GRCh38
rs773547071 Chr.21: 33426909-33426909 on Build GRCh38
rs968591114 Chr.21: 33426866-33426866 on Build GRCh38
rs563502088 Chr.21: 33426773-33426773 on Build GRCh38
rs936531276 Chr.21: 33426925-33426925 on Build GRCh38
rs753283715 Chr.21: 33427069-33427069 on Build GRCh38
rs780601430 Chr.21: 33426881-33426881 on Build GRCh38
rs184664014 Chr.21: 33427038-33427038 on Build GRCh38
rs115346998 Chr.21: 33427004-33427004 on Build GRCh38
rs961821936 Chr.21: 33426795-33426796 on Build GRCh38
rs1015768527 Chr.21: 33426794-33426795 on Build GRCh38
rs917581605 Chr.21: 33426847-33426847 on Build GRCh38
rs1051428325 Chr.21: 33426724-33426724 on Build GRCh38
rs759324312 Chr.21: 33427026-33427026 on Build GRCh38
rs747498076 Chr.21: 33426888-33426888 on Build GRCh38
rs754691445 Chr.21: 33426844-33426844 on Build GRCh38
rs755628412 Chr.21: 33426955-33426955 on Build GRCh38
rs756905466 Chr.21: 33427084-33427084 on Build GRCh38
rs11910627 Chr.21: 33427043-33427043 on Build GRCh38
rs911597815 Chr.21: 33426783-33426783 on Build GRCh38
rs896258091 Chr.21: 33426685-33426685 on Build GRCh38
rs121913209 Chr.21: 33426981-33426981 on Build GRCh38
rs771387730 Chr.21: 33427061-33427065 on Build GRCh38
rs769963583 Chr.21: 33427003-33427003 on Build GRCh38
rs121913202 Chr.21: 33426687-33426687 on Build GRCh38
rs769220371 Chr.21: 33426778-33426779 on Build GRCh38
rs368333636 Chr.21: 33426995-33426995 on Build GRCh38
rs17878711 Chr.21: 33427015-33427015 on Build GRCh38
rs1034975460 Chr.21: 33426828-33426828 on Build GRCh38
rs563280647 Chr.21: 33427092-33427092 on Build GRCh38
rs905268347 Chr.21: 33426798-33426798 on Build GRCh38
rs137854906 Chr.21: 33426831-33426836 on Build GRCh38
rs1032645492 Chr.21: 33426738-33426741 on Build GRCh38
rs121913205 Chr.21: 33426825-33426825 on Build GRCh38
rs375520816 Chr.21: 33426919-33426919 on Build GRCh38
rs746279788 Chr.21: 33426860-33426860 on Build GRCh38
rs987257772 Chr.21: 33426792-33426793 on Build GRCh38
rs766274599 Chr.21: 33426913-33426913 on Build GRCh38
rs902309393 Chr.21: 33426717-33426717 on Build GRCh38
rs767448310 Chr.21: 33426945-33426945 on Build GRCh38
rs121913207 Chr.21: 33426859-33426859 on Build GRCh38
rs554716509 Chr.21: 33426992-33426992 on Build GRCh38
rs758160187 Chr.21: 33427010-33427010 on Build GRCh38
rs755652308 Chr.21: 33426679-33426679 on Build GRCh38
rs752170562 Chr.21: 33426952-33426952 on Build GRCh38
rs146481328 Chr.21: 33426777-33426778 on Build GRCh38
rs779518163 Chr.21: 33426779-33426780 on Build GRCh38
rs542657768 Chr.21: 33427024-33427024 on Build GRCh38
rs188848981 Chr.21: 33426908-33426908 on Build GRCh38
rs121913203 Chr.21: 33426792-33426792 on Build GRCh38
rs996827085 Chr.21: 33426778-33426779 on Build GRCh38
rs757685822 Chr.21: 33426796-33426796 on Build GRCh38
rs749509332 Chr.21: 33427014-33427014 on Build GRCh38
rs776581325 Chr.21: 33426907-33426907 on Build GRCh38
rs201440484 Chr.21: 33427000-33427000 on Build GRCh38
rs530773732 Chr.21: 33426793-33426793 on Build GRCh38
rs892436780 Chr.21: 33427112-33427112 on Build GRCh38
rs775466154 Chr.21: 33427039-33427039 on Build GRCh38
rs17878639 Chr.21: 33426910-33426910 on Build GRCh38
rs78072678 Chr.21: 33426891-33426891 on Build GRCh38
rs1000791560 Chr.21: 33426717-33426717 on Build GRCh38
rs570794289 Chr.21: 33426875-33426875 on Build GRCh38
rs142997921 Chr.21: 33426980-33426980 on Build GRCh38
rs1003486830 Chr.21: 33426611-33426611 on Build GRCh38
rs761197245 Chr.21: 33427054-33427054 on Build GRCh38
rs547939989 Chr.21: 33426770-33426770 on Build GRCh38
rs1028277774 Chr.21: 33426791-33426791 on Build GRCh38
rs940571580 Chr.21: 33426790-33426790 on Build GRCh38
rs121913204 Chr.21: 33426805-33426806 on Build GRCh38
rs770966080 Chr.21: 33427103-33427103 on Build GRCh38
rs78407108 Chr.21: 33426675-33426675 on Build GRCh38

View Full Product Details