Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 11877218-11877634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSPT1
Entrez Gene ID
2935
Gene Name
G1 to S phase transition 1
Gene Aliases
551G9.2, ETF3A, GST1, eRF3a
UniGene
Hs.528780
Gene Chromosome Location
Chr.16: 11868128-11916662 on Build GRCh38
Transcripts
NM_001130006.1 NM_001130007.1 NM_002094.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5850165 c.1156C>G p.L386V Chr.16: 11877439-11877439 on Build GRCh38
1189182 c.1066G>T p.V356L Chr.16: 11877529-11877529 on Build GRCh38
2142497 c.1492G>A p.E498K Chr.16: 11877517-11877517 on Build GRCh38
4186972 c.1024G>A p.E342K Chr.16: 11877571-11877571 on Build GRCh38
4849346 c.1522G>A p.E508K Chr.16: 11877487-11877487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774096473 Chr.16: 11877414-11877414 on Build GRCh38
rs901008787 Chr.16: 11877242-11877242 on Build GRCh38
rs184130992 Chr.16: 11877594-11877594 on Build GRCh38
rs776207350 Chr.16: 11877488-11877488 on Build GRCh38
rs767283791 Chr.16: 11877585-11877585 on Build GRCh38
rs1051553897 Chr.16: 11877441-11877441 on Build GRCh38
rs376143242 Chr.16: 11877464-11877464 on Build GRCh38
rs768671780 Chr.16: 11877503-11877503 on Build GRCh38
rs559521042 Chr.16: 11877424-11877424 on Build GRCh38
rs529532806 Chr.16: 11877330-11877330 on Build GRCh38
rs1025032056 Chr.16: 11877368-11877368 on Build GRCh38
rs1012296654 Chr.16: 11877361-11877361 on Build GRCh38
rs750574311 Chr.16: 11877537-11877537 on Build GRCh38
rs982965596 Chr.16: 11877261-11877261 on Build GRCh38
rs369309563 Chr.16: 11877529-11877529 on Build GRCh38
rs751454198 Chr.16: 11877345-11877345 on Build GRCh38
rs377596394 Chr.16: 11877575-11877575 on Build GRCh38
rs778901109 Chr.16: 11877393-11877395 on Build GRCh38
rs187336511 Chr.16: 11877382-11877382 on Build GRCh38
rs979297213 Chr.16: 11877245-11877245 on Build GRCh38
rs868638142 Chr.16: 11877564-11877564 on Build GRCh38
rs756047943 Chr.16: 11877589-11877589 on Build GRCh38
rs563858912 Chr.16: 11877328-11877328 on Build GRCh38
rs1032344359 Chr.16: 11877341-11877341 on Build GRCh38
rs373950871 Chr.16: 11877401-11877401 on Build GRCh38
rs1038688994 Chr.16: 11877569-11877569 on Build GRCh38
rs984368049 Chr.16: 11877552-11877552 on Build GRCh38
rs761230839 Chr.16: 11877489-11877489 on Build GRCh38
rs781110337 Chr.16: 11877373-11877373 on Build GRCh38
rs143412070 Chr.16: 11877260-11877260 on Build GRCh38
rs1025032743 Chr.16: 11877288-11877288 on Build GRCh38
rs557275356 Chr.16: 11877247-11877247 on Build GRCh38
rs1027140808 Chr.16: 11877254-11877254 on Build GRCh38
rs770646823 Chr.16: 11877402-11877402 on Build GRCh38
rs200314229 Chr.16: 11877393-11877393 on Build GRCh38
rs776744295 Chr.16: 11877525-11877525 on Build GRCh38
rs191855187 Chr.16: 11877530-11877530 on Build GRCh38
rs376552811 Chr.16: 11877381-11877381 on Build GRCh38
rs199609208 Chr.16: 11877586-11877586 on Build GRCh38

View Full Product Details