Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39360122-39360640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
UniGene
Hs.588202
Gene Chromosome Location
Chr.6: 39335100-39725405 on Build GRCh38
Transcripts
NM_001289020.1 NM_001289021.1 NM_001289024.1 NM_145027.4 XM_005248904.4 XM_011514357.2 XM_011514358.2 XM_011514359.2 XM_017010427.1 XM_017010428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5405254 c.322C>T p.L108L Chr.6: 39360508-39360508 on Build GRCh38
6189200 c.2021C>T p.A674V Chr.6: 39360456-39360456 on Build GRCh38
3626991 c.385C>T p.L129L Chr.6: 39360445-39360445 on Build GRCh38
3626989 c.2032C>T p.L678L Chr.6: 39360445-39360445 on Build GRCh38
3394174 c.1824C>T p.I608I Chr.6: 39360485-39360485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942178370 Chr.6: 39360227-39360227 on Build GRCh38
rs772034031 Chr.6: 39360563-39360563 on Build GRCh38
rs879558916 Chr.6: 39360172-39360172 on Build GRCh38
rs753814552 Chr.6: 39360349-39360349 on Build GRCh38
rs372318916 Chr.6: 39360550-39360550 on Build GRCh38
rs1057451600 Chr.6: 39360285-39360285 on Build GRCh38
rs552165968 Chr.6: 39360294-39360294 on Build GRCh38
rs949831826 Chr.6: 39360428-39360428 on Build GRCh38
rs530978858 Chr.6: 39360562-39360562 on Build GRCh38
rs757947423 Chr.6: 39360385-39360385 on Build GRCh38
rs1000842619 Chr.6: 39360608-39360608 on Build GRCh38
rs752729007 Chr.6: 39360538-39360538 on Build GRCh38
rs891786590 Chr.6: 39360292-39360292 on Build GRCh38
rs904376550 Chr.6: 39360317-39360317 on Build GRCh38
rs777015734 Chr.6: 39360579-39360579 on Build GRCh38
rs938749738 Chr.6: 39360261-39360261 on Build GRCh38
rs200514768 Chr.6: 39360535-39360535 on Build GRCh38
rs751197410 Chr.6: 39360544-39360544 on Build GRCh38
rs746712680 Chr.6: 39360406-39360406 on Build GRCh38
rs899222023 Chr.6: 39360233-39360233 on Build GRCh38
rs771238160 Chr.6: 39360578-39360578 on Build GRCh38
rs565511465 Chr.6: 39360298-39360298 on Build GRCh38
rs574729542 Chr.6: 39360481-39360481 on Build GRCh38
rs150690403 Chr.6: 39360561-39360561 on Build GRCh38
rs770136928 Chr.6: 39360420-39360420 on Build GRCh38
rs200668834 Chr.6: 39360381-39360381 on Build GRCh38
rs769081370 Chr.6: 39360482-39360482 on Build GRCh38
rs773286926 Chr.6: 39360566-39360566 on Build GRCh38
rs59151863 Chr.6: 39360258-39360258 on Build GRCh38
rs905787775 Chr.6: 39360143-39360143 on Build GRCh38
rs1017776270 Chr.6: 39360328-39360328 on Build GRCh38
rs879838278 Chr.6: 39360321-39360321 on Build GRCh38
rs1025737326 Chr.6: 39360418-39360418 on Build GRCh38
rs759722997 Chr.6: 39360437-39360437 on Build GRCh38
rs769061232 Chr.6: 39360560-39360560 on Build GRCh38
rs1044471866 Chr.6: 39360234-39360234 on Build GRCh38
rs557087227 Chr.6: 39360484-39360484 on Build GRCh38
rs569968168 Chr.6: 39360257-39360257 on Build GRCh38
rs1038125603 Chr.6: 39360189-39360189 on Build GRCh38
rs150466485 Chr.6: 39360511-39360511 on Build GRCh38
rs142324072 Chr.6: 39360401-39360401 on Build GRCh38
rs929579060 Chr.6: 39360167-39360167 on Build GRCh38
rs766723348 Chr.6: 39360522-39360522 on Build GRCh38
rs367760215 Chr.6: 39360350-39360350 on Build GRCh38
rs747570249 Chr.6: 39360384-39360384 on Build GRCh38
rs879715835 Chr.6: 39360314-39360314 on Build GRCh38
rs148383716 Chr.6: 39360486-39360486 on Build GRCh38
rs141566437 Chr.6: 39360510-39360510 on Build GRCh38
rs533905665 Chr.6: 39360556-39360556 on Build GRCh38
rs142999075 Chr.6: 39360483-39360483 on Build GRCh38
rs879098318 Chr.6: 39360358-39360358 on Build GRCh38
rs778915098 Chr.6: 39360366-39360366 on Build GRCh38
rs372897992 Chr.6: 39360575-39360575 on Build GRCh38
rs191950377 Chr.6: 39360146-39360146 on Build GRCh38
rs201403179 Chr.6: 39360534-39360534 on Build GRCh38
rs755944748 Chr.6: 39360345-39360345 on Build GRCh38
rs751282225 Chr.6: 39360378-39360378 on Build GRCh38
rs983685684 Chr.6: 39360169-39360169 on Build GRCh38
rs753621027 Chr.6: 39360530-39360530 on Build GRCh38
rs767641701 Chr.6: 39360217-39360217 on Build GRCh38
rs777407181 Chr.6: 39360543-39360543 on Build GRCh38
rs368687338 Chr.6: 39360466-39360466 on Build GRCh38
rs1021503513 Chr.6: 39360300-39360300 on Build GRCh38

View Full Product Details