Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15787401-15787954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA5B
Entrez Gene ID
11238
Gene Name
carbonic anhydrase 5B
Gene Aliases
CA-VB
UniGene
Hs.653287
Gene Chromosome Location
Chr.X: 15738281-15787625 on Build GRCh38
Transcripts
NM_007220.3 NR_002725.2

Gene Symbol
INE2
Entrez Gene ID
8551
Gene Name
inactivation escape 2 (non-protein coding)
Gene Aliases
NCRNA00011
UniGene
Hs.653287
Gene Chromosome Location
Chr.X: 15785716-15787589 on Build GRCh38
Transcripts
NM_007220.3 NR_002725.2

SNP Detail

SNP ID Chromosome Location
rs945789446 Chr.X: 15787522-15787522 on Build GRCh38
rs183607236 Chr.X: 15787786-15787786 on Build GRCh38
rs931999588 Chr.X: 15787687-15787687 on Build GRCh38
rs1054085669 Chr.X: 15787465-15787465 on Build GRCh38
rs1050820923 Chr.X: 15787546-15787546 on Build GRCh38
rs913179136 Chr.X: 15787885-15787885 on Build GRCh38
rs957285498 Chr.X: 15787854-15787854 on Build GRCh38
rs760528280 Chr.X: 15787680-15787680 on Build GRCh38
rs890720862 Chr.X: 15787749-15787749 on Build GRCh38
rs5936053 Chr.X: 15787448-15787448 on Build GRCh38
rs1025012070 Chr.X: 15787781-15787781 on Build GRCh38
rs190631964 Chr.X: 15787473-15787473 on Build GRCh38
rs150447419 Chr.X: 15787903-15787903 on Build GRCh38
rs763494770 Chr.X: 15787886-15787886 on Build GRCh38
rs927130864 Chr.X: 15787847-15787847 on Build GRCh38
rs1052622632 Chr.X: 15787765-15787765 on Build GRCh38
rs973502246 Chr.X: 15787510-15787510 on Build GRCh38
rs576289981 Chr.X: 15787675-15787675 on Build GRCh38
rs766381494 Chr.X: 15787717-15787717 on Build GRCh38
rs6527639 Chr.X: 15787735-15787735 on Build GRCh38
rs927203089 Chr.X: 15787425-15787425 on Build GRCh38
rs5021752 Chr.X: 15787668-15787668 on Build GRCh38
rs758768071 Chr.X: 15787869-15787869 on Build GRCh38
rs905563459 Chr.X: 15787825-15787825 on Build GRCh38
rs757919885 Chr.X: 15787884-15787884 on Build GRCh38
rs776270596 Chr.X: 15787723-15787723 on Build GRCh38
rs996165324 Chr.X: 15787729-15787729 on Build GRCh38
rs951445604 Chr.X: 15787750-15787750 on Build GRCh38
rs979869021 Chr.X: 15787804-15787804 on Build GRCh38
rs756996289 Chr.X: 15787673-15787673 on Build GRCh38
rs1021356127 Chr.X: 15787789-15787789 on Build GRCh38
rs1011770551 Chr.X: 15787783-15787783 on Build GRCh38
rs764947235 Chr.X: 15787766-15787766 on Build GRCh38

View Full Product Details