Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 90618588-90619137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRV1
Entrez Gene ID
84059
Gene Name
adhesion G protein-coupled receptor V1
Gene Aliases
FEB4, GPR98, MASS1, USH2B, USH2C, VLGR1, VLGR1b
UniGene
Hs.591777
Gene Chromosome Location
Chr.5: 90558681-91164226 on Build GRCh38
Transcripts
NM_032119.3 NR_003149.1 XM_017009963.1 XM_017009964.1 XM_017009965.1 XM_017009966.1 XM_017009967.1 XM_017009968.1 XM_017009969.1 XM_017009970.1 XM_017009971.1 XM_017009974.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
261167 c.381T>C p.L127L Chr.5: 90619109-90619109 on Build GRCh38
6251251 c.403G>T p.V135L Chr.5: 90619131-90619131 on Build GRCh38
6047372 c.382G>T p.G128* Chr.5: 90619110-90619110 on Build GRCh38
1439051 c.392G>T p.R131M Chr.5: 90619120-90619120 on Build GRCh38
5868928 c.401C>T p.T134I Chr.5: 90619129-90619129 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs965197407 Chr.5: 90618838-90618838 on Build GRCh38
rs1055655808 Chr.5: 90618704-90618704 on Build GRCh38
rs773481083 Chr.5: 90619047-90619047 on Build GRCh38
rs1023497184 Chr.5: 90618968-90618968 on Build GRCh38
rs73181609 Chr.5: 90618887-90618887 on Build GRCh38
rs200730114 Chr.5: 90619090-90619090 on Build GRCh38
rs778709054 Chr.5: 90619111-90619111 on Build GRCh38
rs1018477739 Chr.5: 90618831-90618831 on Build GRCh38
rs998043294 Chr.5: 90618863-90618863 on Build GRCh38
rs981677383 Chr.5: 90618725-90618725 on Build GRCh38
rs990907076 Chr.5: 90618965-90618965 on Build GRCh38
rs1046611054 Chr.5: 90618734-90618734 on Build GRCh38
rs1031898067 Chr.5: 90619073-90619073 on Build GRCh38
rs531920516 Chr.5: 90618877-90618877 on Build GRCh38
rs201592369 Chr.5: 90619089-90619089 on Build GRCh38
rs914744762 Chr.5: 90619100-90619100 on Build GRCh38
rs1016051929 Chr.5: 90618923-90618923 on Build GRCh38
rs1673379 Chr.5: 90618870-90618870 on Build GRCh38
rs990328641 Chr.5: 90618752-90618752 on Build GRCh38
rs751316421 Chr.5: 90618905-90618905 on Build GRCh38
rs915909627 Chr.5: 90618668-90618668 on Build GRCh38
rs934791991 Chr.5: 90618738-90618738 on Build GRCh38
rs16868853 Chr.5: 90618937-90618937 on Build GRCh38
rs924880681 Chr.5: 90618737-90618737 on Build GRCh38
rs180844992 Chr.5: 90618785-90618785 on Build GRCh38
rs751886247 Chr.5: 90619072-90619072 on Build GRCh38
rs534005881 Chr.5: 90618974-90618974 on Build GRCh38
rs764455116 Chr.5: 90619057-90619057 on Build GRCh38
rs776657073 Chr.5: 90618995-90618998 on Build GRCh38
rs186234349 Chr.5: 90619095-90619095 on Build GRCh38
rs554253879 Chr.5: 90619074-90619074 on Build GRCh38
rs10462487 Chr.5: 90618797-90618797 on Build GRCh38
rs371742461 Chr.5: 90618694-90618694 on Build GRCh38
rs971403813 Chr.5: 90618992-90618992 on Build GRCh38
rs763156083 Chr.5: 90619071-90619071 on Build GRCh38
rs914274364 Chr.5: 90618781-90618781 on Build GRCh38
rs62375078 Chr.5: 90618729-90618729 on Build GRCh38
rs753522355 Chr.5: 90619103-90619103 on Build GRCh38
rs560452612 Chr.5: 90618708-90618708 on Build GRCh38
rs933012509 Chr.5: 90618841-90618841 on Build GRCh38
rs916738564 Chr.5: 90618712-90618712 on Build GRCh38
rs901502218 Chr.5: 90618833-90618833 on Build GRCh38
rs41311333 Chr.5: 90619108-90619108 on Build GRCh38
rs949529145 Chr.5: 90619039-90619043 on Build GRCh38
rs1012992915 Chr.5: 90618952-90618952 on Build GRCh38
rs766519491 Chr.5: 90619089-90619089 on Build GRCh38

View Full Product Details