Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119431278-119431829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK5
Entrez Gene ID
2869
Gene Name
G protein-coupled receptor kinase 5
Gene Aliases
GPRK5
UniGene
Hs.524625
Gene Chromosome Location
Chr.10: 119207685-119455619 on Build GRCh38
Transcripts
NM_005308.2 XM_005269707.1 XM_005269708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1209002 c.646C>T p.R216C Chr.10: 119431435-119431435 on Build GRCh38
6518345 c.616C>A p.R206R Chr.10: 119431405-119431405 on Build GRCh38
4688856 c.665T>C p.I222T Chr.10: 119431454-119431454 on Build GRCh38
3806561 c.715G>A p.E239K Chr.10: 119431504-119431504 on Build GRCh38
4965672 c.738+9G>A p.? Chr.10: 119431536-119431536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2275038 Chr.10: 119431334-119431334 on Build GRCh38
rs545943145 Chr.10: 119431366-119431366 on Build GRCh38
rs779123615 Chr.10: 119431348-119431348 on Build GRCh38
rs929058584 Chr.10: 119431754-119431754 on Build GRCh38
rs372398313 Chr.10: 119431749-119431749 on Build GRCh38
rs375035540 Chr.10: 119431600-119431600 on Build GRCh38
rs140741672 Chr.10: 119431538-119431538 on Build GRCh38
rs1027950200 Chr.10: 119431760-119431760 on Build GRCh38
rs1022906437 Chr.10: 119431592-119431592 on Build GRCh38
rs550115144 Chr.10: 119431613-119431613 on Build GRCh38
rs761362012 Chr.10: 119431443-119431443 on Build GRCh38
rs1011590646 Chr.10: 119431766-119431766 on Build GRCh38
rs796192047 Chr.10: 119431743-119431743 on Build GRCh38
rs7099478 Chr.10: 119431674-119431674 on Build GRCh38
rs1022576616 Chr.10: 119431774-119431774 on Build GRCh38
rs370118216 Chr.10: 119431555-119431555 on Build GRCh38
rs12721545 Chr.10: 119431614-119431614 on Build GRCh38
rs967398207 Chr.10: 119431417-119431417 on Build GRCh38
rs564130593 Chr.10: 119431435-119431435 on Build GRCh38
rs12721550 Chr.10: 119431561-119431561 on Build GRCh38
rs925928770 Chr.10: 119431456-119431456 on Build GRCh38
rs202120111 Chr.10: 119431510-119431510 on Build GRCh38
rs757441015 Chr.10: 119431559-119431559 on Build GRCh38
rs887468357 Chr.10: 119431770-119431770 on Build GRCh38
rs939328357 Chr.10: 119431619-119431619 on Build GRCh38
rs767270791 Chr.10: 119431501-119431501 on Build GRCh38
rs1051275057 Chr.10: 119431564-119431564 on Build GRCh38
rs927970335 Chr.10: 119431322-119431322 on Build GRCh38
rs4752309 Chr.10: 119431729-119431729 on Build GRCh38
rs948647145 Chr.10: 119431677-119431677 on Build GRCh38
rs778571488 Chr.10: 119431573-119431573 on Build GRCh38
rs748330453 Chr.10: 119431349-119431349 on Build GRCh38
rs958881983 Chr.10: 119431328-119431328 on Build GRCh38
rs758704633 Chr.10: 119431358-119431358 on Build GRCh38
rs990851959 Chr.10: 119431654-119431654 on Build GRCh38
rs200647050 Chr.10: 119431554-119431554 on Build GRCh38
rs561429004 Chr.10: 119431599-119431599 on Build GRCh38
rs74643082 Chr.10: 119431696-119431696 on Build GRCh38
rs528343617 Chr.10: 119431448-119431448 on Build GRCh38
rs775987354 Chr.10: 119431408-119431409 on Build GRCh38
rs777710029 Chr.10: 119431367-119431367 on Build GRCh38
rs754375992 Chr.10: 119431518-119431518 on Build GRCh38
rs1006238306 Chr.10: 119431598-119431598 on Build GRCh38
rs866759998 Chr.10: 119431475-119431475 on Build GRCh38
rs578063095 Chr.10: 119431777-119431777 on Build GRCh38
rs757012905 Chr.10: 119431320-119431320 on Build GRCh38
rs770472713 Chr.10: 119431406-119431407 on Build GRCh38
rs890014443 Chr.10: 119431763-119431763 on Build GRCh38
rs555557988 Chr.10: 119431308-119431308 on Build GRCh38
rs563597805 Chr.10: 119431530-119431530 on Build GRCh38
rs777360386 Chr.10: 119431581-119431584 on Build GRCh38
rs986047396 Chr.10: 119431536-119431536 on Build GRCh38
rs533488194 Chr.10: 119431615-119431615 on Build GRCh38
rs55719738 Chr.10: 119431413-119431413 on Build GRCh38
rs769828788 Chr.10: 119431405-119431405 on Build GRCh38
rs760749742 Chr.10: 119431675-119431675 on Build GRCh38
rs569347181 Chr.10: 119431725-119431725 on Build GRCh38
rs546977938 Chr.10: 119431626-119431626 on Build GRCh38
rs755360711 Chr.10: 119431337-119431337 on Build GRCh38
rs1024395392 Chr.10: 119431618-119431618 on Build GRCh38
rs774000868 Chr.10: 119431436-119431436 on Build GRCh38
rs546124750 Chr.10: 119431785-119431785 on Build GRCh38
rs967259049 Chr.10: 119431780-119431780 on Build GRCh38
rs751637124 Chr.10: 119431556-119431556 on Build GRCh38
rs766824009 Chr.10: 119431504-119431504 on Build GRCh38
rs140965782 Chr.10: 119431502-119431502 on Build GRCh38
rs780068014 Chr.10: 119431560-119431560 on Build GRCh38
rs747121115 Chr.10: 119431378-119431378 on Build GRCh38
rs939893423 Chr.10: 119431732-119431732 on Build GRCh38
rs548292082 Chr.10: 119431591-119431591 on Build GRCh38
rs900965297 Chr.10: 119431744-119431744 on Build GRCh38
rs760228524 Chr.10: 119431503-119431503 on Build GRCh38
rs567082719 Chr.10: 119431656-119431656 on Build GRCh38
rs112871819 Chr.10: 119431511-119431511 on Build GRCh38
rs944583319 Chr.10: 119431731-119431731 on Build GRCh38
rs775587738 Chr.10: 119431410-119431410 on Build GRCh38
rs748020504 Chr.10: 119431577-119431577 on Build GRCh38
rs1004387219 Chr.10: 119431519-119431519 on Build GRCh38

View Full Product Details