Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109668816-109669365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTM2
Entrez Gene ID
2946
Gene Name
glutathione S-transferase mu 2 (muscle)
Gene Aliases
GST4, GSTM, GSTM2-2, GTHMUS
UniGene
Hs.279837
Gene Chromosome Location
Chr.1: 109668022-109683997 on Build GRCh38
Transcripts
NM_000848.3 NM_001142368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1294692 c.141G>A p.L47L Chr.1: 109668953-109668953 on Build GRCh38
7102852 c.219C>T p.S73S Chr.1: 109669331-109669331 on Build GRCh38
1193737 c.166G>T p.D56Y Chr.1: 109668978-109668978 on Build GRCh38
4020428 c.245G>A p.R82H Chr.1: 109669357-109669357 on Build GRCh38
4633232 c.141G>C p.L47L Chr.1: 109668953-109668953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111477734 Chr.1: 109669251-109669251 on Build GRCh38
rs371310525 Chr.1: 109668883-109668883 on Build GRCh38
rs924229081 Chr.1: 109669138-109669138 on Build GRCh38
rs1036096997 Chr.1: 109669345-109669345 on Build GRCh38
rs592792 Chr.1: 109669334-109669334 on Build GRCh38
rs772473017 Chr.1: 109669241-109669241 on Build GRCh38
rs753438244 Chr.1: 109668922-109668922 on Build GRCh38
rs780542067 Chr.1: 109669027-109669027 on Build GRCh38
rs897765713 Chr.1: 109669060-109669060 on Build GRCh38
rs866258791 Chr.1: 109669278-109669278 on Build GRCh38
rs990953018 Chr.1: 109668853-109668853 on Build GRCh38
rs946718113 Chr.1: 109669118-109669118 on Build GRCh38
rs570094396 Chr.1: 109669107-109669107 on Build GRCh38
rs762631602 Chr.1: 109669014-109669014 on Build GRCh38
rs779218262 Chr.1: 109668983-109668983 on Build GRCh38
rs755479703 Chr.1: 109669322-109669322 on Build GRCh38
rs752792325 Chr.1: 109668990-109668990 on Build GRCh38
rs562313739 Chr.1: 109668953-109668953 on Build GRCh38
rs767431563 Chr.1: 109668905-109668905 on Build GRCh38
rs374179090 Chr.1: 109668909-109668909 on Build GRCh38
rs188487928 Chr.1: 109669028-109669028 on Build GRCh38
rs758585781 Chr.1: 109669340-109669340 on Build GRCh38
rs753611820 Chr.1: 109669073-109669073 on Build GRCh38
rs201923867 Chr.1: 109668946-109668946 on Build GRCh38
rs1019040765 Chr.1: 109668938-109668938 on Build GRCh38
rs760507992 Chr.1: 109668996-109668996 on Build GRCh38
rs759022986 Chr.1: 109669009-109669009 on Build GRCh38
rs770640171 Chr.1: 109669268-109669268 on Build GRCh38
rs768622566 Chr.1: 109668957-109668957 on Build GRCh38
rs944924302 Chr.1: 109669146-109669146 on Build GRCh38
rs758994986 Chr.1: 109668923-109668923 on Build GRCh38
rs373932116 Chr.1: 109669083-109669083 on Build GRCh38
rs755070116 Chr.1: 109669031-109669031 on Build GRCh38
rs765775699 Chr.1: 109668920-109668920 on Build GRCh38
rs402505 Chr.1: 109669213-109669213 on Build GRCh38
rs2227960 Chr.1: 109669311-109669311 on Build GRCh38
rs776467717 Chr.1: 109669008-109669008 on Build GRCh38
rs761042411 Chr.1: 109669292-109669292 on Build GRCh38
rs750733142 Chr.1: 109669025-109669025 on Build GRCh38
rs2229047 Chr.1: 109669346-109669346 on Build GRCh38
rs996585547 Chr.1: 109669198-109669198 on Build GRCh38
rs754364196 Chr.1: 109669319-109669319 on Build GRCh38
rs1002818452 Chr.1: 109669072-109669072 on Build GRCh38
rs147235683 Chr.1: 109669345-109669345 on Build GRCh38
rs143027391 Chr.1: 109668929-109668929 on Build GRCh38
rs961256783 Chr.1: 109669016-109669016 on Build GRCh38
rs959485000 Chr.1: 109669066-109669066 on Build GRCh38
rs374930518 Chr.1: 109669277-109669277 on Build GRCh38
rs900675870 Chr.1: 109669192-109669192 on Build GRCh38
rs1055133470 Chr.1: 109668859-109668859 on Build GRCh38
rs768873693 Chr.1: 109668876-109668876 on Build GRCh38
rs764670031 Chr.1: 109669010-109669010 on Build GRCh38
rs778079639 Chr.1: 109669344-109669344 on Build GRCh38
rs756466063 Chr.1: 109669026-109669026 on Build GRCh38
rs397834986 Chr.1: 109668837-109668837 on Build GRCh38
rs779203820 Chr.1: 109669037-109669037 on Build GRCh38
rs958602337 Chr.1: 109669077-109669077 on Build GRCh38
rs368017907 Chr.1: 109668936-109668936 on Build GRCh38
rs765386523 Chr.1: 109669323-109669323 on Build GRCh38
rs748351281 Chr.1: 109669039-109669039 on Build GRCh38
rs1042755868 Chr.1: 109669126-109669126 on Build GRCh38
rs748050285 Chr.1: 109669260-109669289 on Build GRCh38
rs879046352 Chr.1: 109668992-109668992 on Build GRCh38
rs781600778 Chr.1: 109668940-109668940 on Build GRCh38
rs181297205 Chr.1: 109669271-109669271 on Build GRCh38
rs750896418 Chr.1: 109668944-109668944 on Build GRCh38
rs376678043 Chr.1: 109668868-109668868 on Build GRCh38
rs199939618 Chr.1: 109669015-109669015 on Build GRCh38
rs770857267 Chr.1: 109669007-109669007 on Build GRCh38
rs1013123138 Chr.1: 109669091-109669091 on Build GRCh38
rs201645830 Chr.1: 109669259-109669259 on Build GRCh38
rs367858323 Chr.1: 109669282-109669282 on Build GRCh38
rs403337 Chr.1: 109668886-109668886 on Build GRCh38
rs769261240 Chr.1: 109669272-109669272 on Build GRCh38
rs428135 Chr.1: 109669059-109669059 on Build GRCh38
rs773182714 Chr.1: 109668994-109668994 on Build GRCh38
rs771990135 Chr.1: 109668989-109668989 on Build GRCh38
rs749445198 Chr.1: 109668875-109668875 on Build GRCh38
rs935560853 Chr.1: 109668911-109668911 on Build GRCh38
rs773995935 Chr.1: 109669286-109669286 on Build GRCh38
rs41283500 Chr.1: 109669184-109669184 on Build GRCh38
rs371911407 Chr.1: 109668988-109668988 on Build GRCh38
rs545475320 Chr.1: 109668847-109668847 on Build GRCh38
rs746783959 Chr.1: 109669260-109669260 on Build GRCh38
rs1049473216 Chr.1: 109669210-109669210 on Build GRCh38
rs1005232743 Chr.1: 109669240-109669240 on Build GRCh38
rs371258412 Chr.1: 109669283-109669283 on Build GRCh38
rs403294 Chr.1: 109668932-109668932 on Build GRCh38
rs956692472 Chr.1: 109669215-109669215 on Build GRCh38
rs550297391 Chr.1: 109669088-109669088 on Build GRCh38

View Full Product Details