Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42179717-42180261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCA1A
Entrez Gene ID
2978
Gene Name
guanylate cyclase activator 1A
Gene Aliases
C6orf131, COD3, CORD14, GCAP, GCAP1, GUCA, GUCA1
UniGene
Hs.92858
Gene Chromosome Location
Chr.6: 42155377-42180083 on Build GRCh38
Transcripts
NM_000409.4 NM_001319061.1 NM_001319062.1 XM_006715073.3 XM_011514537.2 XM_011514539.1

SNP Detail

SNP ID Chromosome Location
rs919574898 Chr.6: 42179883-42179883 on Build GRCh38
rs958464570 Chr.6: 42179877-42179877 on Build GRCh38
rs1026696244 Chr.6: 42180234-42180234 on Build GRCh38
rs886577802 Chr.6: 42179743-42179743 on Build GRCh38
rs951475915 Chr.6: 42180164-42180164 on Build GRCh38
rs142239616 Chr.6: 42180135-42180135 on Build GRCh38
rs996692150 Chr.6: 42179779-42179779 on Build GRCh38
rs1019975063 Chr.6: 42180105-42180105 on Build GRCh38
rs115822911 Chr.6: 42180199-42180199 on Build GRCh38
rs568311923 Chr.6: 42180093-42180093 on Build GRCh38
rs749422610 Chr.6: 42180154-42180154 on Build GRCh38
rs115636833 Chr.6: 42179748-42179748 on Build GRCh38
rs957375088 Chr.6: 42179809-42179809 on Build GRCh38
rs577697141 Chr.6: 42179890-42179890 on Build GRCh38
rs182201426 Chr.6: 42179818-42179818 on Build GRCh38
rs981264848 Chr.6: 42179870-42179870 on Build GRCh38
rs886061392 Chr.6: 42179951-42179951 on Build GRCh38
rs968973817 Chr.6: 42180109-42180109 on Build GRCh38
rs770888865 Chr.6: 42179848-42179848 on Build GRCh38
rs932060078 Chr.6: 42180094-42180094 on Build GRCh38
rs76894479 Chr.6: 42180088-42180088 on Build GRCh38
rs545710600 Chr.6: 42179955-42179955 on Build GRCh38
rs1034626264 Chr.6: 42179871-42179871 on Build GRCh38
rs1032995321 Chr.6: 42179798-42179798 on Build GRCh38
rs771878848 Chr.6: 42180192-42180192 on Build GRCh38
rs1037520138 Chr.6: 42179933-42179933 on Build GRCh38
rs190097335 Chr.6: 42180020-42180020 on Build GRCh38
rs533125883 Chr.6: 42180217-42180217 on Build GRCh38
rs1000880239 Chr.6: 42180111-42180111 on Build GRCh38
rs893108000 Chr.6: 42180116-42180116 on Build GRCh38
rs992871061 Chr.6: 42180136-42180136 on Build GRCh38
rs1010243239 Chr.6: 42180118-42180118 on Build GRCh38
rs941959301 Chr.6: 42179932-42179932 on Build GRCh38
rs144595632 Chr.6: 42179904-42179904 on Build GRCh38
rs774225319 Chr.6: 42180194-42180194 on Build GRCh38
rs1003227830 Chr.6: 42180159-42180159 on Build GRCh38
rs944991400 Chr.6: 42180069-42180069 on Build GRCh38
rs1020219999 Chr.6: 42179843-42179843 on Build GRCh38
rs111640279 Chr.6: 42180092-42180092 on Build GRCh38
rs575889559 Chr.6: 42180112-42180112 on Build GRCh38
rs546703052 Chr.6: 42180236-42180236 on Build GRCh38
rs780275035 Chr.6: 42179900-42179900 on Build GRCh38
rs983630875 Chr.6: 42179951-42179951 on Build GRCh38
rs549270436 Chr.6: 42179747-42179747 on Build GRCh38
rs917242378 Chr.6: 42179884-42179885 on Build GRCh38
rs774560916 Chr.6: 42179760-42179760 on Build GRCh38
rs1038055234 Chr.6: 42180228-42180228 on Build GRCh38
rs56285673 Chr.6: 42180030-42180030 on Build GRCh38
rs900360037 Chr.6: 42180012-42180012 on Build GRCh38
rs937424279 Chr.6: 42179862-42179862 on Build GRCh38
rs929654998 Chr.6: 42179896-42179896 on Build GRCh38
rs186789929 Chr.6: 42179957-42179957 on Build GRCh38
rs749051608 Chr.6: 42179746-42179746 on Build GRCh38
rs969384561 Chr.6: 42179790-42179790 on Build GRCh38
rs886327541 Chr.6: 42179906-42179906 on Build GRCh38
rs1020331279 Chr.6: 42180120-42180120 on Build GRCh38

View Full Product Details