Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44828149-44828701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H2AFV
Entrez Gene ID
94239
Gene Name
H2A histone family member V
Gene Aliases
H2A.Z-2, H2AV
UniGene
Hs.488189
Gene Chromosome Location
Chr.7: 44826889-44848126 on Build GRCh38
Transcripts
NM_138635.3

SNP Detail

SNP ID Chromosome Location
rs1029842880 Chr.7: 44828186-44828186 on Build GRCh38
rs991882410 Chr.7: 44828317-44828317 on Build GRCh38
rs571552548 Chr.7: 44828627-44828627 on Build GRCh38
rs747606817 Chr.7: 44828605-44828605 on Build GRCh38
rs77705479 Chr.7: 44828611-44828611 on Build GRCh38
rs908906845 Chr.7: 44828540-44828540 on Build GRCh38
rs971322379 Chr.7: 44828423-44828423 on Build GRCh38
rs1032345813 Chr.7: 44828336-44828336 on Build GRCh38
rs563275661 Chr.7: 44828546-44828546 on Build GRCh38
rs562757235 Chr.7: 44828406-44828406 on Build GRCh38
rs576727844 Chr.7: 44828248-44828248 on Build GRCh38
rs1047388166 Chr.7: 44828225-44828225 on Build GRCh38
rs941975753 Chr.7: 44828235-44828235 on Build GRCh38
rs946630830 Chr.7: 44828483-44828483 on Build GRCh38
rs1023397303 Chr.7: 44828593-44828593 on Build GRCh38
rs540780566 Chr.7: 44828393-44828393 on Build GRCh38
rs1039787806 Chr.7: 44828502-44828502 on Build GRCh38
rs933736770 Chr.7: 44828506-44828506 on Build GRCh38
rs951486401 Chr.7: 44828221-44828221 on Build GRCh38
rs567714917 Chr.7: 44828655-44828655 on Build GRCh38
rs927804735 Chr.7: 44828389-44828389 on Build GRCh38
rs1051809082 Chr.7: 44828284-44828284 on Build GRCh38
rs935454541 Chr.7: 44828613-44828613 on Build GRCh38
rs142787279 Chr.7: 44828566-44828566 on Build GRCh38
rs983189951 Chr.7: 44828505-44828505 on Build GRCh38
rs981515934 Chr.7: 44828346-44828346 on Build GRCh38
rs975081420 Chr.7: 44828587-44828587 on Build GRCh38
rs915890245 Chr.7: 44828323-44828323 on Build GRCh38
rs201226730 Chr.7: 44828661-44828661 on Build GRCh38
rs1004373262 Chr.7: 44828556-44828556 on Build GRCh38
rs1013140084 Chr.7: 44828588-44828588 on Build GRCh38
rs908874163 Chr.7: 44828229-44828229 on Build GRCh38
rs928970270 Chr.7: 44828225-44828227 on Build GRCh38
rs11544537 Chr.7: 44828468-44828468 on Build GRCh38
rs971384146 Chr.7: 44828604-44828604 on Build GRCh38
rs898761642 Chr.7: 44828579-44828579 on Build GRCh38
rs11770684 Chr.7: 44828503-44828503 on Build GRCh38
rs367796657 Chr.7: 44828387-44828390 on Build GRCh38
rs1050827048 Chr.7: 44828538-44828538 on Build GRCh38
rs559363494 Chr.7: 44828413-44828413 on Build GRCh38
rs1004508723 Chr.7: 44828243-44828243 on Build GRCh38
rs956561638 Chr.7: 44828619-44828619 on Build GRCh38
rs1038937250 Chr.7: 44828241-44828241 on Build GRCh38
rs958059488 Chr.7: 44828378-44828378 on Build GRCh38
rs971901507 Chr.7: 44828325-44828325 on Build GRCh38
rs962774047 Chr.7: 44828254-44828254 on Build GRCh38

View Full Product Details