Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99950068-99950614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H2AFZ
Entrez Gene ID
3015
Gene Name
H2A histone family member Z
Gene Aliases
H2A.Z-1, H2A.z, H2A/z, H2AZ
UniGene
Hs.119192
Gene Chromosome Location
Chr.4: 99948087-99950355 on Build GRCh38
Transcripts
NM_002106.3 NR_125923.1

Gene Symbol
LOC256880
Entrez Gene ID
256880
Gene Name
uncharacterized LOC256880
Gene Aliases
-
UniGene
Hs.448554
Gene Chromosome Location
Chr.4: 99950494-100037705 on Build GRCh38
Transcripts
NM_002106.3 NR_125923.1

SNP Detail

SNP ID Chromosome Location
rs761283253 Chr.4: 99950217-99950217 on Build GRCh38
rs915150357 Chr.4: 99950111-99950111 on Build GRCh38
rs755919205 Chr.4: 99950223-99950223 on Build GRCh38
rs930651449 Chr.4: 99950571-99950571 on Build GRCh38
rs976026887 Chr.4: 99950498-99950498 on Build GRCh38
rs1022115895 Chr.4: 99950409-99950409 on Build GRCh38
rs1041617238 Chr.4: 99950526-99950526 on Build GRCh38
rs745774401 Chr.4: 99950180-99950180 on Build GRCh38
rs989297011 Chr.4: 99950407-99950407 on Build GRCh38
rs948905342 Chr.4: 99950588-99950588 on Build GRCh38
rs564728683 Chr.4: 99950250-99950250 on Build GRCh38
rs566661390 Chr.4: 99950459-99950459 on Build GRCh38
rs951003611 Chr.4: 99950447-99950447 on Build GRCh38
rs972023763 Chr.4: 99950559-99950559 on Build GRCh38
rs953206491 Chr.4: 99950425-99950425 on Build GRCh38
rs1013091957 Chr.4: 99950299-99950299 on Build GRCh38
rs1053859508 Chr.4: 99950515-99950515 on Build GRCh38
rs1000955299 Chr.4: 99950229-99950229 on Build GRCh38
rs995681283 Chr.4: 99950591-99950591 on Build GRCh38
rs1003657721 Chr.4: 99950324-99950324 on Build GRCh38
rs766850582 Chr.4: 99950139-99950139 on Build GRCh38
rs944190472 Chr.4: 99950470-99950470 on Build GRCh38
rs948889955 Chr.4: 99950494-99950494 on Build GRCh38
rs916033560 Chr.4: 99950578-99950578 on Build GRCh38
rs558965865 Chr.4: 99950516-99950516 on Build GRCh38
rs1050354093 Chr.4: 99950138-99950138 on Build GRCh38
rs986032656 Chr.4: 99950427-99950427 on Build GRCh38
rs1049242785 Chr.4: 99950268-99950268 on Build GRCh38
rs542816803 Chr.4: 99950146-99950146 on Build GRCh38
rs986408375 Chr.4: 99950545-99950545 on Build GRCh38
rs936782285 Chr.4: 99950503-99950503 on Build GRCh38
rs912786227 Chr.4: 99950464-99950464 on Build GRCh38
rs892487129 Chr.4: 99950517-99950517 on Build GRCh38
rs761236197 Chr.4: 99950136-99950136 on Build GRCh38
rs763525436 Chr.4: 99950133-99950133 on Build GRCh38
rs919324041 Chr.4: 99950466-99950466 on Build GRCh38
rs894664014 Chr.4: 99950278-99950278 on Build GRCh38
rs577179857 Chr.4: 99950541-99950541 on Build GRCh38
rs1010134655 Chr.4: 99950241-99950241 on Build GRCh38
rs989344524 Chr.4: 99950089-99950089 on Build GRCh38
rs944722063 Chr.4: 99950462-99950463 on Build GRCh38
rs762616085 Chr.4: 99950124-99950124 on Build GRCh38
rs544980949 Chr.4: 99950088-99950088 on Build GRCh38
rs370608688 Chr.4: 99950220-99950220 on Build GRCh38
rs373152482 Chr.4: 99950227-99950227 on Build GRCh38
rs748898430 Chr.4: 99950205-99950205 on Build GRCh38
rs768666100 Chr.4: 99950537-99950537 on Build GRCh38
rs970379006 Chr.4: 99950314-99950314 on Build GRCh38
rs532091895 Chr.4: 99950253-99950253 on Build GRCh38
rs566602444 Chr.4: 99950537-99950538 on Build GRCh38
rs137864981 Chr.4: 99950224-99950224 on Build GRCh38
rs773908326 Chr.4: 99950212-99950212 on Build GRCh38
rs1020136191 Chr.4: 99950254-99950254 on Build GRCh38
rs958732733 Chr.4: 99950362-99950362 on Build GRCh38
rs945343460 Chr.4: 99950523-99950523 on Build GRCh38
rs747075872 Chr.4: 99950153-99950153 on Build GRCh38
rs984798967 Chr.4: 99950574-99950574 on Build GRCh38
rs548357401 Chr.4: 99950393-99950393 on Build GRCh38
rs966380302 Chr.4: 99950554-99950554 on Build GRCh38
rs377144974 Chr.4: 99950135-99950135 on Build GRCh38
rs892156398 Chr.4: 99950518-99950518 on Build GRCh38
rs1010632970 Chr.4: 99950529-99950529 on Build GRCh38
rs768348413 Chr.4: 99950210-99950210 on Build GRCh38
rs973836096 Chr.4: 99950366-99950366 on Build GRCh38
rs1003079407 Chr.4: 99950275-99950275 on Build GRCh38
rs560995494 Chr.4: 99950203-99950203 on Build GRCh38
rs376055860 Chr.4: 99950256-99950256 on Build GRCh38
rs372241950 Chr.4: 99950215-99950215 on Build GRCh38
rs1030745739 Chr.4: 99950335-99950335 on Build GRCh38
rs1034182170 Chr.4: 99950339-99950339 on Build GRCh38
rs781242350 Chr.4: 99950159-99950159 on Build GRCh38
rs765588625 Chr.4: 99950144-99950144 on Build GRCh38
rs907165431 Chr.4: 99950536-99950536 on Build GRCh38
rs534023465 Chr.4: 99950499-99950499 on Build GRCh38
rs894289776 Chr.4: 99950560-99950560 on Build GRCh38
rs758584043 Chr.4: 99950150-99950150 on Build GRCh38
rs1054232455 Chr.4: 99950230-99950230 on Build GRCh38
rs892455351 Chr.4: 99950237-99950237 on Build GRCh38
rs536473560 Chr.4: 99950349-99950349 on Build GRCh38
rs1017725802 Chr.4: 99950504-99950504 on Build GRCh38
rs757306566 Chr.4: 99950158-99950158 on Build GRCh38
rs1024983960 Chr.4: 99950307-99950307 on Build GRCh38
rs930705154 Chr.4: 99950468-99950468 on Build GRCh38
rs1006375244 Chr.4: 99950480-99950480 on Build GRCh38
rs1049147420 Chr.4: 99950482-99950482 on Build GRCh38
rs894613017 Chr.4: 99950493-99950493 on Build GRCh38
rs747895861 Chr.4: 99950345-99950345 on Build GRCh38
rs1034625892 Chr.4: 99950552-99950552 on Build GRCh38
rs919594387 Chr.4: 99950098-99950098 on Build GRCh38
rs755330454 Chr.4: 99950142-99950142 on Build GRCh38
rs769495557 Chr.4: 99950190-99950190 on Build GRCh38
rs775032760 Chr.4: 99950118-99950118 on Build GRCh38
rs575366571 Chr.4: 99950140-99950140 on Build GRCh38
rs569504818 Chr.4: 99950294-99950294 on Build GRCh38

View Full Product Details