Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 31306686-31307236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR5A
Entrez Gene ID
25938
Gene Name
HEAT repeat containing 5A
Gene Aliases
C14orf125
UniGene
Hs.744979
Gene Chromosome Location
Chr.14: 31291788-31420582 on Build GRCh38
Transcripts
NM_015473.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4428137 c.4934G>C p.R1645T Chr.14: 31306746-31306746 on Build GRCh38
6585996 c.4919_4921delATG p.H1640_V1641>L Chr.14: 31306759-31306761 on Build GRCh38
5891476 c.4936C>G p.R1646G Chr.14: 31306744-31306744 on Build GRCh38
5178716 c.4907C>T p.A1636V Chr.14: 31306773-31306773 on Build GRCh38
6841675 c.4936C>T p.R1646* Chr.14: 31306744-31306744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763584258 Chr.14: 31306798-31306798 on Build GRCh38
rs909057311 Chr.14: 31307046-31307046 on Build GRCh38
rs558562996 Chr.14: 31307136-31307136 on Build GRCh38
rs563776038 Chr.14: 31307049-31307049 on Build GRCh38
rs201553100 Chr.14: 31306778-31306778 on Build GRCh38
rs553069825 Chr.14: 31307185-31307185 on Build GRCh38
rs528220201 Chr.14: 31306859-31306859 on Build GRCh38
rs765793969 Chr.14: 31306802-31306802 on Build GRCh38
rs747183967 Chr.14: 31306760-31306760 on Build GRCh38
rs372893569 Chr.14: 31306729-31306729 on Build GRCh38
rs781507843 Chr.14: 31306876-31306876 on Build GRCh38
rs976807489 Chr.14: 31306750-31306750 on Build GRCh38
rs377240432 Chr.14: 31306744-31306744 on Build GRCh38
rs929097105 Chr.14: 31307014-31307014 on Build GRCh38
rs907280976 Chr.14: 31307101-31307101 on Build GRCh38
rs1030644956 Chr.14: 31307089-31307089 on Build GRCh38
rs541001895 Chr.14: 31307143-31307143 on Build GRCh38
rs557070264 Chr.14: 31306960-31306960 on Build GRCh38
rs773790453 Chr.14: 31306793-31306793 on Build GRCh38
rs780831642 Chr.14: 31306719-31306719 on Build GRCh38
rs544679245 Chr.14: 31306858-31306858 on Build GRCh38
rs570444545 Chr.14: 31306848-31306848 on Build GRCh38
rs895085352 Chr.14: 31307070-31307070 on Build GRCh38
rs767024499 Chr.14: 31306921-31306921 on Build GRCh38
rs1031806708 Chr.14: 31307169-31307169 on Build GRCh38
rs897805060 Chr.14: 31307003-31307003 on Build GRCh38
rs1017662535 Chr.14: 31306940-31306940 on Build GRCh38
rs749739876 Chr.14: 31306890-31306890 on Build GRCh38
rs893328913 Chr.14: 31307180-31307180 on Build GRCh38
rs754721479 Chr.14: 31306711-31306711 on Build GRCh38
rs267603979 Chr.14: 31306743-31306743 on Build GRCh38
rs148868323 Chr.14: 31307083-31307083 on Build GRCh38
rs375990603 Chr.14: 31306895-31306895 on Build GRCh38
rs535299834 Chr.14: 31306930-31306930 on Build GRCh38
rs552240410 Chr.14: 31306845-31306845 on Build GRCh38
rs759041359 Chr.14: 31306916-31306916 on Build GRCh38
rs983240160 Chr.14: 31307033-31307033 on Build GRCh38
rs780243441 Chr.14: 31306751-31306751 on Build GRCh38
rs17091516 Chr.14: 31306770-31306770 on Build GRCh38
rs546739835 Chr.14: 31306883-31306883 on Build GRCh38
rs371300043 Chr.14: 31306785-31306785 on Build GRCh38
rs767118544 Chr.14: 31306830-31306830 on Build GRCh38
rs1055063590 Chr.14: 31307068-31307068 on Build GRCh38
rs563838390 Chr.14: 31306768-31306768 on Build GRCh38
rs990405431 Chr.14: 31306932-31306932 on Build GRCh38
rs542412780 Chr.14: 31306981-31306981 on Build GRCh38
rs923025362 Chr.14: 31307006-31307006 on Build GRCh38
rs759083049 Chr.14: 31306820-31306820 on Build GRCh38
rs757272638 Chr.14: 31306855-31306855 on Build GRCh38
rs941911706 Chr.14: 31307062-31307062 on Build GRCh38
rs1007132137 Chr.14: 31307043-31307043 on Build GRCh38
rs956334199 Chr.14: 31307098-31307098 on Build GRCh38
rs914986604 Chr.14: 31307106-31307106 on Build GRCh38
rs530194529 Chr.14: 31306857-31306857 on Build GRCh38
rs771974895 Chr.14: 31306749-31306749 on Build GRCh38
rs368393016 Chr.14: 31306910-31306910 on Build GRCh38
rs775114358 Chr.14: 31306726-31306729 on Build GRCh38
rs755937244 Chr.14: 31306735-31306735 on Build GRCh38
rs1027704234 Chr.14: 31307025-31307025 on Build GRCh38
rs964420526 Chr.14: 31306950-31306950 on Build GRCh38
rs770279241 Chr.14: 31306877-31306877 on Build GRCh38
rs35341014 Chr.14: 31307111-31307111 on Build GRCh38
rs994925032 Chr.14: 31307015-31307016 on Build GRCh38
rs768138378 Chr.14: 31306926-31306928 on Build GRCh38
rs989199808 Chr.14: 31307105-31307105 on Build GRCh38
rs576804212 Chr.14: 31307141-31307141 on Build GRCh38
rs755843937 Chr.14: 31306846-31306846 on Build GRCh38
rs768647340 Chr.14: 31306874-31306874 on Build GRCh38
rs751132937 Chr.14: 31306810-31306810 on Build GRCh38
rs1024381269 Chr.14: 31306838-31306838 on Build GRCh38
rs1046187388 Chr.14: 31307092-31307092 on Build GRCh38
rs1038741562 Chr.14: 31306762-31306762 on Build GRCh38
rs746845471 Chr.14: 31306889-31306891 on Build GRCh38
rs774754634 Chr.14: 31306914-31306914 on Build GRCh38

View Full Product Details