Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18938542-18939088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOMER3
Entrez Gene ID
9454
Gene Name
homer scaffolding protein 3
Gene Aliases
HOMER-3, VESL3
UniGene
Hs.720208
Gene Chromosome Location
Chr.19: 18929201-18941258 on Build GRCh38
Transcripts
NM_001145721.1 NM_001145722.1 NM_001145724.1 NM_004838.3 NR_027297.1 XM_006722943.1 XM_006722944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6026167 c.105C>T p.L35L Chr.19: 18938794-18938794 on Build GRCh38
7095644 c.15-1G>A p.? Chr.19: 18938885-18938885 on Build GRCh38
6255236 c.45C>A p.H15Q Chr.19: 18938854-18938854 on Build GRCh38
6681506 c.99C>T p.H33H Chr.19: 18938800-18938800 on Build GRCh38
6681507 c.99C>T p.H33H Chr.19: 18938800-18938800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767982331 Chr.19: 18938812-18938812 on Build GRCh38
rs769261242 Chr.19: 18938790-18938790 on Build GRCh38
rs1013084282 Chr.19: 18938580-18938580 on Build GRCh38
rs750189511 Chr.19: 18938946-18938946 on Build GRCh38
rs756501786 Chr.19: 18939049-18939049 on Build GRCh38
rs771483227 Chr.19: 18939021-18939021 on Build GRCh38
rs182760613 Chr.19: 18938838-18938838 on Build GRCh38
rs747633654 Chr.19: 18939007-18939007 on Build GRCh38
rs946935446 Chr.19: 18939055-18939055 on Build GRCh38
rs374794386 Chr.19: 18938893-18938893 on Build GRCh38
rs764071294 Chr.19: 18938653-18938653 on Build GRCh38
rs759881904 Chr.19: 18939030-18939030 on Build GRCh38
rs765428463 Chr.19: 18938919-18938919 on Build GRCh38
rs776993357 Chr.19: 18939027-18939027 on Build GRCh38
rs757264120 Chr.19: 18938939-18938939 on Build GRCh38
rs557445776 Chr.19: 18938951-18938951 on Build GRCh38
rs779711800 Chr.19: 18938880-18938880 on Build GRCh38
rs774707487 Chr.19: 18938889-18938889 on Build GRCh38
rs894718549 Chr.19: 18938913-18938913 on Build GRCh38
rs980874803 Chr.19: 18938987-18938987 on Build GRCh38
rs564270653 Chr.19: 18938591-18938591 on Build GRCh38
rs939013539 Chr.19: 18938899-18938899 on Build GRCh38
rs139040912 Chr.19: 18938810-18938810 on Build GRCh38
rs752131888 Chr.19: 18938587-18938587 on Build GRCh38
rs762268645 Chr.19: 18938742-18938742 on Build GRCh38
rs758363323 Chr.19: 18938768-18938768 on Build GRCh38
rs1016470706 Chr.19: 18938593-18938594 on Build GRCh38
rs759679355 Chr.19: 18938911-18938911 on Build GRCh38
rs372209530 Chr.19: 18938705-18938705 on Build GRCh38
rs756145131 Chr.19: 18938754-18938754 on Build GRCh38
rs774926070 Chr.19: 18938809-18938809 on Build GRCh38
rs766645513 Chr.19: 18938902-18938902 on Build GRCh38
rs200689130 Chr.19: 18938826-18938826 on Build GRCh38
rs374896199 Chr.19: 18938783-18938783 on Build GRCh38
rs781734612 Chr.19: 18938766-18938766 on Build GRCh38
rs778535110 Chr.19: 18939006-18939006 on Build GRCh38
rs150734422 Chr.19: 18938799-18938799 on Build GRCh38
rs367777459 Chr.19: 18938785-18938785 on Build GRCh38
rs371721055 Chr.19: 18938776-18938776 on Build GRCh38
rs750670213 Chr.19: 18938753-18938753 on Build GRCh38
rs751417446 Chr.19: 18938923-18938923 on Build GRCh38
rs780322294 Chr.19: 18938761-18938761 on Build GRCh38
rs35399855 Chr.19: 18938740-18938740 on Build GRCh38
rs773598753 Chr.19: 18939011-18939011 on Build GRCh38
rs747323107 Chr.19: 18939017-18939017 on Build GRCh38
rs899347566 Chr.19: 18938943-18938943 on Build GRCh38
rs763487122 Chr.19: 18938733-18938733 on Build GRCh38
rs748998186 Chr.19: 18938993-18938993 on Build GRCh38
rs914160895 Chr.19: 18939052-18939052 on Build GRCh38
rs371617955 Chr.19: 18938854-18938854 on Build GRCh38
rs750581782 Chr.19: 18938823-18938823 on Build GRCh38
rs569415353 Chr.19: 18938857-18938857 on Build GRCh38
rs772302100 Chr.19: 18938894-18938894 on Build GRCh38
rs993548759 Chr.19: 18938989-18938989 on Build GRCh38
rs1026817146 Chr.19: 18938994-18938994 on Build GRCh38
rs748728371 Chr.19: 18938881-18938881 on Build GRCh38
rs373905922 Chr.19: 18938716-18938716 on Build GRCh38
rs577845446 Chr.19: 18939062-18939062 on Build GRCh38
rs1043834975 Chr.19: 18938752-18938752 on Build GRCh38
rs910455552 Chr.19: 18938787-18938787 on Build GRCh38
rs533238982 Chr.19: 18938672-18938672 on Build GRCh38
rs754480091 Chr.19: 18939003-18939003 on Build GRCh38
rs767375398 Chr.19: 18938944-18938944 on Build GRCh38
rs149871070 Chr.19: 18938862-18938862 on Build GRCh38
rs768167673 Chr.19: 18938888-18938888 on Build GRCh38
rs777009637 Chr.19: 18938910-18938910 on Build GRCh38
rs8111121 Chr.19: 18938689-18938689 on Build GRCh38
rs781077555 Chr.19: 18938859-18938859 on Build GRCh38
rs374923493 Chr.19: 18938801-18938801 on Build GRCh38
rs761009617 Chr.19: 18938825-18938825 on Build GRCh38
rs775968603 Chr.19: 18938722-18938722 on Build GRCh38
rs753930843 Chr.19: 18938828-18938828 on Build GRCh38
rs371683344 Chr.19: 18938800-18938800 on Build GRCh38
rs777800588 Chr.19: 18938848-18938848 on Build GRCh38
rs764417013 Chr.19: 18938739-18938739 on Build GRCh38
rs948082266 Chr.19: 18938626-18938626 on Build GRCh38
rs777892611 Chr.19: 18938775-18938775 on Build GRCh38
rs769933351 Chr.19: 18938950-18938950 on Build GRCh38
rs750834938 Chr.19: 18938861-18938861 on Build GRCh38
rs773578948 Chr.19: 18938896-18938896 on Build GRCh38
rs376592730 Chr.19: 18938687-18938687 on Build GRCh38
rs199645270 Chr.19: 18938874-18938874 on Build GRCh38
rs954853947 Chr.19: 18938995-18938995 on Build GRCh38
rs928024798 Chr.19: 18938696-18938696 on Build GRCh38
rs759471688 Chr.19: 18938920-18938921 on Build GRCh38
rs772594020 Chr.19: 18939008-18939008 on Build GRCh38
rs899600717 Chr.19: 18938760-18938760 on Build GRCh38
rs761154056 Chr.19: 18938900-18938900 on Build GRCh38
rs775025710 Chr.19: 18938796-18938796 on Build GRCh38
rs939328964 Chr.19: 18938709-18938709 on Build GRCh38
rs878976182 Chr.19: 18938949-18938950 on Build GRCh38
rs374227320 Chr.19: 18939060-18939060 on Build GRCh38
rs767567385 Chr.19: 18938748-18938748 on Build GRCh38

View Full Product Details