Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27173922-27174420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA10
Entrez Gene ID
3206
Gene Name
homeobox A10
Gene Aliases
HOX1, HOX1.8, HOX1H, PL
UniGene
Hs.110637
Gene Chromosome Location
Chr.7: 27170591-27180261 on Build GRCh38
Transcripts
NM_018951.3 NR_037939.1 NR_037940.1

Gene Symbol
HOXA10-HOXA9
Entrez Gene ID
100534589
Gene Name
HOXA10-HOXA9 readthrough
Gene Aliases
-
UniGene
Hs.659350
Gene Chromosome Location
Chr.7: 27162438-27180261 on Build GRCh38
Transcripts
NM_018951.3 NR_037939.1 NR_037940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6681568 c.101delG p.G34fs*53 Chr.7: 27174155-27174155 on Build GRCh38
6359840 c.251G>A p.G84E Chr.7: 27174005-27174005 on Build GRCh38
4690958 c.55A>C p.S19R Chr.7: 27174201-27174201 on Build GRCh38
1292490 c.27G>A p.A9A Chr.7: 27174229-27174229 on Build GRCh38
5681359 c.285C>T p.P95P Chr.7: 27173971-27173971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758088260 Chr.7: 27174106-27174106 on Build GRCh38
rs990501473 Chr.7: 27174189-27174189 on Build GRCh38
rs776118615 Chr.7: 27174149-27174172 on Build GRCh38
rs752233773 Chr.7: 27174103-27174103 on Build GRCh38
rs900580293 Chr.7: 27174346-27174346 on Build GRCh38
rs202173149 Chr.7: 27174208-27174208 on Build GRCh38
rs758538241 Chr.7: 27174185-27174185 on Build GRCh38
rs752283373 Chr.7: 27174158-27174158 on Build GRCh38
rs772971961 Chr.7: 27174128-27174128 on Build GRCh38
rs774838700 Chr.7: 27174206-27174206 on Build GRCh38
rs750802108 Chr.7: 27174160-27174160 on Build GRCh38
rs746505663 Chr.7: 27174139-27174140 on Build GRCh38
rs774090455 Chr.7: 27174223-27174223 on Build GRCh38
rs774373521 Chr.7: 27174056-27174056 on Build GRCh38
rs373421816 Chr.7: 27174291-27174291 on Build GRCh38
rs781630554 Chr.7: 27174002-27174002 on Build GRCh38
rs765521585 Chr.7: 27174342-27174342 on Build GRCh38
rs776849742 Chr.7: 27174333-27174333 on Build GRCh38
rs916409472 Chr.7: 27174197-27174197 on Build GRCh38
rs771715381 Chr.7: 27174059-27174059 on Build GRCh38
rs760576037 Chr.7: 27174069-27174069 on Build GRCh38
rs945024648 Chr.7: 27174248-27174248 on Build GRCh38
rs760014602 Chr.7: 27174335-27174335 on Build GRCh38
rs773022987 Chr.7: 27174064-27174064 on Build GRCh38
rs770757784 Chr.7: 27174145-27174146 on Build GRCh38
rs778939773 Chr.7: 27174004-27174005 on Build GRCh38
rs866631902 Chr.7: 27173982-27173982 on Build GRCh38
rs746323844 Chr.7: 27174003-27174003 on Build GRCh38
rs780248307 Chr.7: 27174024-27174024 on Build GRCh38
rs747182497 Chr.7: 27174247-27174247 on Build GRCh38
rs921007073 Chr.7: 27173983-27173983 on Build GRCh38
rs1003665653 Chr.7: 27174085-27174085 on Build GRCh38
rs764114704 Chr.7: 27174352-27174352 on Build GRCh38
rs748311251 Chr.7: 27174178-27174178 on Build GRCh38
rs748283182 Chr.7: 27174126-27174126 on Build GRCh38
rs753331027 Chr.7: 27174097-27174097 on Build GRCh38
rs34720764 Chr.7: 27173943-27173943 on Build GRCh38
rs747233773 Chr.7: 27174187-27174187 on Build GRCh38
rs914983745 Chr.7: 27174230-27174230 on Build GRCh38
rs763886371 Chr.7: 27174159-27174159 on Build GRCh38
rs1053845544 Chr.7: 27174013-27174013 on Build GRCh38
rs965322614 Chr.7: 27174147-27174147 on Build GRCh38
rs772925194 Chr.7: 27174138-27174139 on Build GRCh38
rs749818947 Chr.7: 27174121-27174121 on Build GRCh38
rs935217045 Chr.7: 27174218-27174218 on Build GRCh38
rs768233328 Chr.7: 27174324-27174324 on Build GRCh38
rs994905785 Chr.7: 27174105-27174105 on Build GRCh38
rs199591141 Chr.7: 27174331-27174331 on Build GRCh38
rs766809355 Chr.7: 27174229-27174229 on Build GRCh38
rs759112411 Chr.7: 27174100-27174100 on Build GRCh38
rs777840276 Chr.7: 27174244-27174244 on Build GRCh38
rs779072679 Chr.7: 27174125-27174125 on Build GRCh38
rs778033462 Chr.7: 27174186-27174186 on Build GRCh38
rs764598649 Chr.7: 27174157-27174157 on Build GRCh38
rs568043829 Chr.7: 27174051-27174051 on Build GRCh38
rs749198971 Chr.7: 27174310-27174310 on Build GRCh38
rs755422334 Chr.7: 27174237-27174237 on Build GRCh38
rs143452717 Chr.7: 27174161-27174166 on Build GRCh38
rs774029295 Chr.7: 27174325-27174325 on Build GRCh38
rs757497712 Chr.7: 27174262-27174262 on Build GRCh38
rs750857163 Chr.7: 27174118-27174118 on Build GRCh38
rs771722221 Chr.7: 27174061-27174061 on Build GRCh38
rs756665289 Chr.7: 27174120-27174120 on Build GRCh38
rs776428407 Chr.7: 27174194-27174194 on Build GRCh38
rs749439718 Chr.7: 27174025-27174025 on Build GRCh38
rs932446610 Chr.7: 27173992-27173992 on Build GRCh38
rs372671579 Chr.7: 27174207-27174207 on Build GRCh38
rs950555260 Chr.7: 27174119-27174119 on Build GRCh38
rs752718123 Chr.7: 27174239-27174239 on Build GRCh38
rs1040938030 Chr.7: 27173975-27173975 on Build GRCh38
rs755545078 Chr.7: 27174163-27174163 on Build GRCh38
rs906788961 Chr.7: 27174266-27174266 on Build GRCh38
rs538881123 Chr.7: 27174172-27174172 on Build GRCh38
rs780579803 Chr.7: 27174161-27174161 on Build GRCh38
rs535450007 Chr.7: 27174058-27174058 on Build GRCh38
rs765834065 Chr.7: 27174238-27174238 on Build GRCh38
rs951023413 Chr.7: 27174372-27174372 on Build GRCh38
rs983110503 Chr.7: 27174155-27174155 on Build GRCh38
rs745723503 Chr.7: 27174290-27174290 on Build GRCh38
rs1028226128 Chr.7: 27174359-27174359 on Build GRCh38
rs766355235 Chr.7: 27174076-27174076 on Build GRCh38
rs781368034 Chr.7: 27174110-27174110 on Build GRCh38
rs759973639 Chr.7: 27174152-27174172 on Build GRCh38
rs1043616734 Chr.7: 27174006-27174006 on Build GRCh38
rs1026578178 Chr.7: 27174116-27174116 on Build GRCh38
rs758404063 Chr.7: 27174243-27174243 on Build GRCh38
rs894956881 Chr.7: 27173956-27173956 on Build GRCh38
rs1027698163 Chr.7: 27174362-27174362 on Build GRCh38
rs998901060 Chr.7: 27174090-27174090 on Build GRCh38
rs765467369 Chr.7: 27174341-27174342 on Build GRCh38
rs770651896 Chr.7: 27174192-27174192 on Build GRCh38
rs770848155 Chr.7: 27174143-27174143 on Build GRCh38
rs570291423 Chr.7: 27174137-27174137 on Build GRCh38
rs1003781721 Chr.7: 27174267-27174267 on Build GRCh38
rs778831085 Chr.7: 27174176-27174176 on Build GRCh38
rs1033846356 Chr.7: 27174087-27174087 on Build GRCh38
rs780934546 Chr.7: 27174287-27174287 on Build GRCh38
rs768561565 Chr.7: 27174123-27174123 on Build GRCh38
rs754274142 Chr.7: 27174235-27174235 on Build GRCh38
rs951027963 Chr.7: 27174154-27174154 on Build GRCh38
rs776482213 Chr.7: 27174153-27174153 on Build GRCh38
rs139581121 Chr.7: 27174378-27174378 on Build GRCh38
rs761581039 Chr.7: 27174226-27174226 on Build GRCh38
rs747986816 Chr.7: 27174005-27174010 on Build GRCh38
rs753388180 Chr.7: 27173940-27173940 on Build GRCh38
rs747393933 Chr.7: 27173973-27173973 on Build GRCh38
rs753186988 Chr.7: 27174347-27174347 on Build GRCh38
rs908456392 Chr.7: 27174145-27174145 on Build GRCh38
rs368847177 Chr.7: 27174141-27174141 on Build GRCh38
rs893449741 Chr.7: 27174016-27174016 on Build GRCh38
rs778485760 Chr.7: 27173958-27173958 on Build GRCh38
rs770451301 Chr.7: 27174018-27174018 on Build GRCh38
rs556571937 Chr.7: 27174102-27174102 on Build GRCh38
rs768920878 Chr.7: 27174029-27174029 on Build GRCh38
rs902456062 Chr.7: 27174215-27174215 on Build GRCh38
rs769808963 Chr.7: 27174205-27174205 on Build GRCh38

View Full Product Details