Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83295121-83295665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPSE
Entrez Gene ID
10855
Gene Name
heparanase
Gene Aliases
HPA, HPA1, HPR1, HPSE1, HSE1
UniGene
Hs.44227
Gene Chromosome Location
Chr.4: 83292461-83335153 on Build GRCh38
Transcripts
NM_001098540.2 NM_001166498.2 NM_001199830.1 NM_006665.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6101288 c.1498C>T p.L500L Chr.4: 83295478-83295478 on Build GRCh38
734456 c.1576G>A p.A526T Chr.4: 83295400-83295400 on Build GRCh38
3409583 c.1506G>C p.M502I Chr.4: 83295470-83295470 on Build GRCh38
6048241 c.1596delT p.F532fs*2 Chr.4: 83295380-83295380 on Build GRCh38
448245 c.1587_1588delTA p.Y529fs*1 Chr.4: 83295388-83295389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75695154 Chr.4: 83295573-83295573 on Build GRCh38
rs752676303 Chr.4: 83295297-83295305 on Build GRCh38
rs778180409 Chr.4: 83295533-83295533 on Build GRCh38
rs139515004 Chr.4: 83295151-83295151 on Build GRCh38
rs753321321 Chr.4: 83295403-83295404 on Build GRCh38
rs6856901 Chr.4: 83295215-83295215 on Build GRCh38
rs776427696 Chr.4: 83295325-83295325 on Build GRCh38
rs1046191144 Chr.4: 83295602-83295602 on Build GRCh38
rs139500120 Chr.4: 83295473-83295473 on Build GRCh38
rs762787392 Chr.4: 83295480-83295480 on Build GRCh38
rs925176552 Chr.4: 83295553-83295553 on Build GRCh38
rs756732153 Chr.4: 83295406-83295410 on Build GRCh38
rs570847655 Chr.4: 83295331-83295331 on Build GRCh38
rs576008054 Chr.4: 83295521-83295521 on Build GRCh38
rs771371566 Chr.4: 83295534-83295534 on Build GRCh38
rs756023219 Chr.4: 83295307-83295307 on Build GRCh38
rs1015077396 Chr.4: 83295463-83295463 on Build GRCh38
rs763103308 Chr.4: 83295548-83295548 on Build GRCh38
rs987951106 Chr.4: 83295597-83295598 on Build GRCh38
rs759407538 Chr.4: 83295422-83295422 on Build GRCh38
rs920118122 Chr.4: 83295371-83295371 on Build GRCh38
rs576844478 Chr.4: 83295568-83295568 on Build GRCh38
rs748501273 Chr.4: 83295441-83295441 on Build GRCh38
rs913706901 Chr.4: 83295598-83295598 on Build GRCh38
rs540856329 Chr.4: 83295161-83295161 on Build GRCh38
rs779711147 Chr.4: 83295525-83295525 on Build GRCh38
rs759625684 Chr.4: 83295457-83295457 on Build GRCh38
rs548047868 Chr.4: 83295261-83295261 on Build GRCh38
rs569637487 Chr.4: 83295266-83295266 on Build GRCh38
rs539217123 Chr.4: 83295562-83295562 on Build GRCh38
rs754032506 Chr.4: 83295328-83295328 on Build GRCh38
rs991856793 Chr.4: 83295291-83295291 on Build GRCh38
rs1020806286 Chr.4: 83295183-83295183 on Build GRCh38
rs778487365 Chr.4: 83295427-83295428 on Build GRCh38
rs557656789 Chr.4: 83295518-83295518 on Build GRCh38
rs145553296 Chr.4: 83295389-83295389 on Build GRCh38
rs936502837 Chr.4: 83295259-83295259 on Build GRCh38
rs931518753 Chr.4: 83295391-83295391 on Build GRCh38
rs773627443 Chr.4: 83295308-83295308 on Build GRCh38
rs770997326 Chr.4: 83295452-83295452 on Build GRCh38
rs80122687 Chr.4: 83295315-83295315 on Build GRCh38
rs772559711 Chr.4: 83295299-83295299 on Build GRCh38
rs752479765 Chr.4: 83295364-83295364 on Build GRCh38
rs926236098 Chr.4: 83295257-83295257 on Build GRCh38
rs765434485 Chr.4: 83295459-83295459 on Build GRCh38
rs543434903 Chr.4: 83295526-83295526 on Build GRCh38
rs543399017 Chr.4: 83295580-83295580 on Build GRCh38
rs755732529 Chr.4: 83295431-83295431 on Build GRCh38
rs149701279 Chr.4: 83295330-83295330 on Build GRCh38
rs775591944 Chr.4: 83295512-83295512 on Build GRCh38
rs758761736 Chr.4: 83295527-83295527 on Build GRCh38
rs758133160 Chr.4: 83295388-83295388 on Build GRCh38
rs144185023 Chr.4: 83295496-83295496 on Build GRCh38
rs6855404 Chr.4: 83295217-83295217 on Build GRCh38
rs959302725 Chr.4: 83295598-83295598 on Build GRCh38
rs922901693 Chr.4: 83295196-83295196 on Build GRCh38
rs755239721 Chr.4: 83295290-83295291 on Build GRCh38
rs781335966 Chr.4: 83295519-83295519 on Build GRCh38
rs571906118 Chr.4: 83295481-83295481 on Build GRCh38
rs558571724 Chr.4: 83295541-83295541 on Build GRCh38
rs751596282 Chr.4: 83295403-83295403 on Build GRCh38
rs745921688 Chr.4: 83295535-83295535 on Build GRCh38
rs199555797 Chr.4: 83295551-83295551 on Build GRCh38
rs925182511 Chr.4: 83295219-83295219 on Build GRCh38
rs386676731 Chr.4: 83295215-83295217 on Build GRCh38
rs200924614 Chr.4: 83295427-83295427 on Build GRCh38
rs367786140 Chr.4: 83295433-83295433 on Build GRCh38
rs372237278 Chr.4: 83295338-83295338 on Build GRCh38
rs371462727 Chr.4: 83295448-83295448 on Build GRCh38
rs777639789 Chr.4: 83295379-83295380 on Build GRCh38
rs747557302 Chr.4: 83295451-83295451 on Build GRCh38
rs1037581677 Chr.4: 83295340-83295340 on Build GRCh38
rs764275173 Chr.4: 83295623-83295623 on Build GRCh38
rs907163888 Chr.4: 83295378-83295378 on Build GRCh38
rs373648825 Chr.4: 83295426-83295426 on Build GRCh38
rs767572781 Chr.4: 83295515-83295515 on Build GRCh38
rs772349039 Chr.4: 83295447-83295447 on Build GRCh38
rs776752498 Chr.4: 83295453-83295453 on Build GRCh38
rs568670121 Chr.4: 83295489-83295489 on Build GRCh38
rs763934869 Chr.4: 83295552-83295552 on Build GRCh38

View Full Product Details