Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 96835703-96836204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS6ST3
Entrez Gene ID
266722
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Gene Aliases
HS6ST-3
UniGene
Hs.171001
Gene Chromosome Location
Chr.13: 96090634-96839562 on Build GRCh38
Transcripts
NM_153456.3

SNP Detail

SNP ID Chromosome Location
rs986729597 Chr.13: 96835943-96835943 on Build GRCh38
rs747819784 Chr.13: 96836024-96836031 on Build GRCh38
rs903913582 Chr.13: 96835750-96835750 on Build GRCh38
rs1016777169 Chr.13: 96836129-96836129 on Build GRCh38
rs556576706 Chr.13: 96836087-96836087 on Build GRCh38
rs1039136294 Chr.13: 96835754-96835754 on Build GRCh38
rs914124119 Chr.13: 96835955-96835955 on Build GRCh38
rs1004022961 Chr.13: 96835795-96835795 on Build GRCh38
rs886959503 Chr.13: 96835793-96835793 on Build GRCh38
rs3742123 Chr.13: 96835862-96835862 on Build GRCh38
rs962893419 Chr.13: 96836121-96836121 on Build GRCh38
rs553717720 Chr.13: 96836104-96836104 on Build GRCh38
rs999754110 Chr.13: 96835844-96835844 on Build GRCh38
rs1043803428 Chr.13: 96835736-96835736 on Build GRCh38
rs901826139 Chr.13: 96836045-96836045 on Build GRCh38
rs974258699 Chr.13: 96836131-96836131 on Build GRCh38
rs75428341 Chr.13: 96835997-96835997 on Build GRCh38
rs113528661 Chr.13: 96835934-96835934 on Build GRCh38
rs551709474 Chr.13: 96835987-96835987 on Build GRCh38
rs749554636 Chr.13: 96836170-96836170 on Build GRCh38
rs1012493590 Chr.13: 96836096-96836096 on Build GRCh38
rs560911592 Chr.13: 96835839-96835839 on Build GRCh38
rs530034008 Chr.13: 96835849-96835849 on Build GRCh38
rs899783537 Chr.13: 96835773-96835773 on Build GRCh38
rs1023394437 Chr.13: 96835903-96835903 on Build GRCh38
rs898275921 Chr.13: 96836134-96836134 on Build GRCh38
rs1007974574 Chr.13: 96835759-96835759 on Build GRCh38
rs902517496 Chr.13: 96835828-96835828 on Build GRCh38
rs375778420 Chr.13: 96835911-96835911 on Build GRCh38
rs1039475667 Chr.13: 96836023-96836023 on Build GRCh38
rs3864 Chr.13: 96836095-96836095 on Build GRCh38
rs531990022 Chr.13: 96835983-96835983 on Build GRCh38
rs754892407 Chr.13: 96835792-96835792 on Build GRCh38
rs962812400 Chr.13: 96835904-96835904 on Build GRCh38
rs80022048 Chr.13: 96836070-96836070 on Build GRCh38
rs943457986 Chr.13: 96836007-96836007 on Build GRCh38
rs186849870 Chr.13: 96836166-96836166 on Build GRCh38
rs539035520 Chr.13: 96836113-96836113 on Build GRCh38
rs151202437 Chr.13: 96835861-96835861 on Build GRCh38
rs979530083 Chr.13: 96835992-96835992 on Build GRCh38
rs558378750 Chr.13: 96836090-96836090 on Build GRCh38
rs747908952 Chr.13: 96835985-96835985 on Build GRCh38
rs145731698 Chr.13: 96836105-96836105 on Build GRCh38
rs558986566 Chr.13: 96836145-96836145 on Build GRCh38
rs1010913860 Chr.13: 96835774-96835774 on Build GRCh38
rs575600612 Chr.13: 96835763-96835763 on Build GRCh38
rs1001941533 Chr.13: 96836159-96836159 on Build GRCh38
rs1033336909 Chr.13: 96835910-96835910 on Build GRCh38
rs992386846 Chr.13: 96835935-96835935 on Build GRCh38
rs962454335 Chr.13: 96835845-96835845 on Build GRCh38
rs912460365 Chr.13: 96835957-96835957 on Build GRCh38
rs990911666 Chr.13: 96835884-96835884 on Build GRCh38
rs770978876 Chr.13: 96836173-96836173 on Build GRCh38
rs10492534 Chr.13: 96835776-96835776 on Build GRCh38
rs1039031312 Chr.13: 96835767-96835767 on Build GRCh38
rs551188640 Chr.13: 96836081-96836081 on Build GRCh38
rs961270633 Chr.13: 96835930-96835930 on Build GRCh38
rs1043798177 Chr.13: 96836081-96836082 on Build GRCh38

View Full Product Details