Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55368476-55368980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL11
Entrez Gene ID
3589
Gene Name
interleukin 11
Gene Aliases
AGIF, IL-11
UniGene
Hs.467304
Gene Chromosome Location
Chr.19: 55364382-55370463 on Build GRCh38
Transcripts
NM_000641.3 NM_001267718.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
137187 c.227C>T p.P76L Chr.19: 55368523-55368523 on Build GRCh38
4847835 c.257G>A p.G86E Chr.19: 55368493-55368493 on Build GRCh38
1396575 c.125C>T p.T42I Chr.19: 55368824-55368824 on Build GRCh38
1396574 c.202G>A p.G68R Chr.19: 55368548-55368548 on Build GRCh38
7371999 c.100G>C p.D34H Chr.19: 55368849-55368849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758437473 Chr.19: 55368896-55368896 on Build GRCh38
rs758809983 Chr.19: 55368519-55368519 on Build GRCh38
rs1051129166 Chr.19: 55368668-55368668 on Build GRCh38
rs201067487 Chr.19: 55368933-55368933 on Build GRCh38
rs926655964 Chr.19: 55368616-55368616 on Build GRCh38
rs748856545 Chr.19: 55368760-55368760 on Build GRCh38
rs570837384 Chr.19: 55368838-55368838 on Build GRCh38
rs891097603 Chr.19: 55368702-55368702 on Build GRCh38
rs539881082 Chr.19: 55368906-55368906 on Build GRCh38
rs773998938 Chr.19: 55368505-55368505 on Build GRCh38
rs748079395 Chr.19: 55368548-55368548 on Build GRCh38
rs773143821 Chr.19: 55368809-55368809 on Build GRCh38
rs764519258 Chr.19: 55368839-55368839 on Build GRCh38
rs755768865 Chr.19: 55368748-55368748 on Build GRCh38
rs1017496871 Chr.19: 55368795-55368795 on Build GRCh38
rs1016866170 Chr.19: 55368510-55368510 on Build GRCh38
rs867527711 Chr.19: 55368911-55368911 on Build GRCh38
rs759771192 Chr.19: 55368823-55368823 on Build GRCh38
rs779630554 Chr.19: 55368538-55368538 on Build GRCh38
rs778078221 Chr.19: 55368524-55368524 on Build GRCh38
rs752601728 Chr.19: 55368921-55368922 on Build GRCh38
rs753476624 Chr.19: 55368856-55368856 on Build GRCh38
rs771202937 Chr.19: 55368810-55368810 on Build GRCh38
rs901341360 Chr.19: 55368704-55368704 on Build GRCh38
rs375031113 Chr.19: 55368825-55368825 on Build GRCh38
rs770584215 Chr.19: 55368705-55368705 on Build GRCh38
rs753240724 Chr.19: 55368600-55368600 on Build GRCh38
rs747592950 Chr.19: 55368879-55368879 on Build GRCh38
rs527461248 Chr.19: 55368669-55368669 on Build GRCh38
rs746133596 Chr.19: 55368919-55368919 on Build GRCh38
rs757325072 Chr.19: 55368723-55368723 on Build GRCh38
rs753799788 Chr.19: 55368500-55368500 on Build GRCh38
rs930040769 Chr.19: 55368666-55368666 on Build GRCh38
rs758900122 Chr.19: 55368612-55368612 on Build GRCh38
rs776799433 Chr.19: 55368585-55368585 on Build GRCh38
rs751952561 Chr.19: 55368847-55368847 on Build GRCh38
rs760988555 Chr.19: 55368563-55368563 on Build GRCh38
rs748165583 Chr.19: 55368797-55368797 on Build GRCh38
rs979895815 Chr.19: 55368961-55368961 on Build GRCh38
rs945599049 Chr.19: 55368694-55368694 on Build GRCh38
rs757518582 Chr.19: 55368852-55368852 on Build GRCh38
rs751866348 Chr.19: 55368726-55368726 on Build GRCh38
rs776767433 Chr.19: 55368811-55368811 on Build GRCh38
rs368133522 Chr.19: 55368959-55368959 on Build GRCh38
rs955968619 Chr.19: 55368499-55368499 on Build GRCh38
rs548038066 Chr.19: 55368587-55368587 on Build GRCh38
rs150100243 Chr.19: 55368806-55368806 on Build GRCh38
rs971000324 Chr.19: 55368924-55368924 on Build GRCh38
rs531922469 Chr.19: 55368718-55368718 on Build GRCh38
rs1042589040 Chr.19: 55368698-55368698 on Build GRCh38
rs183174646 Chr.19: 55368945-55368945 on Build GRCh38
rs767906633 Chr.19: 55368855-55368855 on Build GRCh38
rs774111814 Chr.19: 55368546-55368546 on Build GRCh38
rs980341731 Chr.19: 55368925-55368925 on Build GRCh38
rs568268577 Chr.19: 55368649-55368649 on Build GRCh38
rs762981585 Chr.19: 55368935-55368935 on Build GRCh38
rs531513552 Chr.19: 55368549-55368549 on Build GRCh38
rs770002734 Chr.19: 55368923-55368923 on Build GRCh38
rs937999938 Chr.19: 55368627-55368627 on Build GRCh38
rs138170385 Chr.19: 55368537-55368537 on Build GRCh38
rs779248310 Chr.19: 55368506-55368506 on Build GRCh38
rs773125492 Chr.19: 55368557-55368557 on Build GRCh38
rs766441538 Chr.19: 55368577-55368577 on Build GRCh38
rs568521538 Chr.19: 55368939-55368939 on Build GRCh38
rs754658079 Chr.19: 55368864-55368864 on Build GRCh38
rs1008820792 Chr.19: 55368768-55368768 on Build GRCh38
rs754499717 Chr.19: 55368781-55368781 on Build GRCh38
rs897259854 Chr.19: 55368634-55368634 on Build GRCh38
rs141630906 Chr.19: 55368545-55368545 on Build GRCh38
rs770670953 Chr.19: 55368909-55368909 on Build GRCh38
rs778457489 Chr.19: 55368787-55368787 on Build GRCh38
rs368402118 Chr.19: 55368918-55368918 on Build GRCh38
rs753127489 Chr.19: 55368508-55368508 on Build GRCh38
rs778297416 Chr.19: 55368878-55368878 on Build GRCh38
rs775535260 Chr.19: 55368837-55368837 on Build GRCh38
rs765337084 Chr.19: 55368594-55368594 on Build GRCh38
rs1038929416 Chr.19: 55368619-55368619 on Build GRCh38
rs915512017 Chr.19: 55368638-55368638 on Build GRCh38
rs867906886 Chr.19: 55368791-55368791 on Build GRCh38
rs779533258 Chr.19: 55368758-55368758 on Build GRCh38
rs745589252 Chr.19: 55368525-55368525 on Build GRCh38

View Full Product Details