Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6034406-6034813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNAB2
Entrez Gene ID
8514
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 2
Gene Aliases
AKR6A5, HKvbeta2, HKvbeta2.1, HKvbeta2.2, KCNA2B, KV-BETA-2
UniGene
Hs.440497
Gene Chromosome Location
Chr.1: 5992298-6101193 on Build GRCh38
Transcripts
NM_001199860.1 NM_001199861.1 NM_003636.3 NM_172130.2 XM_005263514.2 XM_011542321.2 XM_011542322.2 XM_017002618.1 XM_017002619.1 XM_017002621.1

SNP Detail

SNP ID Chromosome Location
rs527697847 Chr.1: 6034447-6034447 on Build GRCh38
rs150796221 Chr.1: 6034565-6034565 on Build GRCh38
rs890641092 Chr.1: 6034532-6034532 on Build GRCh38
rs942852338 Chr.1: 6034522-6034522 on Build GRCh38
rs931477600 Chr.1: 6034777-6034777 on Build GRCh38
rs1039685284 Chr.1: 6034681-6034681 on Build GRCh38
rs537269261 Chr.1: 6034504-6034504 on Build GRCh38
rs1024365887 Chr.1: 6034730-6034730 on Build GRCh38
rs77094171 Chr.1: 6034733-6034733 on Build GRCh38
rs781558022 Chr.1: 6034600-6034600 on Build GRCh38
rs11584686 Chr.1: 6034751-6034751 on Build GRCh38
rs868096037 Chr.1: 6034545-6034545 on Build GRCh38
rs987484268 Chr.1: 6034489-6034489 on Build GRCh38
rs756495017 Chr.1: 6034627-6034627 on Build GRCh38
rs542359620 Chr.1: 6034563-6034563 on Build GRCh38
rs989149855 Chr.1: 6034781-6034781 on Build GRCh38
rs1046075866 Chr.1: 6034597-6034597 on Build GRCh38
rs1001328959 Chr.1: 6034638-6034638 on Build GRCh38
rs899379067 Chr.1: 6034691-6034691 on Build GRCh38
rs886455058 Chr.1: 6034711-6034711 on Build GRCh38
rs1024061636 Chr.1: 6034593-6034593 on Build GRCh38
rs912047586 Chr.1: 6034653-6034653 on Build GRCh38
rs912948726 Chr.1: 6034492-6034492 on Build GRCh38
rs548363980 Chr.1: 6034449-6034449 on Build GRCh38
rs12076209 Chr.1: 6034740-6034740 on Build GRCh38
rs943555283 Chr.1: 6034660-6034660 on Build GRCh38
rs1295095 Chr.1: 6034534-6034534 on Build GRCh38
rs1001342569 Chr.1: 6034739-6034739 on Build GRCh38
rs114215906 Chr.1: 6034453-6034453 on Build GRCh38
rs995497836 Chr.1: 6034699-6034699 on Build GRCh38
rs755622372 Chr.1: 6034725-6034725 on Build GRCh38
rs912907785 Chr.1: 6034620-6034620 on Build GRCh38
rs994881169 Chr.1: 6034564-6034564 on Build GRCh38
rs933731391 Chr.1: 6034495-6034495 on Build GRCh38
rs577501300 Chr.1: 6034540-6034540 on Build GRCh38
rs561224151 Chr.1: 6034779-6034779 on Build GRCh38
rs1047699527 Chr.1: 6034704-6034704 on Build GRCh38
rs967801986 Chr.1: 6034732-6034732 on Build GRCh38
rs138107254 Chr.1: 6034588-6034588 on Build GRCh38
rs190444541 Chr.1: 6034601-6034601 on Build GRCh38
rs775122371 Chr.1: 6034771-6034771 on Build GRCh38
rs755579801 Chr.1: 6034599-6034599 on Build GRCh38
rs576161508 Chr.1: 6034701-6034701 on Build GRCh38
rs935721578 Chr.1: 6034435-6034435 on Build GRCh38
rs1050860938 Chr.1: 6034636-6034636 on Build GRCh38
rs1034187005 Chr.1: 6034647-6034647 on Build GRCh38
rs1014279047 Chr.1: 6034716-6034716 on Build GRCh38
rs560971127 Chr.1: 6034613-6034613 on Build GRCh38
rs751581643 Chr.1: 6034595-6034595 on Build GRCh38
rs866030499 Chr.1: 6034518-6034518 on Build GRCh38
rs1012103460 Chr.1: 6034696-6034696 on Build GRCh38
rs950760746 Chr.1: 6034566-6034566 on Build GRCh38

View Full Product Details