Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44925009-44925504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF18B
Entrez Gene ID
146909
Gene Name
kinesin family member 18B
Gene Aliases
-
UniGene
Hs.135094
Gene Chromosome Location
Chr.17: 44924711-44947746 on Build GRCh38
Transcripts
NM_001264573.1 NM_001265577.1 XM_011524385.2 XM_011524386.2 XM_011524387.2 XM_011524388.2 XM_011524389.2 XM_011524390.2

SNP Detail

SNP ID Chromosome Location
rs1019586951 Chr.17: 44925182-44925182 on Build GRCh38
rs893652937 Chr.17: 44925071-44925071 on Build GRCh38
rs534905426 Chr.17: 44925075-44925075 on Build GRCh38
rs771974813 Chr.17: 44925176-44925176 on Build GRCh38
rs991655965 Chr.17: 44925389-44925389 on Build GRCh38
rs1010750895 Chr.17: 44925303-44925303 on Build GRCh38
rs1010971586 Chr.17: 44925101-44925101 on Build GRCh38
rs982571985 Chr.17: 44925243-44925243 on Build GRCh38
rs1011704587 Chr.17: 44925308-44925308 on Build GRCh38
rs962084713 Chr.17: 44925353-44925353 on Build GRCh38
rs975816110 Chr.17: 44925480-44925480 on Build GRCh38
rs1011694797 Chr.17: 44925167-44925167 on Build GRCh38
rs893389994 Chr.17: 44925307-44925307 on Build GRCh38
rs542248064 Chr.17: 44925315-44925315 on Build GRCh38
rs576689438 Chr.17: 44925407-44925407 on Build GRCh38
rs552223618 Chr.17: 44925179-44925179 on Build GRCh38
rs111305569 Chr.17: 44925336-44925336 on Build GRCh38
rs937660543 Chr.17: 44925276-44925276 on Build GRCh38
rs949035309 Chr.17: 44925113-44925113 on Build GRCh38
rs141842046 Chr.17: 44925319-44925319 on Build GRCh38
rs986817403 Chr.17: 44925039-44925039 on Build GRCh38
rs893603968 Chr.17: 44925297-44925297 on Build GRCh38
rs373356612 Chr.17: 44925435-44925435 on Build GRCh38
rs778260189 Chr.17: 44925179-44925180 on Build GRCh38
rs778695814 Chr.17: 44925151-44925151 on Build GRCh38
rs975382101 Chr.17: 44925198-44925198 on Build GRCh38
rs571920568 Chr.17: 44925222-44925222 on Build GRCh38
rs942602594 Chr.17: 44925061-44925061 on Build GRCh38
rs116017776 Chr.17: 44925143-44925143 on Build GRCh38
rs369923386 Chr.17: 44925132-44925132 on Build GRCh38
rs919833311 Chr.17: 44925216-44925216 on Build GRCh38
rs377614488 Chr.17: 44925476-44925476 on Build GRCh38
rs748068031 Chr.17: 44925173-44925173 on Build GRCh38
rs931120928 Chr.17: 44925089-44925089 on Build GRCh38
rs529944650 Chr.17: 44925131-44925131 on Build GRCh38
rs540920081 Chr.17: 44925416-44925416 on Build GRCh38
rs967388421 Chr.17: 44925030-44925030 on Build GRCh38
rs375478979 Chr.17: 44925060-44925060 on Build GRCh38
rs927887613 Chr.17: 44925259-44925259 on Build GRCh38
rs558534062 Chr.17: 44925391-44925391 on Build GRCh38
rs1046506093 Chr.17: 44925263-44925263 on Build GRCh38
rs1046880559 Chr.17: 44925093-44925093 on Build GRCh38
rs559296513 Chr.17: 44925114-44925114 on Build GRCh38
rs547146850 Chr.17: 44925108-44925108 on Build GRCh38
rs904881698 Chr.17: 44925275-44925275 on Build GRCh38
rs1031278193 Chr.17: 44925068-44925068 on Build GRCh38
rs919779234 Chr.17: 44925087-44925087 on Build GRCh38
rs537008587 Chr.17: 44925177-44925177 on Build GRCh38
rs7214258 Chr.17: 44925304-44925304 on Build GRCh38
rs920734645 Chr.17: 44925183-44925183 on Build GRCh38
rs1008333384 Chr.17: 44925323-44925323 on Build GRCh38
rs564804919 Chr.17: 44925036-44925036 on Build GRCh38
rs569765345 Chr.17: 44925172-44925172 on Build GRCh38
rs994134260 Chr.17: 44925377-44925377 on Build GRCh38
rs972246018 Chr.17: 44925181-44925181 on Build GRCh38
rs373484501 Chr.17: 44925378-44925378 on Build GRCh38
rs949136918 Chr.17: 44925258-44925258 on Build GRCh38
rs532297723 Chr.17: 44925038-44925038 on Build GRCh38
rs554489123 Chr.17: 44925271-44925271 on Build GRCh38
rs539350511 Chr.17: 44925251-44925251 on Build GRCh38
rs193194380 Chr.17: 44925379-44925379 on Build GRCh38
rs536878537 Chr.17: 44925317-44925317 on Build GRCh38
rs746440556 Chr.17: 44925474-44925474 on Build GRCh38
rs964369389 Chr.17: 44925188-44925188 on Build GRCh38
rs893262204 Chr.17: 44925447-44925447 on Build GRCh38
rs1026904702 Chr.17: 44925485-44925485 on Build GRCh38
rs574857212 Chr.17: 44925313-44925313 on Build GRCh38
rs893337673 Chr.17: 44925148-44925148 on Build GRCh38
rs913308707 Chr.17: 44925029-44925029 on Build GRCh38

View Full Product Details