Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46686711-46687224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB14
Entrez Gene ID
9813
Gene Name
EF-hand calcium binding domain 14
Gene Aliases
KIAA0494
UniGene
Hs.707688
Gene Chromosome Location
Chr.1: 46675159-46719064 on Build GRCh38
Transcripts
NM_014774.2 NR_038827.1 NR_038828.1

Gene Symbol
EFCAB14-AS1
Entrez Gene ID
100130197
Gene Name
EFCAB14 antisense RNA 1
Gene Aliases
KIAA0494-AS1
UniGene
Hs.568643
Gene Chromosome Location
Chr.1: 46674036-46692098 on Build GRCh38
Transcripts
NM_014774.2 NR_038827.1 NR_038828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
910356 c.1062C>T p.I354I Chr.1: 46686796-46686796 on Build GRCh38
7170796 c.994G>C p.D332H Chr.1: 46686864-46686864 on Build GRCh38
6332657 c.1030C>A p.Q344K Chr.1: 46686828-46686828 on Build GRCh38
1343166 c.1016G>T p.R339I Chr.1: 46686842-46686842 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs922912383 Chr.1: 46687034-46687034 on Build GRCh38
rs765049426 Chr.1: 46686770-46686770 on Build GRCh38
rs752186815 Chr.1: 46686881-46686881 on Build GRCh38
rs78161481 Chr.1: 46686768-46686768 on Build GRCh38
rs763769582 Chr.1: 46686904-46686904 on Build GRCh38
rs1057462119 Chr.1: 46686818-46686818 on Build GRCh38
rs372912057 Chr.1: 46686887-46686887 on Build GRCh38
rs1003045936 Chr.1: 46686788-46686788 on Build GRCh38
rs558214619 Chr.1: 46687109-46687109 on Build GRCh38
rs753438736 Chr.1: 46686765-46686765 on Build GRCh38
rs576648159 Chr.1: 46687148-46687148 on Build GRCh38
rs987353507 Chr.1: 46686986-46686986 on Build GRCh38
rs774732028 Chr.1: 46686738-46686738 on Build GRCh38
rs200625014 Chr.1: 46686806-46686806 on Build GRCh38
rs778697585 Chr.1: 46687191-46687191 on Build GRCh38
rs764926660 Chr.1: 46686885-46686885 on Build GRCh38
rs1057331052 Chr.1: 46687137-46687137 on Build GRCh38
rs781229853 Chr.1: 46686812-46686812 on Build GRCh38
rs924897063 Chr.1: 46687082-46687082 on Build GRCh38
rs367944001 Chr.1: 46686796-46686796 on Build GRCh38
rs537439516 Chr.1: 46687169-46687169 on Build GRCh38
rs549415887 Chr.1: 46686816-46686816 on Build GRCh38
rs993347854 Chr.1: 46686938-46686938 on Build GRCh38
rs539281268 Chr.1: 46687102-46687102 on Build GRCh38
rs1025080219 Chr.1: 46687174-46687174 on Build GRCh38
rs555800055 Chr.1: 46687180-46687180 on Build GRCh38
rs746009501 Chr.1: 46686813-46686813 on Build GRCh38
rs903661554 Chr.1: 46686755-46686755 on Build GRCh38
rs774879054 Chr.1: 46686841-46686841 on Build GRCh38
rs940467285 Chr.1: 46687001-46687001 on Build GRCh38
rs981405397 Chr.1: 46686895-46686895 on Build GRCh38
rs1052319060 Chr.1: 46687097-46687097 on Build GRCh38
rs931685473 Chr.1: 46687075-46687075 on Build GRCh38
rs148914051 Chr.1: 46687032-46687032 on Build GRCh38
rs375767892 Chr.1: 46686836-46686836 on Build GRCh38
rs764592017 Chr.1: 46686737-46686737 on Build GRCh38
rs907626796 Chr.1: 46686961-46686961 on Build GRCh38
rs758143073 Chr.1: 46686805-46686805 on Build GRCh38
rs1013707713 Chr.1: 46687170-46687170 on Build GRCh38
rs1050731269 Chr.1: 46687080-46687080 on Build GRCh38
rs752317575 Chr.1: 46686894-46686894 on Build GRCh38
rs758174471 Chr.1: 46686896-46686896 on Build GRCh38
rs914881428 Chr.1: 46687049-46687049 on Build GRCh38
rs905152784 Chr.1: 46687176-46687176 on Build GRCh38
rs751423089 Chr.1: 46686922-46686922 on Build GRCh38
rs776266973 Chr.1: 46686874-46686874 on Build GRCh38
rs753883079 Chr.1: 46687093-46687093 on Build GRCh38
rs60098765 Chr.1: 46686739-46686739 on Build GRCh38
rs751712807 Chr.1: 46686969-46686969 on Build GRCh38
rs371263884 Chr.1: 46686809-46686809 on Build GRCh38
rs757262165 Chr.1: 46686807-46686807 on Build GRCh38
rs780188598 Chr.1: 46687178-46687178 on Build GRCh38
rs1031469559 Chr.1: 46686833-46686833 on Build GRCh38
rs749606157 Chr.1: 46686830-46686830 on Build GRCh38
rs989652980 Chr.1: 46687047-46687047 on Build GRCh38
rs758632989 Chr.1: 46687151-46687151 on Build GRCh38
rs543870856 Chr.1: 46686954-46686954 on Build GRCh38
rs1027238937 Chr.1: 46686943-46686943 on Build GRCh38
rs945033305 Chr.1: 46687073-46687073 on Build GRCh38
rs768186418 Chr.1: 46686853-46686853 on Build GRCh38
rs3766223 Chr.1: 46686958-46686958 on Build GRCh38
rs368244329 Chr.1: 46686879-46686879 on Build GRCh38
rs763855605 Chr.1: 46686887-46686888 on Build GRCh38
rs931928176 Chr.1: 46686736-46686736 on Build GRCh38
rs766198549 Chr.1: 46687091-46687091 on Build GRCh38
rs892591343 Chr.1: 46686834-46686834 on Build GRCh38
rs760188415 Chr.1: 46686888-46686891 on Build GRCh38
rs6665021 Chr.1: 46686848-46686848 on Build GRCh38
rs531197471 Chr.1: 46686815-46686815 on Build GRCh38

View Full Product Details