Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155925798-155926287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0907
Entrez Gene ID
22889
Gene Name
KIAA0907
Gene Aliases
BLOM7
UniGene
Hs.24656
Gene Chromosome Location
Chr.1: 155913043-155934442 on Build GRCh38
Transcripts
NM_014949.3 NR_003005.1

Gene Symbol
SCARNA4
Entrez Gene ID
677771
Gene Name
small Cajal body-specific RNA 4
Gene Aliases
ACA26
UniGene
Hs.678903
Gene Chromosome Location
Chr.1: 155925958-155926086 on Build GRCh38
Transcripts
NM_014949.3 NR_003005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5729247 c.685C>T p.H229Y Chr.1: 155925840-155925840 on Build GRCh38
3979883 c.712G>T p.G238C Chr.1: 155925813-155925813 on Build GRCh38
6378022 c.697G>T p.D233Y Chr.1: 155925828-155925828 on Build GRCh38
4433974 c.712G>A p.G238S Chr.1: 155925813-155925813 on Build GRCh38
6437687 c.697G>A p.D233N Chr.1: 155925828-155925828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054319684 Chr.1: 155926251-155926251 on Build GRCh38
rs748829267 Chr.1: 155925965-155925965 on Build GRCh38
rs759911527 Chr.1: 155926136-155926136 on Build GRCh38
rs755073369 Chr.1: 155925917-155925919 on Build GRCh38
rs774545373 Chr.1: 155926135-155926135 on Build GRCh38
rs773214293 Chr.1: 155926116-155926116 on Build GRCh38
rs770578721 Chr.1: 155925866-155925866 on Build GRCh38
rs751962484 Chr.1: 155926013-155926013 on Build GRCh38
rs752322115 Chr.1: 155925955-155925955 on Build GRCh38
rs767748506 Chr.1: 155926059-155926064 on Build GRCh38
rs77914413 Chr.1: 155925868-155925868 on Build GRCh38
rs576460230 Chr.1: 155926015-155926015 on Build GRCh38
rs757015091 Chr.1: 155926050-155926050 on Build GRCh38
rs373141280 Chr.1: 155925935-155925935 on Build GRCh38
rs752083634 Chr.1: 155925950-155925950 on Build GRCh38
rs568573806 Chr.1: 155925947-155925947 on Build GRCh38
rs778674768 Chr.1: 155926075-155926075 on Build GRCh38
rs769704417 Chr.1: 155926003-155926003 on Build GRCh38
rs536466961 Chr.1: 155926004-155926004 on Build GRCh38
rs377370483 Chr.1: 155925937-155925937 on Build GRCh38
rs780054101 Chr.1: 155926090-155926090 on Build GRCh38
rs1039742872 Chr.1: 155926191-155926191 on Build GRCh38
rs377061715 Chr.1: 155925862-155925862 on Build GRCh38
rs138592971 Chr.1: 155926206-155926206 on Build GRCh38
rs960612894 Chr.1: 155926185-155926185 on Build GRCh38
rs185604567 Chr.1: 155926123-155926123 on Build GRCh38
rs777289424 Chr.1: 155925864-155925864 on Build GRCh38
rs761766548 Chr.1: 155925915-155925915 on Build GRCh38
rs755529903 Chr.1: 155925951-155925951 on Build GRCh38
rs766566504 Chr.1: 155926007-155926007 on Build GRCh38
rs112149566 Chr.1: 155926236-155926236 on Build GRCh38
rs777498008 Chr.1: 155925960-155925960 on Build GRCh38
rs755570043 Chr.1: 155925958-155925958 on Build GRCh38
rs763476331 Chr.1: 155926036-155926036 on Build GRCh38
rs550306911 Chr.1: 155925856-155925856 on Build GRCh38
rs955875710 Chr.1: 155925908-155925908 on Build GRCh38
rs746603060 Chr.1: 155926019-155926019 on Build GRCh38
rs768876734 Chr.1: 155925889-155925889 on Build GRCh38
rs75537286 Chr.1: 155925867-155925867 on Build GRCh38
rs962371261 Chr.1: 155926020-155926020 on Build GRCh38
rs753546688 Chr.1: 155926046-155926046 on Build GRCh38
rs754515385 Chr.1: 155925848-155925848 on Build GRCh38
rs747997499 Chr.1: 155926149-155926149 on Build GRCh38
rs753090402 Chr.1: 155926023-155926023 on Build GRCh38
rs751756965 Chr.1: 155925861-155925861 on Build GRCh38
rs772174070 Chr.1: 155925989-155925989 on Build GRCh38
rs772054904 Chr.1: 155926162-155926162 on Build GRCh38
rs755657311 Chr.1: 155926026-155926026 on Build GRCh38
rs771211996 Chr.1: 155926132-155926132 on Build GRCh38
rs1030957595 Chr.1: 155926017-155926017 on Build GRCh38
rs918817550 Chr.1: 155926037-155926037 on Build GRCh38
rs766690984 Chr.1: 155925944-155925944 on Build GRCh38
rs759503456 Chr.1: 155925883-155925883 on Build GRCh38
rs762009421 Chr.1: 155925998-155925998 on Build GRCh38
rs930321125 Chr.1: 155926225-155926225 on Build GRCh38
rs745633699 Chr.1: 155925973-155925973 on Build GRCh38
rs1048780345 Chr.1: 155926255-155926255 on Build GRCh38
rs758253057 Chr.1: 155926081-155926081 on Build GRCh38
rs746808572 Chr.1: 155926220-155926220 on Build GRCh38
rs372642266 Chr.1: 155926141-155926141 on Build GRCh38
rs781642222 Chr.1: 155925954-155925954 on Build GRCh38
rs371393030 Chr.1: 155926092-155926092 on Build GRCh38
rs778799054 Chr.1: 155925971-155925971 on Build GRCh38
rs770787624 Chr.1: 155925970-155925970 on Build GRCh38
rs759891403 Chr.1: 155926014-155926014 on Build GRCh38
rs774438506 Chr.1: 155925872-155925872 on Build GRCh38
rs71628698 Chr.1: 155926171-155926171 on Build GRCh38
rs775561353 Chr.1: 155925993-155925993 on Build GRCh38
rs745722008 Chr.1: 155926076-155926076 on Build GRCh38
rs189641504 Chr.1: 155926219-155926219 on Build GRCh38
rs749167938 Chr.1: 155925865-155925865 on Build GRCh38
rs956707216 Chr.1: 155926025-155926025 on Build GRCh38
rs769939661 Chr.1: 155926105-155926105 on Build GRCh38
rs910276421 Chr.1: 155926256-155926256 on Build GRCh38
rs776897195 Chr.1: 155925892-155925892 on Build GRCh38

View Full Product Details